Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84482824-84483085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPF1
Entrez Gene ID
80135
Gene Name
ribosome production factor 1 homolog
Gene Aliases
BXDC5
UniGene
Hs.481202
Gene Chromosome Location
Chr.1: 84479237-84498350 on Build GRCh38
Transcripts
NM_025065.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5693272 c.324G>T p.Q108H Chr.1: 84482953-84482953 on Build GRCh38
5728887 c.324G>A p.Q108Q Chr.1: 84482953-84482953 on Build GRCh38
5701449 c.324_325insT p.R109fs*4 Chr.1: 84482953-84482954 on Build GRCh38
6540598 c.286G>T p.A96S Chr.1: 84482915-84482915 on Build GRCh38
7156085 c.337G>A p.E113K Chr.1: 84482966-84482966 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs150021810 Chr.1: 84482970-84482970 on Build GRCh38
rs761594589 Chr.1: 84482948-84482948 on Build GRCh38
rs752996347 Chr.1: 84483001-84483001 on Build GRCh38
rs771857514 Chr.1: 84483047-84483047 on Build GRCh38
rs553966595 Chr.1: 84482917-84482917 on Build GRCh38
rs989875097 Chr.1: 84482972-84482972 on Build GRCh38
rs765670625 Chr.1: 84482999-84482999 on Build GRCh38
rs760871701 Chr.1: 84483053-84483054 on Build GRCh38
rs147475122 Chr.1: 84483058-84483058 on Build GRCh38
rs777893791 Chr.1: 84482873-84482873 on Build GRCh38
rs143563141 Chr.1: 84482993-84482993 on Build GRCh38
rs945580692 Chr.1: 84483000-84483000 on Build GRCh38
rs547250384 Chr.1: 84482961-84482961 on Build GRCh38
rs749068369 Chr.1: 84482935-84482935 on Build GRCh38
rs772962273 Chr.1: 84483052-84483052 on Build GRCh38
rs747076367 Chr.1: 84482892-84482892 on Build GRCh38
rs777912171 Chr.1: 84483014-84483014 on Build GRCh38
rs372518170 Chr.1: 84483030-84483030 on Build GRCh38
rs773987130 Chr.1: 84482943-84482943 on Build GRCh38
rs758675054 Chr.1: 84483010-84483010 on Build GRCh38
rs1050202558 Chr.1: 84483061-84483061 on Build GRCh38
rs113387621 Chr.1: 84482863-84482863 on Build GRCh38
rs778699675 Chr.1: 84483008-84483009 on Build GRCh38
rs149373797 Chr.1: 84483015-84483015 on Build GRCh38
rs775288820 Chr.1: 84483053-84483053 on Build GRCh38
rs912967964 Chr.1: 84482979-84482979 on Build GRCh38
rs576561861 Chr.1: 84482926-84482926 on Build GRCh38
rs768546399 Chr.1: 84482942-84482942 on Build GRCh38
rs772054963 Chr.1: 84483043-84483047 on Build GRCh38
rs576630002 Chr.1: 84483055-84483055 on Build GRCh38
rs376926225 Chr.1: 84483035-84483035 on Build GRCh38
rs769735979 Chr.1: 84482897-84482897 on Build GRCh38
rs760199081 Chr.1: 84482962-84482962 on Build GRCh38
rs17110157 Chr.1: 84482857-84482857 on Build GRCh38
rs534222222 Chr.1: 84483051-84483051 on Build GRCh38
rs754325676 Chr.1: 84482987-84482987 on Build GRCh38
rs767267951 Chr.1: 84482949-84482949 on Build GRCh38
rs747841593 Chr.1: 84483037-84483037 on Build GRCh38
rs369492407 Chr.1: 84482864-84482864 on Build GRCh38
rs1026425331 Chr.1: 84482940-84482940 on Build GRCh38
rs201962769 Chr.1: 84483027-84483027 on Build GRCh38
rs867229500 Chr.1: 84482929-84482929 on Build GRCh38
rs774262001 Chr.1: 84483036-84483036 on Build GRCh38
rs757287507 Chr.1: 84483019-84483019 on Build GRCh38
rs771251607 Chr.1: 84483011-84483015 on Build GRCh38

View Full Product Details