Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 112703667-112703906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHOC
Entrez Gene ID
389
Gene Name
ras homolog family member C
Gene Aliases
ARH9, ARHC, H9, RHOH9
UniGene
Hs.502659
Gene Chromosome Location
Chr.1: 112701127-112707403 on Build GRCh38
Transcripts
NM_001042678.1 NM_001042679.1 NM_175744.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5014137 c.65T>A p.L22H Chr.1: 112703735-112703735 on Build GRCh38
3385035 c.132G>A p.A44A Chr.1: 112703668-112703668 on Build GRCh38
7256701 c.69C>T p.I23I Chr.1: 112703731-112703731 on Build GRCh38
5037638 c.103G>A p.V35I Chr.1: 112703697-112703697 on Build GRCh38
6507631 c.93G>T p.P31P Chr.1: 112703707-112703707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774806474 Chr.1: 112703753-112703753 on Build GRCh38
rs370372319 Chr.1: 112703821-112703821 on Build GRCh38
rs554766644 Chr.1: 112703860-112703860 on Build GRCh38
rs202097009 Chr.1: 112703824-112703824 on Build GRCh38
rs774557812 Chr.1: 112703802-112703802 on Build GRCh38
rs746283016 Chr.1: 112703770-112703770 on Build GRCh38
rs778278882 Chr.1: 112703843-112703843 on Build GRCh38
rs202032648 Chr.1: 112703809-112703809 on Build GRCh38
rs370374549 Chr.1: 112703698-112703698 on Build GRCh38
rs868187294 Chr.1: 112703802-112703803 on Build GRCh38
rs767496260 Chr.1: 112703807-112703807 on Build GRCh38
rs189082614 Chr.1: 112703731-112703731 on Build GRCh38
rs780800937 Chr.1: 112703849-112703849 on Build GRCh38
rs754441878 Chr.1: 112703833-112703833 on Build GRCh38
rs1017891315 Chr.1: 112703866-112703866 on Build GRCh38
rs980513702 Chr.1: 112703868-112703868 on Build GRCh38
rs775884207 Chr.1: 112703789-112703789 on Build GRCh38
rs549460700 Chr.1: 112703790-112703790 on Build GRCh38
rs2306937 Chr.1: 112703884-112703884 on Build GRCh38
rs751438534 Chr.1: 112703787-112703787 on Build GRCh38
rs1014495631 Chr.1: 112703822-112703822 on Build GRCh38
rs372772606 Chr.1: 112703707-112703707 on Build GRCh38
rs764087172 Chr.1: 112703791-112703791 on Build GRCh38
rs532814642 Chr.1: 112703741-112703741 on Build GRCh38
rs754465217 Chr.1: 112703733-112703733 on Build GRCh38
rs779724269 Chr.1: 112703708-112703708 on Build GRCh38
rs771531775 Chr.1: 112703764-112703764 on Build GRCh38
rs896061382 Chr.1: 112703818-112703818 on Build GRCh38
rs761643994 Chr.1: 112703804-112703804 on Build GRCh38
rs757010375 Chr.1: 112703695-112703695 on Build GRCh38
rs777122070 Chr.1: 112703769-112703769 on Build GRCh38
rs763777776 Chr.1: 112703769-112703773 on Build GRCh38
rs755937044 Chr.1: 112703812-112703812 on Build GRCh38
rs778283490 Chr.1: 112703736-112703736 on Build GRCh38
rs1804292 Chr.1: 112703779-112703779 on Build GRCh38

View Full Product Details