Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 181053484-181053757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MR1
Entrez Gene ID
3140
Gene Name
major histocompatibility complex, class I-related
Gene Aliases
HLALS
UniGene
Hs.13500
Gene Chromosome Location
Chr.1: 181033379-181061938 on Build GRCh38
Transcripts
NM_001194999.1 NM_001195000.1 NM_001195035.1 NM_001310213.1 NM_001531.2 XM_006711286.3 XM_011509463.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4563067 c.955G>A p.G319S Chr.1: 181053647-181053647 on Build GRCh38
5005016 c.983G>A p.R328Q Chr.1: 181053675-181053675 on Build GRCh38
358285 c.983G>T p.R328L Chr.1: 181053675-181053675 on Build GRCh38
3479495 c.969G>A p.W323* Chr.1: 181053661-181053661 on Build GRCh38
900508 c.888A>G p.E296E Chr.1: 181053580-181053580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757492621 Chr.1: 181053528-181053528 on Build GRCh38
rs1053016417 Chr.1: 181053624-181053624 on Build GRCh38
rs777324974 Chr.1: 181053675-181053675 on Build GRCh38
rs367841673 Chr.1: 181053723-181053723 on Build GRCh38
rs775783960 Chr.1: 181053720-181053720 on Build GRCh38
rs138847795 Chr.1: 181053722-181053722 on Build GRCh38
rs554565768 Chr.1: 181053545-181053545 on Build GRCh38
rs778446144 Chr.1: 181053587-181053587 on Build GRCh38
rs35441386 Chr.1: 181053610-181053611 on Build GRCh38
rs1010892481 Chr.1: 181053674-181053674 on Build GRCh38
rs367826683 Chr.1: 181053713-181053713 on Build GRCh38
rs746835170 Chr.1: 181053604-181053604 on Build GRCh38
rs377640897 Chr.1: 181053593-181053593 on Build GRCh38
rs750766088 Chr.1: 181053661-181053661 on Build GRCh38
rs767816725 Chr.1: 181053651-181053651 on Build GRCh38
rs751981491 Chr.1: 181053526-181053526 on Build GRCh38
rs891084515 Chr.1: 181053609-181053609 on Build GRCh38
rs762380947 Chr.1: 181053642-181053642 on Build GRCh38
rs112996862 Chr.1: 181053634-181053634 on Build GRCh38
rs374617057 Chr.1: 181053706-181053706 on Build GRCh38
rs762277892 Chr.1: 181053728-181053728 on Build GRCh38
rs200587984 Chr.1: 181053582-181053582 on Build GRCh38
rs149165214 Chr.1: 181053619-181053619 on Build GRCh38
rs747610255 Chr.1: 181053589-181053589 on Build GRCh38
rs777505440 Chr.1: 181053597-181053597 on Build GRCh38
rs746985326 Chr.1: 181053682-181053682 on Build GRCh38
rs763084873 Chr.1: 181053632-181053632 on Build GRCh38
rs757845823 Chr.1: 181053671-181053671 on Build GRCh38
rs372194193 Chr.1: 181053727-181053727 on Build GRCh38
rs753505165 Chr.1: 181053566-181053566 on Build GRCh38
rs745607140 Chr.1: 181053707-181053707 on Build GRCh38
rs768827297 Chr.1: 181053511-181053511 on Build GRCh38
rs764830276 Chr.1: 181053670-181053670 on Build GRCh38
rs34434385 Chr.1: 181053544-181053545 on Build GRCh38
rs757186740 Chr.1: 181053689-181053689 on Build GRCh38
rs115981006 Chr.1: 181053714-181053714 on Build GRCh38
rs754650505 Chr.1: 181053669-181053669 on Build GRCh38
rs770689113 Chr.1: 181053605-181053605 on Build GRCh38
rs776481265 Chr.1: 181053610-181053610 on Build GRCh38
rs775730900 Chr.1: 181053622-181053622 on Build GRCh38
rs778902974 Chr.1: 181053711-181053711 on Build GRCh38
rs746131097 Chr.1: 181053613-181053613 on Build GRCh38
rs754597231 Chr.1: 181053571-181053571 on Build GRCh38
rs930849648 Chr.1: 181053702-181053702 on Build GRCh38
rs900895202 Chr.1: 181053724-181053724 on Build GRCh38
rs774658856 Chr.1: 181053635-181053635 on Build GRCh38

View Full Product Details