Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33582040-33582279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DONSON
Entrez Gene ID
29980
Gene Name
downstream neighbor of SON
Gene Aliases
B17, C21orf60
UniGene
Hs.436341
Gene Chromosome Location
Chr.21: 33577551-33588708 on Build GRCh38
Transcripts
NM_017613.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3423927 c.998G>C p.S333T Chr.21: 33582213-33582213 on Build GRCh38
7118758 c.979C>G p.L327V Chr.21: 33582232-33582232 on Build GRCh38
4101382 c.1047-2A>G p.? Chr.21: 33582057-33582057 on Build GRCh38
3841818 c.994G>T p.E332* Chr.21: 33582217-33582217 on Build GRCh38
1030391 c.1014G>A p.E338E Chr.21: 33582197-33582197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887928187 Chr.21: 33582168-33582168 on Build GRCh38
rs368570176 Chr.21: 33582179-33582179 on Build GRCh38
rs917943361 Chr.21: 33582182-33582182 on Build GRCh38
rs185033915 Chr.21: 33582102-33582102 on Build GRCh38
rs766283641 Chr.21: 33582092-33582092 on Build GRCh38
rs745561864 Chr.21: 33582069-33582070 on Build GRCh38
rs752565840 Chr.21: 33582207-33582207 on Build GRCh38
rs188284536 Chr.21: 33582139-33582139 on Build GRCh38
rs199989677 Chr.21: 33582212-33582212 on Build GRCh38
rs774312830 Chr.21: 33582099-33582099 on Build GRCh38
rs753371688 Chr.21: 33582134-33582134 on Build GRCh38
rs775130920 Chr.21: 33582222-33582222 on Build GRCh38
rs759408217 Chr.21: 33582217-33582217 on Build GRCh38
rs371275940 Chr.21: 33582144-33582144 on Build GRCh38
rs367663372 Chr.21: 33582133-33582133 on Build GRCh38
rs146461848 Chr.21: 33582180-33582180 on Build GRCh38
rs369488404 Chr.21: 33582196-33582196 on Build GRCh38
rs375424429 Chr.21: 33582105-33582105 on Build GRCh38
rs548866717 Chr.21: 33582221-33582221 on Build GRCh38
rs755688201 Chr.21: 33582200-33582202 on Build GRCh38
rs375857436 Chr.21: 33582161-33582161 on Build GRCh38
rs780667282 Chr.21: 33582141-33582141 on Build GRCh38
rs755310952 Chr.21: 33582157-33582157 on Build GRCh38
rs755714861 Chr.21: 33582125-33582125 on Build GRCh38
rs763079987 Chr.21: 33582090-33582090 on Build GRCh38
rs769843611 Chr.21: 33582059-33582059 on Build GRCh38
rs779149681 Chr.21: 33582064-33582064 on Build GRCh38

View Full Product Details