Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40141551-40141780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC3
Entrez Gene ID
22794
Gene Name
cancer susceptibility candidate 3
Gene Aliases
BTZ, MLN51
UniGene
Hs.743287
Gene Chromosome Location
Chr.17: 40140254-40172178 on Build GRCh38
Transcripts
NM_007359.4 XM_005257163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4957623 c.282T>C p.S94S Chr.17: 40141592-40141592 on Build GRCh38
6146646 c.270G>T p.S90S Chr.17: 40141580-40141580 on Build GRCh38
5495937 c.277G>A p.E93K Chr.17: 40141587-40141587 on Build GRCh38
2838718 c.294G>A p.S98S Chr.17: 40141604-40141604 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775395017 Chr.17: 40141628-40141628 on Build GRCh38
rs765673382 Chr.17: 40141600-40141600 on Build GRCh38
rs532687270 Chr.17: 40141606-40141606 on Build GRCh38
rs75957101 Chr.17: 40141732-40141732 on Build GRCh38
rs1045577068 Chr.17: 40141709-40141709 on Build GRCh38
rs571213652 Chr.17: 40141724-40141724 on Build GRCh38
rs927145284 Chr.17: 40141717-40141717 on Build GRCh38
rs150571760 Chr.17: 40141672-40141674 on Build GRCh38
rs750200255 Chr.17: 40141634-40141634 on Build GRCh38
rs774910900 Chr.17: 40141603-40141603 on Build GRCh38
rs756942193 Chr.17: 40141630-40141630 on Build GRCh38
rs7212098 Chr.17: 40141684-40141684 on Build GRCh38
rs974939084 Chr.17: 40141580-40141580 on Build GRCh38
rs765689165 Chr.17: 40141720-40141720 on Build GRCh38
rs907167707 Chr.17: 40141742-40141742 on Build GRCh38
rs751191341 Chr.17: 40141620-40141620 on Build GRCh38
rs763687038 Chr.17: 40141618-40141618 on Build GRCh38
rs951765324 Chr.17: 40141631-40141631 on Build GRCh38
rs1004130990 Chr.17: 40141702-40141702 on Build GRCh38
rs756013215 Chr.17: 40141636-40141636 on Build GRCh38
rs899684038 Chr.17: 40141598-40141598 on Build GRCh38
rs949969601 Chr.17: 40141688-40141688 on Build GRCh38
rs567328554 Chr.17: 40141676-40141676 on Build GRCh38
rs549557458 Chr.17: 40141694-40141694 on Build GRCh38
rs931106009 Chr.17: 40141621-40141621 on Build GRCh38
rs767338248 Chr.17: 40141632-40141632 on Build GRCh38

View Full Product Details