Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31094668-31094897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VKORC1
Entrez Gene ID
79001
Gene Name
vitamin K epoxide reductase complex subunit 1
Gene Aliases
EDTP308, MST134, MST576, VKCFD2, VKOR
UniGene
Hs.324844
Gene Chromosome Location
Chr.16: 31090842-31094999 on Build GRCh38
Transcripts
NM_001311311.1 NM_024006.5 NM_206824.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5439291 c.54G>T p.T18T Chr.16: 31094676-31094676 on Build GRCh38
1178984 c.54G>C p.T18T Chr.16: 31094676-31094676 on Build GRCh38
3712022 c.57C>T p.G19G Chr.16: 31094673-31094673 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766721996 Chr.16: 31094701-31094701 on Build GRCh38
rs1028424472 Chr.16: 31094704-31094704 on Build GRCh38
rs886042699 Chr.16: 31094727-31094727 on Build GRCh38
rs1008191003 Chr.16: 31094715-31094715 on Build GRCh38
rs752892359 Chr.16: 31094716-31094716 on Build GRCh38
rs935685325 Chr.16: 31094817-31094817 on Build GRCh38
rs1004095378 Chr.16: 31094871-31094871 on Build GRCh38
rs768158149 Chr.16: 31094756-31094756 on Build GRCh38
rs1017284342 Chr.16: 31094877-31094877 on Build GRCh38
rs772018953 Chr.16: 31094747-31094747 on Build GRCh38
rs778819991 Chr.16: 31094728-31094728 on Build GRCh38
rs745707367 Chr.16: 31094731-31094731 on Build GRCh38
rs988276085 Chr.16: 31094861-31094861 on Build GRCh38
rs539401220 Chr.16: 31094797-31094797 on Build GRCh38
rs755166588 Chr.16: 31094710-31094710 on Build GRCh38
rs979466695 Chr.16: 31094784-31094784 on Build GRCh38
rs914136023 Chr.16: 31094744-31094744 on Build GRCh38
rs949923135 Chr.16: 31094862-31094862 on Build GRCh38
rs1055439614 Chr.16: 31094821-31094821 on Build GRCh38
rs200328478 Chr.16: 31094708-31094708 on Build GRCh38
rs925581293 Chr.16: 31094810-31094810 on Build GRCh38
rs1045605206 Chr.16: 31094863-31094863 on Build GRCh38
rs750454948 Chr.16: 31094867-31094867 on Build GRCh38
rs55894764 Chr.16: 31094694-31094694 on Build GRCh38
rs547713296 Chr.16: 31094699-31094699 on Build GRCh38
rs749237507 Chr.16: 31094720-31094720 on Build GRCh38
rs993371943 Chr.16: 31094690-31094690 on Build GRCh38
rs925409684 Chr.16: 31094801-31094801 on Build GRCh38
rs746650028 Chr.16: 31094755-31094755 on Build GRCh38
rs894254929 Chr.16: 31094826-31094826 on Build GRCh38
rs766916065 Chr.16: 31094761-31094761 on Build GRCh38
rs763482320 Chr.16: 31094758-31094758 on Build GRCh38
rs757332613 Chr.16: 31094725-31094725 on Build GRCh38
rs763425395 Chr.16: 31094700-31094700 on Build GRCh38
rs775171450 Chr.16: 31094748-31094748 on Build GRCh38
rs767508384 Chr.16: 31094712-31094712 on Build GRCh38
rs987009894 Chr.16: 31094743-31094743 on Build GRCh38
rs774860201 Chr.16: 31094766-31094766 on Build GRCh38
rs557787762 Chr.16: 31094864-31094864 on Build GRCh38
rs756150795 Chr.16: 31094719-31094719 on Build GRCh38
rs759852599 Chr.16: 31094774-31094774 on Build GRCh38

View Full Product Details