Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59114459-59114716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB2
Entrez Gene ID
9133
Gene Name
cyclin B2
Gene Aliases
HsT17299
UniGene
Hs.194698
Gene Chromosome Location
Chr.15: 59105085-59125045 on Build GRCh38
Transcripts
NM_004701.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3502796 c.336C>T p.A112A Chr.15: 59114512-59114512 on Build GRCh38
352561 c.316T>G p.C106G Chr.15: 59114492-59114492 on Build GRCh38
7447940 c.362A>G p.D121G Chr.15: 59114538-59114538 on Build GRCh38
7180672 c.426C>T p.L142L Chr.15: 59114602-59114602 on Build GRCh38
2219692 c.318C>T p.C106C Chr.15: 59114494-59114494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779994699 Chr.15: 59114546-59114546 on Build GRCh38
rs772568414 Chr.15: 59114666-59114666 on Build GRCh38
rs975888979 Chr.15: 59114664-59114664 on Build GRCh38
rs747420379 Chr.15: 59114685-59114685 on Build GRCh38
rs764812051 Chr.15: 59114616-59114616 on Build GRCh38
rs887489295 Chr.15: 59114621-59114621 on Build GRCh38
rs550919161 Chr.15: 59114486-59114486 on Build GRCh38
rs774800920 Chr.15: 59114481-59114481 on Build GRCh38
rs371430210 Chr.15: 59114648-59114648 on Build GRCh38
rs778079480 Chr.15: 59114638-59114638 on Build GRCh38
rs534109952 Chr.15: 59114586-59114586 on Build GRCh38
rs775189310 Chr.15: 59114494-59114494 on Build GRCh38
rs1007712455 Chr.15: 59114624-59114624 on Build GRCh38
rs763591966 Chr.15: 59114507-59114507 on Build GRCh38
rs201147134 Chr.15: 59114528-59114528 on Build GRCh38
rs146057559 Chr.15: 59114578-59114578 on Build GRCh38
rs757393271 Chr.15: 59114650-59114650 on Build GRCh38
rs775841822 Chr.15: 59114595-59114595 on Build GRCh38
rs554757385 Chr.15: 59114548-59114548 on Build GRCh38
rs756679957 Chr.15: 59114527-59114527 on Build GRCh38
rs750456709 Chr.15: 59114617-59114617 on Build GRCh38
rs758400651 Chr.15: 59114618-59114618 on Build GRCh38
rs896571394 Chr.15: 59114547-59114547 on Build GRCh38
rs772482349 Chr.15: 59114594-59114594 on Build GRCh38
rs760877193 Chr.15: 59114608-59114608 on Build GRCh38
rs778950981 Chr.15: 59114660-59114660 on Build GRCh38
rs767834376 Chr.15: 59114493-59114493 on Build GRCh38
rs144226046 Chr.15: 59114511-59114511 on Build GRCh38
rs763836666 Chr.15: 59114609-59114609 on Build GRCh38
rs759942264 Chr.15: 59114483-59114483 on Build GRCh38
rs776096009 Chr.15: 59114610-59114610 on Build GRCh38
rs756373517 Chr.15: 59114557-59114557 on Build GRCh38
rs748017216 Chr.15: 59114687-59114687 on Build GRCh38
rs761438863 Chr.15: 59114614-59114614 on Build GRCh38
rs201017479 Chr.15: 59114628-59114628 on Build GRCh38
rs536661238 Chr.15: 59114537-59114537 on Build GRCh38
rs140205864 Chr.15: 59114592-59114592 on Build GRCh38
rs749080700 Chr.15: 59114485-59114485 on Build GRCh38
rs760361771 Chr.15: 59114499-59114499 on Build GRCh38
rs865848383 Chr.15: 59114503-59114503 on Build GRCh38
rs185419194 Chr.15: 59114663-59114663 on Build GRCh38
rs754691646 Chr.15: 59114629-59114629 on Build GRCh38
rs775816244 Chr.15: 59114683-59114683 on Build GRCh38
rs758310424 Chr.15: 59114542-59114542 on Build GRCh38
rs2306785 Chr.15: 59114579-59114579 on Build GRCh38
rs766362424 Chr.15: 59114619-59114619 on Build GRCh38

View Full Product Details