Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 177550794-177551019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC30B
Entrez Gene ID
150737
Gene Name
tetratricopeptide repeat domain 30B
Gene Aliases
IFT70, IFT70B, fleer
UniGene
Hs.447659
Gene Chromosome Location
Chr.2: 177550152-177552796 on Build GRCh38
Transcripts
NM_152517.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5994326 c.1949C>A p.S650Y Chr.2: 177550815-177550815 on Build GRCh38
717616 c.1793C>G p.S598* Chr.2: 177550971-177550971 on Build GRCh38
4737165 c.1902G>C p.L634L Chr.2: 177550862-177550862 on Build GRCh38
7395590 c.1780T>A p.L594I Chr.2: 177550984-177550984 on Build GRCh38
1009944 c.1929G>T p.K643N Chr.2: 177550835-177550835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745309315 Chr.2: 177550853-177550853 on Build GRCh38
rs756353740 Chr.2: 177550821-177550821 on Build GRCh38
rs760084480 Chr.2: 177550964-177550964 on Build GRCh38
rs3204099 Chr.2: 177550953-177550953 on Build GRCh38
rs373148288 Chr.2: 177550860-177550860 on Build GRCh38
rs1052456004 Chr.2: 177550971-177550971 on Build GRCh38
rs771411917 Chr.2: 177550951-177550951 on Build GRCh38
rs753627193 Chr.2: 177550901-177550901 on Build GRCh38
rs764242707 Chr.2: 177550898-177550898 on Build GRCh38
rs774927003 Chr.2: 177550962-177550962 on Build GRCh38
rs758396328 Chr.2: 177550912-177550912 on Build GRCh38
rs746523690 Chr.2: 177550943-177550943 on Build GRCh38
rs770851035 Chr.2: 177550836-177550836 on Build GRCh38
rs974380195 Chr.2: 177550913-177550913 on Build GRCh38
rs760182509 Chr.2: 177550885-177550885 on Build GRCh38
rs768315810 Chr.2: 177550949-177550949 on Build GRCh38
rs775014951 Chr.2: 177550875-177550875 on Build GRCh38
rs756668891 Chr.2: 177550900-177550901 on Build GRCh38
rs752860873 Chr.2: 177550888-177550888 on Build GRCh38
rs776155289 Chr.2: 177550950-177550950 on Build GRCh38
rs200300646 Chr.2: 177550903-177550903 on Build GRCh38
rs767705451 Chr.2: 177550887-177550887 on Build GRCh38
rs185099655 Chr.2: 177550838-177550838 on Build GRCh38
rs756334589 Chr.2: 177550894-177550894 on Build GRCh38
rs773578827 Chr.2: 177550867-177550867 on Build GRCh38
rs866276543 Chr.2: 177550939-177550940 on Build GRCh38
rs117066669 Chr.2: 177550966-177550966 on Build GRCh38
rs749083490 Chr.2: 177550835-177550835 on Build GRCh38
rs745791642 Chr.2: 177550909-177550909 on Build GRCh38
rs766845463 Chr.2: 177550874-177550874 on Build GRCh38
rs190055750 Chr.2: 177550864-177550864 on Build GRCh38
rs369930014 Chr.2: 177550830-177550830 on Build GRCh38
rs376491794 Chr.2: 177550843-177550843 on Build GRCh38
rs200420269 Chr.2: 177550905-177550905 on Build GRCh38
rs779517318 Chr.2: 177550930-177550930 on Build GRCh38
rs778260199 Chr.2: 177550906-177550907 on Build GRCh38
rs760289832 Chr.2: 177550865-177550865 on Build GRCh38

View Full Product Details