Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145765410-145765647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf51
Entrez Gene ID
646603
Gene Name
chromosome 4 open reading frame 51
Gene Aliases
-
UniGene
Hs.452865
Gene Chromosome Location
Chr.4: 145680103-145792248 on Build GRCh38
Transcripts
XM_017008547.1 XM_017008548.1 XM_017008549.1 XM_017008550.1 XM_017008551.1 XM_017008552.1 XM_017008553.1 XM_017008554.1 XM_017008555.1 XM_017008556.1 NM_001306215.1 NM_178835.4 XM_011531634.2 XM_011531635.2 XM_011531645.2 XM_017007768.1 XM_017007769.1 XM_017007770.1 XM_017007771.1 XM_017007772.1 XM_017007773.1 XM_017007774.1 XM_017007775.1 XM_017007777.1

Gene Symbol
ZNF827
Entrez Gene ID
152485
Gene Name
zinc finger protein 827
Gene Aliases
-
UniGene
Hs.133916
Gene Chromosome Location
Chr.4: 145757627-145939008 on Build GRCh38
Transcripts
XM_017008547.1 XM_017008548.1 XM_017008549.1 XM_017008550.1 XM_017008551.1 XM_017008552.1 XM_017008553.1 XM_017008554.1 XM_017008555.1 XM_017008556.1 NM_001306215.1 NM_178835.4 XM_011531634.2 XM_011531635.2 XM_011531645.2 XM_017007768.1 XM_017007769.1 XM_017007770.1 XM_017007771.1 XM_017007772.1 XM_017007773.1 XM_017007774.1 XM_017007775.1 XM_017007777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4891519 c.3052G>T p.G1018W Chr.4: 145765547-145765547 on Build GRCh38
1051773 c.3015C>T p.S1005S Chr.4: 145765584-145765584 on Build GRCh38
233845 c.2959T>G p.S987A Chr.4: 145765640-145765640 on Build GRCh38
7107385 c.3010G>C p.G1004R Chr.4: 145765589-145765589 on Build GRCh38
732477 c.2953G>C p.D985H Chr.4: 145765646-145765646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375267951 Chr.4: 145765510-145765510 on Build GRCh38
rs761834793 Chr.4: 145765551-145765551 on Build GRCh38
rs776654285 Chr.4: 145765539-145765539 on Build GRCh38
rs781521090 Chr.4: 145765592-145765592 on Build GRCh38
rs768721535 Chr.4: 145765532-145765532 on Build GRCh38
rs902344807 Chr.4: 145765465-145765465 on Build GRCh38
rs767784968 Chr.4: 145765506-145765507 on Build GRCh38
rs762685378 Chr.4: 145765560-145765560 on Build GRCh38
rs4327505 Chr.4: 145765431-145765431 on Build GRCh38
rs4348116 Chr.4: 145765438-145765438 on Build GRCh38
rs755276818 Chr.4: 145765574-145765574 on Build GRCh38
rs536222642 Chr.4: 145765494-145765494 on Build GRCh38
rs555437405 Chr.4: 145765496-145765496 on Build GRCh38
rs751957056 Chr.4: 145765567-145765567 on Build GRCh38
rs1042661837 Chr.4: 145765464-145765464 on Build GRCh38
rs780092220 Chr.4: 145765474-145765474 on Build GRCh38
rs756583284 Chr.4: 145765602-145765602 on Build GRCh38
rs748238049 Chr.4: 145765509-145765509 on Build GRCh38
rs900571394 Chr.4: 145765493-145765493 on Build GRCh38
rs112442492 Chr.4: 145765586-145765586 on Build GRCh38
rs569928074 Chr.4: 145765597-145765597 on Build GRCh38
rs768281100 Chr.4: 145765623-145765623 on Build GRCh38
rs949322676 Chr.4: 145765450-145765450 on Build GRCh38
rs781610994 Chr.4: 145765508-145765508 on Build GRCh38
rs372229936 Chr.4: 145765557-145765557 on Build GRCh38
rs769684778 Chr.4: 145765473-145765473 on Build GRCh38
rs749565486 Chr.4: 145765511-145765511 on Build GRCh38
rs777958792 Chr.4: 145765617-145765617 on Build GRCh38
rs765908191 Chr.4: 145765564-145765564 on Build GRCh38
rs771036713 Chr.4: 145765512-145765512 on Build GRCh38
rs771220958 Chr.4: 145765622-145765622 on Build GRCh38
rs760350682 Chr.4: 145765524-145765524 on Build GRCh38
rs112917849 Chr.4: 145765526-145765526 on Build GRCh38
rs764877432 Chr.4: 145765556-145765556 on Build GRCh38
rs115676530 Chr.4: 145765487-145765487 on Build GRCh38
rs931233933 Chr.4: 145765504-145765504 on Build GRCh38
rs199624953 Chr.4: 145765535-145765535 on Build GRCh38
rs368271131 Chr.4: 145765515-145765515 on Build GRCh38
rs1046269280 Chr.4: 145765513-145765513 on Build GRCh38
rs755509501 Chr.4: 145765500-145765500 on Build GRCh38
rs751820757 Chr.4: 145765498-145765498 on Build GRCh38

View Full Product Details