Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34241531-34241776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBAP1
Entrez Gene ID
51271
Gene Name
ubiquitin associated protein 1
Gene Aliases
NAG20, UAP, UBAP, UBAP-1
UniGene
Hs.268963
Gene Chromosome Location
Chr.9: 34179005-34252523 on Build GRCh38
Transcripts
NM_001171201.1 NM_001171202.1 NM_001171203.2 NM_001171204.2 NM_016525.4 NR_033243.2 XM_006716779.3 XM_011517898.2 XM_011517899.1 XM_011517900.1 XM_017014801.1 XM_017014802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1497069 c.838G>A p.A280T Chr.9: 34241671-34241671 on Build GRCh38
6550054 c.942C>T p.S314S Chr.9: 34241775-34241775 on Build GRCh38
7159286 c.562A>G p.T188A Chr.9: 34241587-34241587 on Build GRCh38
2773018 c.606G>C p.R202S Chr.9: 34241631-34241631 on Build GRCh38
6536753 c.877C>G p.H293D Chr.9: 34241710-34241710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778588610 Chr.9: 34241677-34241677 on Build GRCh38
rs1047764232 Chr.9: 34241580-34241580 on Build GRCh38
rs770258434 Chr.9: 34241711-34241711 on Build GRCh38
rs900649680 Chr.9: 34241673-34241673 on Build GRCh38
rs755986740 Chr.9: 34241641-34241641 on Build GRCh38
rs753594887 Chr.9: 34241645-34241645 on Build GRCh38
rs201497858 Chr.9: 34241692-34241692 on Build GRCh38
rs370224733 Chr.9: 34241671-34241671 on Build GRCh38
rs530143231 Chr.9: 34241643-34241643 on Build GRCh38
rs777537431 Chr.9: 34241579-34241579 on Build GRCh38
rs140180675 Chr.9: 34241618-34241618 on Build GRCh38
rs201070690 Chr.9: 34241732-34241732 on Build GRCh38
rs763253925 Chr.9: 34241741-34241741 on Build GRCh38
rs574197954 Chr.9: 34241702-34241702 on Build GRCh38
rs896581712 Chr.9: 34241574-34241574 on Build GRCh38
rs137993779 Chr.9: 34241683-34241683 on Build GRCh38
rs888018595 Chr.9: 34241593-34241593 on Build GRCh38
rs150314272 Chr.9: 34241623-34241623 on Build GRCh38
rs368507896 Chr.9: 34241684-34241684 on Build GRCh38
rs754526509 Chr.9: 34241664-34241664 on Build GRCh38
rs750329588 Chr.9: 34241628-34241628 on Build GRCh38

View Full Product Details