Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74336587-74336858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGM2L1
Entrez Gene ID
283209
Gene Name
phosphoglucomutase 2 like 1
Gene Aliases
BM32A, PMMLP
UniGene
Hs.26612
Gene Chromosome Location
Chr.11: 74330312-74398465 on Build GRCh38
Transcripts
NM_173582.4 XM_011544953.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4944430 c.1859G>A p.R620H Chr.11: 74336662-74336662 on Build GRCh38
1188350 c.1768G>C p.D590H Chr.11: 74336753-74336753 on Build GRCh38
5793249 c.1828C>G p.L610V Chr.11: 74336693-74336693 on Build GRCh38
7180295 c.1819G>C p.E607Q Chr.11: 74336702-74336702 on Build GRCh38
6356394 c.1858C>T p.R620C Chr.11: 74336663-74336663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569490571 Chr.11: 74336691-74336691 on Build GRCh38
rs773687874 Chr.11: 74336664-74336664 on Build GRCh38
rs755187377 Chr.11: 74336785-74336785 on Build GRCh38
rs775158056 Chr.11: 74336737-74336737 on Build GRCh38
rs139050358 Chr.11: 74336824-74336825 on Build GRCh38
rs1030701582 Chr.11: 74336659-74336659 on Build GRCh38
rs994450836 Chr.11: 74336771-74336771 on Build GRCh38
rs368706989 Chr.11: 74336662-74336662 on Build GRCh38
rs764164267 Chr.11: 74336746-74336746 on Build GRCh38
rs772590650 Chr.11: 74336663-74336663 on Build GRCh38
rs372232892 Chr.11: 74336693-74336693 on Build GRCh38
rs757845172 Chr.11: 74336650-74336650 on Build GRCh38
rs749455842 Chr.11: 74336644-74336644 on Build GRCh38
rs759127837 Chr.11: 74336716-74336716 on Build GRCh38
rs985153468 Chr.11: 74336830-74336830 on Build GRCh38
rs371691460 Chr.11: 74336685-74336685 on Build GRCh38
rs749126739 Chr.11: 74336802-74336802 on Build GRCh38
rs1044782 Chr.11: 74336638-74336638 on Build GRCh38
rs140061354 Chr.11: 74336654-74336654 on Build GRCh38
rs36014178 Chr.11: 74336698-74336698 on Build GRCh38
rs374676133 Chr.11: 74336626-74336626 on Build GRCh38
rs927513437 Chr.11: 74336812-74336812 on Build GRCh38
rs185001330 Chr.11: 74336615-74336615 on Build GRCh38
rs767228393 Chr.11: 74336759-74336759 on Build GRCh38
rs751446447 Chr.11: 74336634-74336634 on Build GRCh38
rs756968407 Chr.11: 74336637-74336637 on Build GRCh38
rs376585328 Chr.11: 74336628-74336628 on Build GRCh38
rs762525694 Chr.11: 74336740-74336740 on Build GRCh38
rs768863647 Chr.11: 74336653-74336653 on Build GRCh38
rs750165130 Chr.11: 74336773-74336773 on Build GRCh38
rs200256315 Chr.11: 74336757-74336757 on Build GRCh38
rs948088318 Chr.11: 74336641-74336641 on Build GRCh38
rs752200786 Chr.11: 74336617-74336617 on Build GRCh38
rs989272857 Chr.11: 74336696-74336696 on Build GRCh38
rs764880897 Chr.11: 74336726-74336726 on Build GRCh38

View Full Product Details