Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 85089151-85089398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE8A
Entrez Gene ID
5151
Gene Name
phosphodiesterase 8A
Gene Aliases
HsT19550
UniGene
Hs.9333
Gene Chromosome Location
Chr.15: 84980513-85139145 on Build GRCh38
Transcripts
NM_001243137.1 NM_002605.2 NM_173454.1 XM_017022308.1 XM_017022309.1 XM_017022310.1 XM_017022311.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5384009 c.654C>T p.F218F Chr.15: 85089356-85089356 on Build GRCh38
4982708 c.647C>G p.S216* Chr.15: 85089349-85089349 on Build GRCh38
7383384 c.685G>T p.E229* Chr.15: 85089387-85089387 on Build GRCh38
7208883 c.640T>C p.C214R Chr.15: 85089342-85089342 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs951725417 Chr.15: 85089192-85089192 on Build GRCh38
rs768639137 Chr.15: 85089293-85089293 on Build GRCh38
rs762404521 Chr.15: 85089316-85089317 on Build GRCh38
rs368348788 Chr.15: 85089358-85089358 on Build GRCh38
rs917779470 Chr.15: 85089196-85089196 on Build GRCh38
rs765957522 Chr.15: 85089366-85089367 on Build GRCh38
rs907491830 Chr.15: 85089247-85089247 on Build GRCh38
rs567783245 Chr.15: 85089334-85089334 on Build GRCh38
rs527704035 Chr.15: 85089179-85089179 on Build GRCh38
rs779009148 Chr.15: 85089319-85089319 on Build GRCh38
rs547455133 Chr.15: 85089231-85089231 on Build GRCh38
rs923818298 Chr.15: 85089283-85089283 on Build GRCh38
rs769774840 Chr.15: 85089331-85089331 on Build GRCh38
rs772223713 Chr.15: 85089329-85089329 on Build GRCh38
rs541233505 Chr.15: 85089282-85089282 on Build GRCh38
rs970942000 Chr.15: 85089236-85089236 on Build GRCh38
rs775313361 Chr.15: 85089339-85089339 on Build GRCh38
rs936325190 Chr.15: 85089324-85089324 on Build GRCh38
rs371799625 Chr.15: 85089330-85089330 on Build GRCh38

View Full Product Details