Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151213364-151213589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED12L
Entrez Gene ID
116931
Gene Name
mediator complex subunit 12 like
Gene Aliases
NOPAR, TNRC11L, TRALP, TRALPUSH
UniGene
Hs.744234
Gene Chromosome Location
Chr.3: 151085665-151436677 on Build GRCh38
Transcripts
NM_053002.5 XM_006713487.3 XM_011512390.2 XM_011512394.2 XM_011512399.2 XM_017005676.1 XM_017005677.1 XM_017005678.1 XM_017005679.1 XM_017005680.1 NM_001081455.1 NM_014879.3 XM_005247922.3 XM_005247923.3 XM_011513340.2 XM_017007583.1

Gene Symbol
P2RY14
Entrez Gene ID
9934
Gene Name
purinergic receptor P2Y14
Gene Aliases
BPR105, GPR105, P2Y14
UniGene
Hs.2465
Gene Chromosome Location
Chr.3: 151212115-151279165 on Build GRCh38
Transcripts
NM_053002.5 XM_006713487.3 XM_011512390.2 XM_011512394.2 XM_011512399.2 XM_017005676.1 XM_017005677.1 XM_017005678.1 XM_017005679.1 XM_017005680.1 NM_001081455.1 NM_014879.3 XM_005247922.3 XM_005247923.3 XM_011513340.2 XM_017007583.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7172542 c.845T>C p.L282P Chr.3: 151213472-151213472 on Build GRCh38
1260770 c.784G>A p.E262K Chr.3: 151213533-151213533 on Build GRCh38
5398392 c.937C>T p.P313S Chr.3: 151213380-151213380 on Build GRCh38
6896254 c.851C>G p.S284C Chr.3: 151213466-151213466 on Build GRCh38
7093805 c.765C>T p.P255P Chr.3: 151213552-151213552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750563526 Chr.3: 151213505-151213505 on Build GRCh38
rs146811566 Chr.3: 151213409-151213409 on Build GRCh38
rs145589776 Chr.3: 151213497-151213497 on Build GRCh38
rs571558200 Chr.3: 151213438-151213438 on Build GRCh38
rs763811837 Chr.3: 151213420-151213420 on Build GRCh38
rs567130380 Chr.3: 151213496-151213496 on Build GRCh38
rs755518330 Chr.3: 151213522-151213522 on Build GRCh38
rs201894452 Chr.3: 151213533-151213533 on Build GRCh38
rs771639317 Chr.3: 151213538-151213538 on Build GRCh38
rs182884078 Chr.3: 151213495-151213495 on Build GRCh38
rs763417485 Chr.3: 151213544-151213545 on Build GRCh38
rs148423792 Chr.3: 151213506-151213506 on Build GRCh38
rs187659532 Chr.3: 151213539-151213539 on Build GRCh38
rs781679355 Chr.3: 151213480-151213480 on Build GRCh38
rs780730940 Chr.3: 151213451-151213451 on Build GRCh38
rs528794736 Chr.3: 151213432-151213432 on Build GRCh38
rs754128917 Chr.3: 151213429-151213429 on Build GRCh38
rs769208048 Chr.3: 151213479-151213479 on Build GRCh38
rs758370216 Chr.3: 151213448-151213448 on Build GRCh38
rs747451996 Chr.3: 151213461-151213461 on Build GRCh38
rs1004347927 Chr.3: 151213424-151213424 on Build GRCh38
rs373767329 Chr.3: 151213485-151213485 on Build GRCh38
rs770001752 Chr.3: 151213534-151213534 on Build GRCh38
rs912376640 Chr.3: 151213543-151213543 on Build GRCh38
rs141983619 Chr.3: 151213555-151213555 on Build GRCh38
rs765862509 Chr.3: 151213398-151213398 on Build GRCh38
rs371105315 Chr.3: 151213503-151213503 on Build GRCh38
rs868480435 Chr.3: 151213560-151213560 on Build GRCh38
rs773831838 Chr.3: 151213492-151213492 on Build GRCh38
rs781719077 Chr.3: 151213526-151213526 on Build GRCh38
rs139896697 Chr.3: 151213433-151213433 on Build GRCh38
rs74811185 Chr.3: 151213415-151213415 on Build GRCh38
rs140553172 Chr.3: 151213494-151213494 on Build GRCh38
rs780275102 Chr.3: 151213515-151213515 on Build GRCh38
rs202190979 Chr.3: 151213416-151213416 on Build GRCh38
rs142582353 Chr.3: 151213517-151213517 on Build GRCh38
rs759104990 Chr.3: 151213493-151213493 on Build GRCh38
rs774970575 Chr.3: 151213541-151213541 on Build GRCh38
rs147553856 Chr.3: 151213535-151213535 on Build GRCh38
rs144133888 Chr.3: 151213414-151213414 on Build GRCh38
rs978641151 Chr.3: 151213443-151213443 on Build GRCh38

View Full Product Details