Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2233084-2233355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E4F1
Entrez Gene ID
1877
Gene Name
E4F transcription factor 1
Gene Aliases
E4F
UniGene
Hs.513268
Gene Chromosome Location
Chr.16: 2223488-2235742 on Build GRCh38
Transcripts
NM_001288776.1 NM_001288778.1 NM_004424.4 XM_005255155.1 XM_005255156.1 XM_006720858.1 XM_011522402.1 XM_011522403.1 XM_017023010.1 XM_017023011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4679996 c.1008C>T p.H336H Chr.16: 2233135-2233135 on Build GRCh38
5623611 c.1010T>G p.V337G Chr.16: 2233137-2233137 on Build GRCh38
6651085 c.991G>A p.V331I Chr.16: 2233118-2233118 on Build GRCh38
4748654 c.1024C>A p.L342M Chr.16: 2233151-2233151 on Build GRCh38
3279231 c.1039A>G p.K347E Chr.16: 2233166-2233166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764947330 Chr.16: 2233185-2233186 on Build GRCh38
rs766832827 Chr.16: 2233116-2233116 on Build GRCh38
rs757793831 Chr.16: 2233195-2233195 on Build GRCh38
rs920295834 Chr.16: 2233127-2233127 on Build GRCh38
rs370693095 Chr.16: 2233306-2233306 on Build GRCh38
rs773481655 Chr.16: 2233192-2233192 on Build GRCh38
rs369317092 Chr.16: 2233118-2233118 on Build GRCh38
rs753134859 Chr.16: 2233233-2233233 on Build GRCh38
rs200055937 Chr.16: 2233108-2233108 on Build GRCh38
rs547092769 Chr.16: 2233284-2233284 on Build GRCh38
rs534827168 Chr.16: 2233318-2233318 on Build GRCh38
rs530921933 Chr.16: 2233169-2233169 on Build GRCh38
rs997610691 Chr.16: 2233201-2233201 on Build GRCh38
rs771770063 Chr.16: 2233159-2233159 on Build GRCh38
rs996294534 Chr.16: 2233200-2233200 on Build GRCh38
rs554520634 Chr.16: 2233326-2233326 on Build GRCh38
rs889130567 Chr.16: 2233213-2233213 on Build GRCh38
rs879377196 Chr.16: 2233329-2233329 on Build GRCh38
rs59845216 Chr.16: 2233327-2233327 on Build GRCh38
rs565702323 Chr.16: 2233314-2233314 on Build GRCh38
rs750129537 Chr.16: 2233194-2233195 on Build GRCh38
rs1026062428 Chr.16: 2233266-2233266 on Build GRCh38
rs373359809 Chr.16: 2233191-2233191 on Build GRCh38
rs551031835 Chr.16: 2233195-2233195 on Build GRCh38
rs539965441 Chr.16: 2233255-2233255 on Build GRCh38
rs756713317 Chr.16: 2233232-2233232 on Build GRCh38
rs371113367 Chr.16: 2233189-2233189 on Build GRCh38
rs570940739 Chr.16: 2233205-2233205 on Build GRCh38
rs778472592 Chr.16: 2233135-2233135 on Build GRCh38
rs758005235 Chr.16: 2233234-2233234 on Build GRCh38
rs745517753 Chr.16: 2233157-2233157 on Build GRCh38
rs765598650 Chr.16: 2233204-2233204 on Build GRCh38
rs781584850 Chr.16: 2233120-2233120 on Build GRCh38
rs374953286 Chr.16: 2233126-2233126 on Build GRCh38
rs768589395 Chr.16: 2233180-2233180 on Build GRCh38
rs761884575 Chr.16: 2233190-2233190 on Build GRCh38
rs1016159921 Chr.16: 2233253-2233253 on Build GRCh38
rs369274242 Chr.16: 2233117-2233117 on Build GRCh38
rs1031394028 Chr.16: 2233254-2233254 on Build GRCh38
rs970845245 Chr.16: 2233285-2233285 on Build GRCh38
rs999553305 Chr.16: 2233218-2233218 on Build GRCh38
rs186588193 Chr.16: 2233163-2233163 on Build GRCh38
rs765920788 Chr.16: 2233210-2233211 on Build GRCh38
rs750675436 Chr.16: 2233106-2233106 on Build GRCh38
rs751916028 Chr.16: 2233202-2233202 on Build GRCh38
rs1014107499 Chr.16: 2233262-2233262 on Build GRCh38

View Full Product Details