Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108449120-108449347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCC2
Entrez Gene ID
9648
Gene Name
GRIP and coiled-coil domain containing 2
Gene Aliases
GCC185, RANBP2L4, REN53
UniGene
Hs.436505
Gene Chromosome Location
Chr.2: 108448561-108509398 on Build GRCh38
Transcripts
NM_181453.3 NR_028063.1 XM_006712870.3 XM_006712871.1 XM_006712872.3

SNP Detail

SNP ID Chromosome Location
rs1032848549 Chr.2: 108449295-108449295 on Build GRCh38
rs900127381 Chr.2: 108449169-108449169 on Build GRCh38
rs769865848 Chr.2: 108449236-108449256 on Build GRCh38
rs759956198 Chr.2: 108449239-108449239 on Build GRCh38
rs920494835 Chr.2: 108449256-108449256 on Build GRCh38
rs747693822 Chr.2: 108449229-108449229 on Build GRCh38
rs574492010 Chr.2: 108449222-108449222 on Build GRCh38
rs983296114 Chr.2: 108449307-108449307 on Build GRCh38
rs957398932 Chr.2: 108449299-108449299 on Build GRCh38
rs1023965750 Chr.2: 108449262-108449262 on Build GRCh38
rs944403618 Chr.2: 108449151-108449152 on Build GRCh38
rs989095657 Chr.2: 108449157-108449157 on Build GRCh38
rs1032200284 Chr.2: 108449232-108449232 on Build GRCh38
rs147355200 Chr.2: 108449139-108449139 on Build GRCh38
rs575208339 Chr.2: 108449252-108449252 on Build GRCh38
rs900616886 Chr.2: 108449213-108449213 on Build GRCh38
rs1040349590 Chr.2: 108449192-108449192 on Build GRCh38
rs370365824 Chr.2: 108449251-108449251 on Build GRCh38
rs765637800 Chr.2: 108449246-108449246 on Build GRCh38
rs541919188 Chr.2: 108449234-108449234 on Build GRCh38
rs904708033 Chr.2: 108449201-108449201 on Build GRCh38
rs532747944 Chr.2: 108449319-108449319 on Build GRCh38
rs764691463 Chr.2: 108449235-108449235 on Build GRCh38
rs887917053 Chr.2: 108449248-108449248 on Build GRCh38
rs1013954399 Chr.2: 108449260-108449260 on Build GRCh38
rs767705277 Chr.2: 108449289-108449289 on Build GRCh38
rs935951664 Chr.2: 108449152-108449152 on Build GRCh38
rs565602080 Chr.2: 108449282-108449282 on Build GRCh38
rs911669135 Chr.2: 108449314-108449314 on Build GRCh38
rs530992367 Chr.2: 108449204-108449204 on Build GRCh38
rs757471595 Chr.2: 108449274-108449274 on Build GRCh38
rs771544904 Chr.2: 108449231-108449231 on Build GRCh38
rs544110704 Chr.2: 108449265-108449265 on Build GRCh38
rs553030659 Chr.2: 108449197-108449197 on Build GRCh38
rs776397797 Chr.2: 108449257-108449267 on Build GRCh38
rs751810629 Chr.2: 108449271-108449271 on Build GRCh38
rs896734136 Chr.2: 108449257-108449257 on Build GRCh38
rs979825410 Chr.2: 108449292-108449292 on Build GRCh38
rs774301106 Chr.2: 108449277-108449277 on Build GRCh38
rs913239792 Chr.2: 108449165-108449165 on Build GRCh38
rs779885564 Chr.2: 108449145-108449145 on Build GRCh38
rs1014717464 Chr.2: 108449242-108449242 on Build GRCh38
rs534536327 Chr.2: 108449189-108449189 on Build GRCh38
rs548050427 Chr.2: 108449321-108449321 on Build GRCh38

View Full Product Details