Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 94129420-94129647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PANX1
Entrez Gene ID
24145
Gene Name
pannexin 1
Gene Aliases
MRS1, PX1, UNQ2529
UniGene
Hs.591976
Gene Chromosome Location
Chr.11: 94128928-94185596 on Build GRCh38
Transcripts
NM_015368.3 XM_017017464.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6070774 c.129G>T p.V43V Chr.11: 94129441-94129441 on Build GRCh38
690915 c.152C>G p.S51W Chr.11: 94129464-94129464 on Build GRCh38
690914 c.162C>A p.F54L Chr.11: 94129474-94129474 on Build GRCh38
253793 c.177G>A p.S59S Chr.11: 94129489-94129489 on Build GRCh38
6135119 c.158C>T p.A53V Chr.11: 94129470-94129470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766980953 Chr.11: 94129438-94129438 on Build GRCh38
rs140681094 Chr.11: 94129459-94129459 on Build GRCh38
rs367864274 Chr.11: 94129536-94129536 on Build GRCh38
rs771754751 Chr.11: 94129524-94129524 on Build GRCh38
rs756088285 Chr.11: 94129506-94129506 on Build GRCh38
rs757480669 Chr.11: 94129479-94129479 on Build GRCh38
rs764404754 Chr.11: 94129464-94129464 on Build GRCh38
rs751922313 Chr.11: 94129538-94129538 on Build GRCh38
rs765578243 Chr.11: 94129609-94129609 on Build GRCh38
rs371580391 Chr.11: 94129503-94129503 on Build GRCh38
rs369317573 Chr.11: 94129571-94129571 on Build GRCh38
rs760031836 Chr.11: 94129447-94129447 on Build GRCh38
rs574756479 Chr.11: 94129495-94129495 on Build GRCh38
rs1049543406 Chr.11: 94129468-94129468 on Build GRCh38
rs1023102526 Chr.11: 94129610-94129610 on Build GRCh38
rs751803002 Chr.11: 94129477-94129477 on Build GRCh38
rs764492607 Chr.11: 94129534-94129534 on Build GRCh38
rs371876803 Chr.11: 94129530-94129530 on Build GRCh38
rs772743657 Chr.11: 94129444-94129444 on Build GRCh38
rs200745097 Chr.11: 94129516-94129516 on Build GRCh38
rs1012013202 Chr.11: 94129579-94129579 on Build GRCh38
rs758769376 Chr.11: 94129462-94129462 on Build GRCh38
rs745947961 Chr.11: 94129496-94129496 on Build GRCh38
rs781195700 Chr.11: 94129481-94129481 on Build GRCh38
rs767793830 Chr.11: 94129541-94129541 on Build GRCh38
rs753146841 Chr.11: 94129460-94129460 on Build GRCh38
rs973297582 Chr.11: 94129483-94129483 on Build GRCh38
rs759700706 Chr.11: 94129540-94129540 on Build GRCh38
rs1022885735 Chr.11: 94129443-94129443 on Build GRCh38
rs760249250 Chr.11: 94129526-94129526 on Build GRCh38
rs765812508 Chr.11: 94129529-94129529 on Build GRCh38
rs138436783 Chr.11: 94129519-94129519 on Build GRCh38
rs973173885 Chr.11: 94129439-94129439 on Build GRCh38
rs750518405 Chr.11: 94129479-94129481 on Build GRCh38
rs896232108 Chr.11: 94129576-94129576 on Build GRCh38
rs750542914 Chr.11: 94129544-94129544 on Build GRCh38
rs772660482 Chr.11: 94129525-94129525 on Build GRCh38
rs766248075 Chr.11: 94129559-94129559 on Build GRCh38
rs763403399 Chr.11: 94129531-94129531 on Build GRCh38
rs747792332 Chr.11: 94129521-94129521 on Build GRCh38
rs758633405 Chr.11: 94129515-94129515 on Build GRCh38
rs751467023 Chr.11: 94129601-94129601 on Build GRCh38
rs999185609 Chr.11: 94129550-94129550 on Build GRCh38
rs539844723 Chr.11: 94129499-94129499 on Build GRCh38
rs908597891 Chr.11: 94129528-94129528 on Build GRCh38
rs374725362 Chr.11: 94129518-94129518 on Build GRCh38
rs780208325 Chr.11: 94129533-94129534 on Build GRCh38
rs780144027 Chr.11: 94129511-94129511 on Build GRCh38
rs1041028068 Chr.11: 94129558-94129558 on Build GRCh38

View Full Product Details