Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 77908616-77908869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHTF2
Entrez Gene ID
57157
Gene Name
putative homeodomain transcription factor 2
Gene Aliases
-
UniGene
Hs.203965
Gene Chromosome Location
Chr.7: 77798792-77957504 on Build GRCh38
Transcripts
NM_001127357.1 NM_001127358.1 NM_001127359.1 NM_001127360.1 NM_020432.4 XM_005250508.4 XM_005250509.4 XM_006716063.3 XM_011516422.2 XM_011516423.2 XM_011516424.2 XM_017012458.1 XM_017012459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1623110 c.463T>C p.F155L Chr.7: 77908810-77908810 on Build GRCh38
1220490 c.512G>A p.G171E Chr.7: 77908859-77908859 on Build GRCh38
6745676 c.395T>C p.L132P Chr.7: 77908844-77908844 on Build GRCh38
6548374 c.505G>T p.V169L Chr.7: 77908852-77908852 on Build GRCh38
1623109 c.361T>C p.F121L Chr.7: 77908810-77908810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375153750 Chr.7: 77908839-77908839 on Build GRCh38
rs768988835 Chr.7: 77908751-77908751 on Build GRCh38
rs769321414 Chr.7: 77908665-77908665 on Build GRCh38
rs765688393 Chr.7: 77908764-77908764 on Build GRCh38
rs763870384 Chr.7: 77908803-77908803 on Build GRCh38
rs552247090 Chr.7: 77908818-77908818 on Build GRCh38
rs751125580 Chr.7: 77908783-77908784 on Build GRCh38
rs773757563 Chr.7: 77908771-77908771 on Build GRCh38
rs777285593 Chr.7: 77908756-77908756 on Build GRCh38
rs757234620 Chr.7: 77908822-77908822 on Build GRCh38
rs993563234 Chr.7: 77908730-77908730 on Build GRCh38
rs751542223 Chr.7: 77908784-77908784 on Build GRCh38
rs760257499 Chr.7: 77908750-77908750 on Build GRCh38
rs201800333 Chr.7: 77908827-77908827 on Build GRCh38
rs766653203 Chr.7: 77908779-77908779 on Build GRCh38
rs745695610 Chr.7: 77908834-77908834 on Build GRCh38
rs762480530 Chr.7: 77908760-77908760 on Build GRCh38
rs848494 Chr.7: 77908647-77908647 on Build GRCh38
rs755098403 Chr.7: 77908789-77908789 on Build GRCh38
rs779772271 Chr.7: 77908843-77908843 on Build GRCh38
rs906217185 Chr.7: 77908767-77908767 on Build GRCh38
rs939687601 Chr.7: 77908719-77908719 on Build GRCh38
rs778648648 Chr.7: 77908824-77908824 on Build GRCh38
rs182131021 Chr.7: 77908774-77908774 on Build GRCh38
rs769643295 Chr.7: 77908741-77908741 on Build GRCh38

View Full Product Details