Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23026141-23026402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMB5
Entrez Gene ID
5693
Gene Name
proteasome subunit beta 5
Gene Aliases
LMPX, MB1, X
UniGene
Hs.422990
Gene Chromosome Location
Chr.14: 23016543-23035220 on Build GRCh38
Transcripts
NM_001130725.1 NM_001144932.2 NM_002797.4 XM_005267871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4933016 c.635A>G p.Y212C Chr.14: 23026246-23026246 on Build GRCh38
6878687 c.542C>T p.S181L Chr.14: 23026339-23026339 on Build GRCh38
6996000 c.622G>C p.V208L Chr.14: 23026259-23026259 on Build GRCh38
7118997 c.734G>C p.R245P Chr.14: 23026147-23026147 on Build GRCh38
3744269 c.652G>A p.A218T Chr.14: 23026229-23026229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748576681 Chr.14: 23026334-23026334 on Build GRCh38
rs377286048 Chr.14: 23026165-23026165 on Build GRCh38
rs749175846 Chr.14: 23026174-23026174 on Build GRCh38
rs140183720 Chr.14: 23026222-23026222 on Build GRCh38
rs200785674 Chr.14: 23026199-23026199 on Build GRCh38
rs759473572 Chr.14: 23026368-23026368 on Build GRCh38
rs372901909 Chr.14: 23026345-23026345 on Build GRCh38
rs140179061 Chr.14: 23026178-23026178 on Build GRCh38
rs760437419 Chr.14: 23026227-23026227 on Build GRCh38
rs759817497 Chr.14: 23026272-23026272 on Build GRCh38
rs779779692 Chr.14: 23026326-23026326 on Build GRCh38
rs763505796 Chr.14: 23026356-23026356 on Build GRCh38
rs765075694 Chr.14: 23026242-23026243 on Build GRCh38
rs768368849 Chr.14: 23026332-23026332 on Build GRCh38
rs113133875 Chr.14: 23026377-23026377 on Build GRCh38
rs745405707 Chr.14: 23026171-23026171 on Build GRCh38
rs57148615 Chr.14: 23026293-23026293 on Build GRCh38
rs891067967 Chr.14: 23026211-23026211 on Build GRCh38
rs369321865 Chr.14: 23026330-23026330 on Build GRCh38
rs60224002 Chr.14: 23026246-23026246 on Build GRCh38
rs968890668 Chr.14: 23026337-23026337 on Build GRCh38
rs58556496 Chr.14: 23026333-23026333 on Build GRCh38
rs768191445 Chr.14: 23026215-23026215 on Build GRCh38
rs758398552 Chr.14: 23026322-23026322 on Build GRCh38
rs369529779 Chr.14: 23026166-23026166 on Build GRCh38
rs375675979 Chr.14: 23026364-23026364 on Build GRCh38
rs766491079 Chr.14: 23026260-23026260 on Build GRCh38
rs778741331 Chr.14: 23026309-23026309 on Build GRCh38
rs12043 Chr.14: 23026265-23026265 on Build GRCh38
rs767565432 Chr.14: 23026283-23026283 on Build GRCh38
rs773542531 Chr.14: 23026346-23026346 on Build GRCh38
rs752906559 Chr.14: 23026375-23026375 on Build GRCh38
rs1047397353 Chr.14: 23026203-23026203 on Build GRCh38
rs750438167 Chr.14: 23026319-23026319 on Build GRCh38
rs56688661 Chr.14: 23026169-23026169 on Build GRCh38
rs774264387 Chr.14: 23026371-23026371 on Build GRCh38
rs770354735 Chr.14: 23026235-23026235 on Build GRCh38
rs751623199 Chr.14: 23026266-23026266 on Build GRCh38
rs377390691 Chr.14: 23026170-23026170 on Build GRCh38
rs562640901 Chr.14: 23026365-23026365 on Build GRCh38
rs373543398 Chr.14: 23026176-23026176 on Build GRCh38
rs746718225 Chr.14: 23026180-23026180 on Build GRCh38
rs753693697 Chr.14: 23026285-23026285 on Build GRCh38
rs763417612 Chr.14: 23026248-23026248 on Build GRCh38
rs748037051 Chr.14: 23026276-23026276 on Build GRCh38
rs145689628 Chr.14: 23026234-23026234 on Build GRCh38
rs768336631 Chr.14: 23026336-23026336 on Build GRCh38

View Full Product Details