Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 132214319-132214544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAP2C
Entrez Gene ID
57826
Gene Name
RAP2C, member of RAS oncogene family
Gene Aliases
-
UniGene
Hs.119889
Gene Chromosome Location
Chr.X: 132203024-132219480 on Build GRCh38
Transcripts
NM_001271186.1 NM_001271187.1 NM_021183.4 NR_073150.1 XM_006724775.2 XM_011531376.1

SNP Detail

SNP ID Chromosome Location
rs747147776 Chr.X: 132214423-132214423 on Build GRCh38
rs758547404 Chr.X: 132214354-132214354 on Build GRCh38
rs748853826 Chr.X: 132214479-132214479 on Build GRCh38
rs35821161 Chr.X: 132214352-132214353 on Build GRCh38
rs369025253 Chr.X: 132214451-132214451 on Build GRCh38
rs41310444 Chr.X: 132214482-132214482 on Build GRCh38
rs752946402 Chr.X: 132214345-132214345 on Build GRCh38
rs901907878 Chr.X: 132214491-132214491 on Build GRCh38
rs148846997 Chr.X: 132214498-132214498 on Build GRCh38
rs376860828 Chr.X: 132214402-132214402 on Build GRCh38

View Full Product Details