Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56121279-56121542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC3H10
Entrez Gene ID
84872
Gene Name
zinc finger CCCH-type containing 10
Gene Aliases
ZC3HDC10
UniGene
Hs.632706
Gene Chromosome Location
Chr.12: 56118220-56122497 on Build GRCh38
Transcripts
NM_001303124.1 NM_001303125.1 NM_032786.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4043481 c.942T>C p.T314T Chr.12: 56121504-56121504 on Build GRCh38
7244967 c.970C>T p.R324C Chr.12: 56121532-56121532 on Build GRCh38
431484 c.749G>T p.R250L Chr.12: 56121311-56121311 on Build GRCh38
7348968 c.826C>T p.R276C Chr.12: 56121388-56121388 on Build GRCh38
4926265 c.891C>T p.P297P Chr.12: 56121453-56121453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770989690 Chr.12: 56121490-56121490 on Build GRCh38
rs150174605 Chr.12: 56121388-56121388 on Build GRCh38
rs146320702 Chr.12: 56121435-56121435 on Build GRCh38
rs747280201 Chr.12: 56121342-56121342 on Build GRCh38
rs907606088 Chr.12: 56121312-56121312 on Build GRCh38
rs867406239 Chr.12: 56121497-56121497 on Build GRCh38
rs759663408 Chr.12: 56121356-56121356 on Build GRCh38
rs141972777 Chr.12: 56121411-56121411 on Build GRCh38
rs569602632 Chr.12: 56121315-56121315 on Build GRCh38
rs138695692 Chr.12: 56121392-56121392 on Build GRCh38
rs755399288 Chr.12: 56121309-56121309 on Build GRCh38
rs139617086 Chr.12: 56121445-56121445 on Build GRCh38
rs1041917107 Chr.12: 56121500-56121500 on Build GRCh38
rs148045185 Chr.12: 56121375-56121375 on Build GRCh38
rs767051817 Chr.12: 56121405-56121405 on Build GRCh38
rs771026927 Chr.12: 56121346-56121346 on Build GRCh38
rs751191460 Chr.12: 56121389-56121389 on Build GRCh38
rs146983581 Chr.12: 56121349-56121349 on Build GRCh38
rs781493573 Chr.12: 56121508-56121508 on Build GRCh38
rs141780038 Chr.12: 56121383-56121383 on Build GRCh38
rs747246765 Chr.12: 56121467-56121467 on Build GRCh38
rs184474754 Chr.12: 56121431-56121431 on Build GRCh38
rs772253304 Chr.12: 56121305-56121305 on Build GRCh38
rs750672741 Chr.12: 56121324-56121326 on Build GRCh38
rs923903701 Chr.12: 56121463-56121463 on Build GRCh38
rs113893957 Chr.12: 56121420-56121420 on Build GRCh38
rs946198827 Chr.12: 56121456-56121456 on Build GRCh38
rs371576118 Chr.12: 56121510-56121510 on Build GRCh38
rs780827493 Chr.12: 56121356-56121357 on Build GRCh38
rs963192123 Chr.12: 56121322-56121322 on Build GRCh38
rs536642969 Chr.12: 56121363-56121363 on Build GRCh38
rs758808670 Chr.12: 56121442-56121442 on Build GRCh38
rs775432609 Chr.12: 56121364-56121364 on Build GRCh38
rs778335553 Chr.12: 56121444-56121444 on Build GRCh38

View Full Product Details