Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 129814577-129814848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC9
Entrez Gene ID
51114
Gene Name
zinc finger DHHC-type containing 9
Gene Aliases
CGI89, CXorf11, DHHC9, MMSA1, MRXSZ, ZDHHC10, ZNF379, ZNF380
UniGene
Hs.193566
Gene Chromosome Location
Chr.X: 129803288-129843934 on Build GRCh38
Transcripts
NM_001008222.2 NM_016032.3

SNP Detail

SNP ID Chromosome Location
rs757136158 Chr.X: 129814673-129814673 on Build GRCh38
rs748827100 Chr.X: 129814704-129814704 on Build GRCh38
rs778798330 Chr.X: 129814663-129814663 on Build GRCh38
rs373699283 Chr.X: 129814638-129814638 on Build GRCh38
rs762762877 Chr.X: 129814807-129814808 on Build GRCh38
rs370408984 Chr.X: 129814821-129814821 on Build GRCh38
rs367846813 Chr.X: 129814812-129814812 on Build GRCh38
rs113127450 Chr.X: 129814804-129814804 on Build GRCh38
rs773721454 Chr.X: 129814710-129814710 on Build GRCh38
rs766688756 Chr.X: 129814636-129814636 on Build GRCh38
rs759659851 Chr.X: 129814810-129814810 on Build GRCh38
rs781147368 Chr.X: 129814674-129814674 on Build GRCh38
rs761289081 Chr.X: 129814752-129814752 on Build GRCh38
rs773907113 Chr.X: 129814734-129814734 on Build GRCh38
rs898868823 Chr.X: 129814823-129814823 on Build GRCh38
rs756591605 Chr.X: 129814635-129814635 on Build GRCh38
rs755229553 Chr.X: 129814662-129814662 on Build GRCh38
rs762304463 Chr.X: 129814614-129814614 on Build GRCh38
rs752687711 Chr.X: 129814814-129814814 on Build GRCh38
rs754241413 Chr.X: 129814793-129814793 on Build GRCh38
rs769527426 Chr.X: 129814685-129814685 on Build GRCh38
rs745638507 Chr.X: 129814678-129814678 on Build GRCh38
rs766780497 Chr.X: 129814764-129814764 on Build GRCh38
rs866000327 Chr.X: 129814661-129814661 on Build GRCh38
rs772679685 Chr.X: 129814718-129814718 on Build GRCh38
rs887261815 Chr.X: 129814686-129814686 on Build GRCh38
rs748313244 Chr.X: 129814668-129814668 on Build GRCh38

View Full Product Details