Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 99327466-99327739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TXNDC9
Entrez Gene ID
10190
Gene Name
thioredoxin domain containing 9
Gene Aliases
APACD, PHLP3
UniGene
Hs.536122
Gene Chromosome Location
Chr.2: 99306184-99336397 on Build GRCh38
Transcripts
NM_005783.3 XM_017003147.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4738496 c.269A>G p.E90G Chr.2: 99327574-99327574 on Build GRCh38
6038335 c.275T>C p.V92A Chr.2: 99327568-99327568 on Build GRCh38
4096589 c.251A>G p.Q84R Chr.2: 99327592-99327592 on Build GRCh38
6902446 c.261G>A p.K87K Chr.2: 99327582-99327582 on Build GRCh38
4096588 c.282C>T p.C94C Chr.2: 99327561-99327561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374799446 Chr.2: 99327513-99327513 on Build GRCh38
rs370820067 Chr.2: 99327505-99327505 on Build GRCh38
rs117462941 Chr.2: 99327705-99327705 on Build GRCh38
rs762519882 Chr.2: 99327498-99327498 on Build GRCh38
rs776728528 Chr.2: 99327693-99327694 on Build GRCh38
rs757430454 Chr.2: 99327581-99327581 on Build GRCh38
rs142231194 Chr.2: 99327594-99327594 on Build GRCh38
rs938344152 Chr.2: 99327530-99327530 on Build GRCh38
rs188413408 Chr.2: 99327693-99327693 on Build GRCh38
rs140023296 Chr.2: 99327527-99327527 on Build GRCh38
rs747256250 Chr.2: 99327552-99327552 on Build GRCh38
rs748796557 Chr.2: 99327622-99327622 on Build GRCh38
rs763877964 Chr.2: 99327499-99327499 on Build GRCh38
rs13390809 Chr.2: 99327606-99327606 on Build GRCh38
rs752789698 Chr.2: 99327543-99327543 on Build GRCh38
rs758477113 Chr.2: 99327544-99327544 on Build GRCh38
rs778019186 Chr.2: 99327550-99327550 on Build GRCh38
rs768851394 Chr.2: 99327683-99327684 on Build GRCh38
rs774123262 Chr.2: 99327504-99327504 on Build GRCh38
rs766879581 Chr.2: 99327509-99327509 on Build GRCh38
rs761558462 Chr.2: 99327675-99327675 on Build GRCh38
rs747369555 Chr.2: 99327497-99327498 on Build GRCh38
rs767217090 Chr.2: 99327689-99327689 on Build GRCh38
rs745661932 Chr.2: 99327605-99327605 on Build GRCh38
rs768300795 Chr.2: 99327648-99327648 on Build GRCh38
rs765764832 Chr.2: 99327536-99327536 on Build GRCh38
rs775464247 Chr.2: 99327610-99327610 on Build GRCh38

View Full Product Details