Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38867304-38867551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN11A
Entrez Gene ID
11280
Gene Name
sodium voltage-gated channel alpha subunit 11
Gene Aliases
FEPS3, HSAN7, NAV1.9, NaN, PN5, SCN12A, SNS-2
UniGene
Hs.591657
Gene Chromosome Location
Chr.3: 38845764-39051986 on Build GRCh38
Transcripts
NM_001287223.1 NM_014139.2 XM_011533321.2 XM_017005647.1 XM_017005648.1 XM_017005649.1 XM_017005650.1 XM_017005651.1 XM_017005652.1 XM_017005653.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1582581 c.3943C>T p.Q1315* Chr.3: 38867329-38867329 on Build GRCh38
3593694 c.3876C>T p.I1292I Chr.3: 38867396-38867396 on Build GRCh38
4634981 c.3908T>C p.F1303S Chr.3: 38867364-38867364 on Build GRCh38
4117423 c.3926A>T p.D1309V Chr.3: 38867346-38867346 on Build GRCh38
1537887 c.3898C>T p.L1300L Chr.3: 38867374-38867374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777977342 Chr.3: 38867331-38867331 on Build GRCh38
rs761590329 Chr.3: 38867395-38867395 on Build GRCh38
rs773570414 Chr.3: 38867453-38867456 on Build GRCh38
rs368772706 Chr.3: 38867415-38867415 on Build GRCh38
rs763336682 Chr.3: 38867356-38867356 on Build GRCh38
rs371852759 Chr.3: 38867345-38867345 on Build GRCh38
rs531276047 Chr.3: 38867357-38867357 on Build GRCh38
rs373458111 Chr.3: 38867461-38867461 on Build GRCh38
rs778934870 Chr.3: 38867460-38867460 on Build GRCh38
rs865937367 Chr.3: 38867427-38867427 on Build GRCh38
rs755416318 Chr.3: 38867421-38867421 on Build GRCh38
rs753000639 Chr.3: 38867423-38867423 on Build GRCh38
rs963175151 Chr.3: 38867420-38867420 on Build GRCh38
rs747904828 Chr.3: 38867470-38867470 on Build GRCh38
rs148945365 Chr.3: 38867453-38867453 on Build GRCh38
rs772937687 Chr.3: 38867404-38867404 on Build GRCh38
rs987168812 Chr.3: 38867502-38867502 on Build GRCh38
rs144377677 Chr.3: 38867432-38867432 on Build GRCh38
rs201791247 Chr.3: 38867379-38867379 on Build GRCh38
rs764798607 Chr.3: 38867398-38867398 on Build GRCh38
rs770940292 Chr.3: 38867344-38867344 on Build GRCh38
rs749374877 Chr.3: 38867337-38867337 on Build GRCh38
rs767982270 Chr.3: 38867422-38867422 on Build GRCh38
rs545544680 Chr.3: 38867482-38867482 on Build GRCh38
rs773984449 Chr.3: 38867500-38867500 on Build GRCh38
rs759678040 Chr.3: 38867518-38867518 on Build GRCh38
rs376861071 Chr.3: 38867361-38867361 on Build GRCh38
rs768469678 Chr.3: 38867467-38867467 on Build GRCh38
rs200920463 Chr.3: 38867380-38867380 on Build GRCh38
rs756436762 Chr.3: 38867429-38867429 on Build GRCh38
rs769578847 Chr.3: 38867478-38867478 on Build GRCh38
rs765838942 Chr.3: 38867483-38867484 on Build GRCh38
rs762662164 Chr.3: 38867491-38867491 on Build GRCh38
rs142412836 Chr.3: 38867444-38867444 on Build GRCh38
rs866777927 Chr.3: 38867446-38867446 on Build GRCh38
rs765774073 Chr.3: 38867446-38867447 on Build GRCh38
rs267599811 Chr.3: 38867396-38867396 on Build GRCh38
rs762160601 Chr.3: 38867363-38867364 on Build GRCh38
rs751187345 Chr.3: 38867516-38867516 on Build GRCh38
rs536925812 Chr.3: 38867407-38867407 on Build GRCh38
rs763469989 Chr.3: 38867468-38867469 on Build GRCh38
rs781114326 Chr.3: 38867468-38867468 on Build GRCh38
rs752151721 Chr.3: 38867416-38867416 on Build GRCh38

View Full Product Details