Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153563675-153563928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
S100A2
Entrez Gene ID
6273
Gene Name
S100 calcium binding protein A2
Gene Aliases
CAN19, S100L
UniGene
Hs.516484
Gene Chromosome Location
Chr.1: 153561108-153565830 on Build GRCh38
Transcripts
NM_005978.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3474458 c.49C>T p.H17Y Chr.1: 153563826-153563826 on Build GRCh38
314962 c.62G>A p.C21Y Chr.1: 153563813-153563813 on Build GRCh38
3801938 c.41C>T p.T14I Chr.1: 153563834-153563834 on Build GRCh38
4022608 c.72C>T p.G24G Chr.1: 153563803-153563803 on Build GRCh38
4172115 c.77A>C p.K26T Chr.1: 153563798-153563798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781693025 Chr.1: 153563823-153563823 on Build GRCh38
rs770160790 Chr.1: 153563842-153563842 on Build GRCh38
rs11550733 Chr.1: 153563751-153563751 on Build GRCh38
rs201315637 Chr.1: 153563710-153563710 on Build GRCh38
rs926549039 Chr.1: 153563839-153563839 on Build GRCh38
rs758899452 Chr.1: 153563802-153563802 on Build GRCh38
rs1027544620 Chr.1: 153563797-153563797 on Build GRCh38
rs183265370 Chr.1: 153563733-153563733 on Build GRCh38
rs769112915 Chr.1: 153563721-153563721 on Build GRCh38
rs148993921 Chr.1: 153563856-153563856 on Build GRCh38
rs775541513 Chr.1: 153563745-153563745 on Build GRCh38
rs753488429 Chr.1: 153563758-153563758 on Build GRCh38
rs374712470 Chr.1: 153563869-153563869 on Build GRCh38
rs749924923 Chr.1: 153563778-153563778 on Build GRCh38
rs764903448 Chr.1: 153563872-153563872 on Build GRCh38
rs112041093 Chr.1: 153563889-153563889 on Build GRCh38
rs766161606 Chr.1: 153563833-153563836 on Build GRCh38
rs199540195 Chr.1: 153563898-153563898 on Build GRCh38
rs113847783 Chr.1: 153563732-153563732 on Build GRCh38
rs762355218 Chr.1: 153563882-153563882 on Build GRCh38
rs760367189 Chr.1: 153563749-153563749 on Build GRCh38
rs769924848 Chr.1: 153563734-153563734 on Build GRCh38
rs367758325 Chr.1: 153563884-153563884 on Build GRCh38
rs755552724 Chr.1: 153563909-153563909 on Build GRCh38
rs200362870 Chr.1: 153563873-153563873 on Build GRCh38
rs760736868 Chr.1: 153563860-153563860 on Build GRCh38
rs2233868 Chr.1: 153563870-153563870 on Build GRCh38
rs376852593 Chr.1: 153563851-153563851 on Build GRCh38
rs370761990 Chr.1: 153563737-153563737 on Build GRCh38
rs974292486 Chr.1: 153563743-153563743 on Build GRCh38
rs575985349 Chr.1: 153563852-153563852 on Build GRCh38
rs967873224 Chr.1: 153563762-153563762 on Build GRCh38
rs371526881 Chr.1: 153563859-153563859 on Build GRCh38
rs780402122 Chr.1: 153563803-153563803 on Build GRCh38
rs752164817 Chr.1: 153563806-153563806 on Build GRCh38
rs764670446 Chr.1: 153563773-153563773 on Build GRCh38
rs577890090 Chr.1: 153563717-153563717 on Build GRCh38
rs769300684 Chr.1: 153563817-153563817 on Build GRCh38
rs761460462 Chr.1: 153563764-153563764 on Build GRCh38
rs906174700 Chr.1: 153563707-153563707 on Build GRCh38
rs532172691 Chr.1: 153563811-153563811 on Build GRCh38
rs768374019 Chr.1: 153563861-153563861 on Build GRCh38
rs773436829 Chr.1: 153563735-153563735 on Build GRCh38
rs912571521 Chr.1: 153563832-153563832 on Build GRCh38
rs748318834 Chr.1: 153563825-153563825 on Build GRCh38

View Full Product Details