Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 8638088-8638343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB12
Entrez Gene ID
201475
Gene Name
RAB12, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.270074
Gene Chromosome Location
Chr.18: 8609428-8639383 on Build GRCh38
Transcripts
NM_001025300.2 XM_006722300.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3821787 c.646G>A p.E216K Chr.18: 8638173-8638173 on Build GRCh38
1730825 c.677C>A p.P226Q Chr.18: 8638204-8638204 on Build GRCh38
6250862 c.681G>T p.E227D Chr.18: 8638208-8638208 on Build GRCh38
5762707 c.669G>A p.S223S Chr.18: 8638196-8638196 on Build GRCh38
4854354 c.697G>C p.E233Q Chr.18: 8638224-8638224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115140842 Chr.18: 8638277-8638277 on Build GRCh38
rs763198374 Chr.18: 8638279-8638279 on Build GRCh38
rs759731885 Chr.18: 8638196-8638196 on Build GRCh38
rs374013427 Chr.18: 8638170-8638170 on Build GRCh38
rs776751207 Chr.18: 8638130-8638130 on Build GRCh38
rs762356282 Chr.18: 8638214-8638214 on Build GRCh38
rs778629538 Chr.18: 8638145-8638145 on Build GRCh38
rs542697781 Chr.18: 8638129-8638132 on Build GRCh38
rs769999520 Chr.18: 8638183-8638183 on Build GRCh38
rs547006682 Chr.18: 8638252-8638252 on Build GRCh38
rs368602516 Chr.18: 8638285-8638285 on Build GRCh38
rs779044651 Chr.18: 8638122-8638123 on Build GRCh38
rs750810141 Chr.18: 8638219-8638219 on Build GRCh38
rs761125389 Chr.18: 8638298-8638298 on Build GRCh38
rs762289949 Chr.18: 8638287-8638287 on Build GRCh38
rs772131118 Chr.18: 8638119-8638119 on Build GRCh38
rs981781065 Chr.18: 8638281-8638281 on Build GRCh38
rs769525201 Chr.18: 8638269-8638269 on Build GRCh38
rs773026027 Chr.18: 8638171-8638171 on Build GRCh38
rs777290251 Chr.18: 8638247-8638247 on Build GRCh38
rs189697693 Chr.18: 8638190-8638190 on Build GRCh38
rs201948442 Chr.18: 8638182-8638182 on Build GRCh38
rs766807365 Chr.18: 8638313-8638313 on Build GRCh38
rs759707470 Chr.18: 8638131-8638131 on Build GRCh38
rs528816647 Chr.18: 8638234-8638234 on Build GRCh38
rs764500324 Chr.18: 8638138-8638138 on Build GRCh38
rs754641460 Chr.18: 8638144-8638144 on Build GRCh38
rs775608519 Chr.18: 8638184-8638184 on Build GRCh38
rs1027819367 Chr.18: 8638200-8638200 on Build GRCh38
rs374700160 Chr.18: 8638218-8638218 on Build GRCh38
rs370554407 Chr.18: 8638259-8638259 on Build GRCh38
rs77888227 Chr.18: 8638128-8638128 on Build GRCh38
rs765350320 Chr.18: 8638132-8638132 on Build GRCh38
rs781567633 Chr.18: 8638142-8638142 on Build GRCh38
rs369554529 Chr.18: 8638143-8638143 on Build GRCh38
rs771092962 Chr.18: 8638123-8638126 on Build GRCh38
rs757875629 Chr.18: 8638246-8638246 on Build GRCh38
rs752050253 Chr.18: 8638140-8638140 on Build GRCh38
rs748132522 Chr.18: 8638118-8638118 on Build GRCh38
rs776390777 Chr.18: 8638181-8638181 on Build GRCh38
rs745763608 Chr.18: 8638123-8638123 on Build GRCh38
rs909695860 Chr.18: 8638213-8638213 on Build GRCh38
rs752743851 Chr.18: 8638133-8638133 on Build GRCh38
rs746923211 Chr.18: 8638178-8638178 on Build GRCh38
rs778721570 Chr.18: 8638223-8638223 on Build GRCh38
rs768051827 Chr.18: 8638296-8638296 on Build GRCh38
rs560999631 Chr.18: 8638122-8638122 on Build GRCh38
rs984091796 Chr.18: 8638203-8638203 on Build GRCh38
rs562391414 Chr.18: 8638253-8638253 on Build GRCh38
rs767576309 Chr.18: 8638305-8638305 on Build GRCh38
rs377561775 Chr.18: 8638220-8638220 on Build GRCh38
rs370446984 Chr.18: 8638189-8638189 on Build GRCh38
rs750902179 Chr.18: 8638297-8638297 on Build GRCh38
rs760399488 Chr.18: 8638120-8638120 on Build GRCh38
rs747646438 Chr.18: 8638154-8638154 on Build GRCh38
rs768663810 Chr.18: 8638280-8638280 on Build GRCh38

View Full Product Details