Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41978054-41978283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A3
Entrez Gene ID
478
Gene Name
ATPase Na+/K+ transporting subunit alpha 3
Gene Aliases
AHC2, ATP1A1, CAPOS, DYT12, RDP
UniGene
Hs.515427
Gene Chromosome Location
Chr.19: 41966582-41994276 on Build GRCh38
Transcripts
NM_001256213.1 NM_001256214.1 NM_152296.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4078614 c.1710G>A p.T570T Chr.19: 41978247-41978247 on Build GRCh38
7215414 c.1776G>A p.A592A Chr.19: 41978181-41978181 on Build GRCh38
3534554 c.1763C>T p.A588V Chr.19: 41978194-41978194 on Build GRCh38
5741604 c.1775C>T p.A592V Chr.19: 41978182-41978182 on Build GRCh38
6358788 c.1807-1G>T p.? Chr.19: 41978073-41978073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782116412 Chr.19: 41978222-41978222 on Build GRCh38
rs782778845 Chr.19: 41978106-41978106 on Build GRCh38
rs782415633 Chr.19: 41978243-41978243 on Build GRCh38
rs377372631 Chr.19: 41978137-41978137 on Build GRCh38
rs782081809 Chr.19: 41978250-41978250 on Build GRCh38
rs781853644 Chr.19: 41978107-41978107 on Build GRCh38
rs781827466 Chr.19: 41978133-41978133 on Build GRCh38
rs781964931 Chr.19: 41978094-41978094 on Build GRCh38
rs782078148 Chr.19: 41978097-41978097 on Build GRCh38
rs375255226 Chr.19: 41978262-41978262 on Build GRCh38
rs977103188 Chr.19: 41978199-41978199 on Build GRCh38
rs944271704 Chr.19: 41978217-41978217 on Build GRCh38
rs782220233 Chr.19: 41978184-41978184 on Build GRCh38
rs2288507 Chr.19: 41978181-41978181 on Build GRCh38
rs781840505 Chr.19: 41978247-41978247 on Build GRCh38
rs3206846 Chr.19: 41978209-41978209 on Build GRCh38
rs370436157 Chr.19: 41978229-41978229 on Build GRCh38
rs782612231 Chr.19: 41978127-41978127 on Build GRCh38
rs782166038 Chr.19: 41978111-41978111 on Build GRCh38
rs782017635 Chr.19: 41978241-41978241 on Build GRCh38
rs782149339 Chr.19: 41978076-41978076 on Build GRCh38
rs782302598 Chr.19: 41978091-41978091 on Build GRCh38
rs782300402 Chr.19: 41978141-41978141 on Build GRCh38
rs782409824 Chr.19: 41978163-41978163 on Build GRCh38
rs886054474 Chr.19: 41978238-41978238 on Build GRCh38
rs782335069 Chr.19: 41978193-41978193 on Build GRCh38
rs782694072 Chr.19: 41978246-41978246 on Build GRCh38
rs202091721 Chr.19: 41978113-41978113 on Build GRCh38
rs782422094 Chr.19: 41978231-41978231 on Build GRCh38
rs781992944 Chr.19: 41978211-41978211 on Build GRCh38
rs1057519113 Chr.19: 41978162-41978162 on Build GRCh38

View Full Product Details