Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 142874953-142875226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP26
Entrez Gene ID
23092
Gene Name
Rho GTPase activating protein 26
Gene Aliases
GRAF, GRAF1, OPHN1L, OPHN1L1
UniGene
Hs.654668
Gene Chromosome Location
Chr.5: 142770370-143229007 on Build GRCh38
Transcripts
NM_001135608.1 NM_015071.4 XM_005268398.4 XM_005268399.4 XM_005268400.4 XM_005268402.4 XM_006714774.3 XM_011537610.2 XM_011537611.2 XM_011537612.2 XM_017009247.1 XM_017009248.1 XM_017009249.1 XM_017009250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3611846 c.309G>A p.R103R Chr.5: 142875168-142875168 on Build GRCh38
7419056 c.308G>A p.R103Q Chr.5: 142875167-142875167 on Build GRCh38
5912184 c.251-4C>T p.? Chr.5: 142875106-142875106 on Build GRCh38
6295058 c.312+8G>A p.? Chr.5: 142875179-142875179 on Build GRCh38
7216828 c.255A>G p.R85R Chr.5: 142875114-142875114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995849580 Chr.5: 142875082-142875082 on Build GRCh38
rs774671486 Chr.5: 142875132-142875132 on Build GRCh38
rs765897153 Chr.5: 142875200-142875200 on Build GRCh38
rs562548632 Chr.5: 142875029-142875029 on Build GRCh38
rs370338871 Chr.5: 142875093-142875093 on Build GRCh38
rs189466953 Chr.5: 142875014-142875014 on Build GRCh38
rs370435398 Chr.5: 142875061-142875061 on Build GRCh38
rs1022024368 Chr.5: 142875101-142875101 on Build GRCh38
rs772467491 Chr.5: 142875156-142875156 on Build GRCh38
rs776566350 Chr.5: 142875161-142875161 on Build GRCh38
rs587778050 Chr.5: 142875166-142875166 on Build GRCh38
rs749343713 Chr.5: 142874980-142874980 on Build GRCh38
rs762486970 Chr.5: 142875190-142875190 on Build GRCh38
rs1056889597 Chr.5: 142875032-142875032 on Build GRCh38
rs1040326332 Chr.5: 142875034-142875035 on Build GRCh38
rs759537245 Chr.5: 142875064-142875065 on Build GRCh38
rs886298397 Chr.5: 142874988-142874988 on Build GRCh38
rs554203882 Chr.5: 142875009-142875009 on Build GRCh38
rs750334908 Chr.5: 142875185-142875185 on Build GRCh38
rs185200 Chr.5: 142875114-142875114 on Build GRCh38
rs770917906 Chr.5: 142875126-142875126 on Build GRCh38
rs373317587 Chr.5: 142875135-142875135 on Build GRCh38
rs761574373 Chr.5: 142875167-142875167 on Build GRCh38
rs538094045 Chr.5: 142875017-142875017 on Build GRCh38
rs900128198 Chr.5: 142875059-142875059 on Build GRCh38
rs778977295 Chr.5: 142875181-142875181 on Build GRCh38

View Full Product Details