Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108616522-108616747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DZIP3
Entrez Gene ID
9666
Gene Name
DAZ interacting zinc finger protein 3
Gene Aliases
PPP1R66, UURF2, hRUL138
UniGene
Hs.409210
Gene Chromosome Location
Chr.3: 108589490-108694846 on Build GRCh38
Transcripts
NM_014648.3 XM_005247914.3 XM_005247915.3 XM_005247916.3 XM_005247917.3 XM_005247918.3 XM_005247919.3 XM_017007540.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5059189 c.343G>A p.A115T Chr.3: 108616625-108616625 on Build GRCh38
1736891 c.275A>G p.N92S Chr.3: 108616557-108616557 on Build GRCh38
445270 c.358C>G p.Q120E Chr.3: 108616640-108616640 on Build GRCh38
3944639 c.363T>C p.A121A Chr.3: 108616645-108616645 on Build GRCh38
6340322 c.349A>T p.R117W Chr.3: 108616631-108616631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923040424 Chr.3: 108616714-108616714 on Build GRCh38
rs143969766 Chr.3: 108616598-108616598 on Build GRCh38
rs755188825 Chr.3: 108616567-108616567 on Build GRCh38
rs984701709 Chr.3: 108616647-108616647 on Build GRCh38
rs1039131705 Chr.3: 108616710-108616710 on Build GRCh38
rs950555928 Chr.3: 108616637-108616637 on Build GRCh38
rs933089083 Chr.3: 108616719-108616719 on Build GRCh38
rs564059785 Chr.3: 108616648-108616648 on Build GRCh38
rs780049137 Chr.3: 108616555-108616555 on Build GRCh38
rs543968481 Chr.3: 108616599-108616599 on Build GRCh38
rs753896992 Chr.3: 108616676-108616676 on Build GRCh38
rs759187615 Chr.3: 108616618-108616618 on Build GRCh38
rs759351599 Chr.3: 108616559-108616559 on Build GRCh38
rs373285274 Chr.3: 108616658-108616658 on Build GRCh38
rs373181033 Chr.3: 108616623-108616623 on Build GRCh38
rs772522908 Chr.3: 108616583-108616583 on Build GRCh38
rs12330782 Chr.3: 108616657-108616657 on Build GRCh38
rs775332144 Chr.3: 108616641-108616641 on Build GRCh38
rs747499362 Chr.3: 108616596-108616596 on Build GRCh38
rs761741416 Chr.3: 108616660-108616660 on Build GRCh38
rs748434749 Chr.3: 108616578-108616578 on Build GRCh38
rs767506687 Chr.3: 108616673-108616673 on Build GRCh38
rs759511983 Chr.3: 108616658-108616659 on Build GRCh38
rs773478113 Chr.3: 108616587-108616587 on Build GRCh38

View Full Product Details