Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42553670-42553931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B1
Entrez Gene ID
3292
Gene Name
hydroxysteroid 17-beta dehydrogenase 1
Gene Aliases
EDH17B2, EDHB17, HSD17, SDR28C1
UniGene
Hs.654385
Gene Chromosome Location
Chr.17: 42551966-42555214 on Build GRCh38
Transcripts
NM_000413.2 XM_005257292.3 XM_006721857.3 XM_006721858.3 XM_006721859.3 XM_011524729.1 XM_011524730.1 XM_011524731.1 XM_011524732.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1737353 c.530T>C p.F177S Chr.17: 42553878-42553878 on Build GRCh38
6682324 c.490G>A p.E164K Chr.17: 42553838-42553838 on Build GRCh38
4844775 c.539+7C>A p.? Chr.17: 42553894-42553894 on Build GRCh38
979523 c.462C>T p.D154D Chr.17: 42553810-42553810 on Build GRCh38
6839402 c.517C>G p.L173V Chr.17: 42553865-42553865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371977976 Chr.17: 42553798-42553798 on Build GRCh38
rs760313890 Chr.17: 42553886-42553886 on Build GRCh38
rs978179640 Chr.17: 42553735-42553735 on Build GRCh38
rs753396022 Chr.17: 42553817-42553817 on Build GRCh38
rs201791531 Chr.17: 42553870-42553870 on Build GRCh38
rs888220255 Chr.17: 42553692-42553693 on Build GRCh38
rs762115742 Chr.17: 42553784-42553784 on Build GRCh38
rs753451117 Chr.17: 42553887-42553887 on Build GRCh38
rs183748964 Chr.17: 42553734-42553734 on Build GRCh38
rs565532649 Chr.17: 42553831-42553831 on Build GRCh38
rs188862602 Chr.17: 42553859-42553859 on Build GRCh38
rs149630844 Chr.17: 42553832-42553832 on Build GRCh38
rs959613715 Chr.17: 42553699-42553699 on Build GRCh38
rs754492233 Chr.17: 42553890-42553890 on Build GRCh38
rs777493205 Chr.17: 42553904-42553904 on Build GRCh38
rs967211454 Chr.17: 42553733-42553733 on Build GRCh38
rs749346120 Chr.17: 42553860-42553860 on Build GRCh38
rs369544710 Chr.17: 42553849-42553849 on Build GRCh38
rs545340852 Chr.17: 42553705-42553705 on Build GRCh38
rs774798762 Chr.17: 42553864-42553864 on Build GRCh38
rs752619767 Chr.17: 42553897-42553897 on Build GRCh38
rs767229862 Chr.17: 42553788-42553788 on Build GRCh38
rs771332059 Chr.17: 42553750-42553750 on Build GRCh38
rs537368847 Chr.17: 42553694-42553694 on Build GRCh38
rs756339089 Chr.17: 42553909-42553909 on Build GRCh38
rs610166 Chr.17: 42553697-42553697 on Build GRCh38
rs775623518 Chr.17: 42553771-42553771 on Build GRCh38
rs764552186 Chr.17: 42553773-42553773 on Build GRCh38
rs768702899 Chr.17: 42553743-42553743 on Build GRCh38
rs763515724 Chr.17: 42553772-42553772 on Build GRCh38
rs755663520 Chr.17: 42553805-42553805 on Build GRCh38
rs146159533 Chr.17: 42553842-42553842 on Build GRCh38
rs192431327 Chr.17: 42553695-42553695 on Build GRCh38
rs773613890 Chr.17: 42553878-42553878 on Build GRCh38
rs377154591 Chr.17: 42553822-42553822 on Build GRCh38
rs779490029 Chr.17: 42553806-42553806 on Build GRCh38
rs769700504 Chr.17: 42553767-42553767 on Build GRCh38
rs72547451 Chr.17: 42553755-42553755 on Build GRCh38
rs767716724 Chr.17: 42553872-42553872 on Build GRCh38
rs769551521 Chr.17: 42553755-42553755 on Build GRCh38
rs1022450001 Chr.17: 42553714-42553714 on Build GRCh38
rs377503215 Chr.17: 42553850-42553850 on Build GRCh38
rs769171720 Chr.17: 42553862-42553862 on Build GRCh38
rs531762225 Chr.17: 42553760-42553760 on Build GRCh38
rs770048386 Chr.17: 42553853-42553853 on Build GRCh38
rs200100050 Chr.17: 42553749-42553749 on Build GRCh38
rs543721042 Chr.17: 42553801-42553801 on Build GRCh38
rs772783090 Chr.17: 42553863-42553863 on Build GRCh38
rs10153 Chr.17: 42553769-42553769 on Build GRCh38
rs778921822 Chr.17: 42553745-42553745 on Build GRCh38
rs762489199 Chr.17: 42553875-42553881 on Build GRCh38
rs1003758694 Chr.17: 42553696-42553696 on Build GRCh38
rs751968719 Chr.17: 42553782-42553782 on Build GRCh38
rs758339758 Chr.17: 42553899-42553899 on Build GRCh38
rs911886629 Chr.17: 42553741-42553741 on Build GRCh38
rs545312595 Chr.17: 42553827-42553827 on Build GRCh38
rs610177 Chr.17: 42553706-42553706 on Build GRCh38
rs746786104 Chr.17: 42553908-42553908 on Build GRCh38
rs765300838 Chr.17: 42553894-42553894 on Build GRCh38

View Full Product Details