Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 223619234-223619503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN8
Entrez Gene ID
388743
Gene Name
calpain 8
Gene Aliases
nCL-2
UniGene
Hs.291487
Gene Chromosome Location
Chr.1: 223541609-223665734 on Build GRCh38
Transcripts
NM_001143962.1 XM_017001265.1 XM_017001266.1 XM_017001267.1 XM_017001268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4448408 c.977C>T p.S326L Chr.1: 223619448-223619448 on Build GRCh38
1960452 c.1039C>G p.L347V Chr.1: 223619386-223619386 on Build GRCh38
5989394 c.1124A>T p.N375I Chr.1: 223619301-223619301 on Build GRCh38
1960454 c.983C>T p.S328L Chr.1: 223619442-223619442 on Build GRCh38
5528126 c.990C>T p.F330F Chr.1: 223619435-223619435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038953936 Chr.1: 223619269-223619269 on Build GRCh38
rs543782414 Chr.1: 223619341-223619341 on Build GRCh38
rs778746550 Chr.1: 223619279-223619279 on Build GRCh38
rs1033343831 Chr.1: 223619402-223619402 on Build GRCh38
rs908506815 Chr.1: 223619428-223619428 on Build GRCh38
rs757171953 Chr.1: 223619355-223619355 on Build GRCh38
rs182284158 Chr.1: 223619469-223619469 on Build GRCh38
rs900946933 Chr.1: 223619364-223619364 on Build GRCh38
rs940105570 Chr.1: 223619453-223619453 on Build GRCh38
rs755607859 Chr.1: 223619378-223619378 on Build GRCh38
rs1003141384 Chr.1: 223619360-223619360 on Build GRCh38
rs759700721 Chr.1: 223619435-223619435 on Build GRCh38
rs904895184 Chr.1: 223619347-223619347 on Build GRCh38
rs139467366 Chr.1: 223619419-223619419 on Build GRCh38
rs751395423 Chr.1: 223619342-223619342 on Build GRCh38
rs748872891 Chr.1: 223619396-223619396 on Build GRCh38
rs955397607 Chr.1: 223619409-223619409 on Build GRCh38
rs558644268 Chr.1: 223619386-223619386 on Build GRCh38
rs79075255 Chr.1: 223619393-223619393 on Build GRCh38
rs776732822 Chr.1: 223619254-223619254 on Build GRCh38
rs536587066 Chr.1: 223619328-223619328 on Build GRCh38
rs879242644 Chr.1: 223619397-223619397 on Build GRCh38
rs572261534 Chr.1: 223619281-223619281 on Build GRCh38
rs1009284375 Chr.1: 223619375-223619375 on Build GRCh38
rs375331004 Chr.1: 223619341-223619342 on Build GRCh38
rs760661935 Chr.1: 223619374-223619374 on Build GRCh38
rs191187914 Chr.1: 223619407-223619407 on Build GRCh38
rs980292281 Chr.1: 223619275-223619275 on Build GRCh38
rs770102905 Chr.1: 223619462-223619462 on Build GRCh38
rs553969720 Chr.1: 223619259-223619259 on Build GRCh38
rs930916972 Chr.1: 223619299-223619299 on Build GRCh38
rs370081508 Chr.1: 223619340-223619340 on Build GRCh38
rs765265205 Chr.1: 223619293-223619293 on Build GRCh38
rs1024378067 Chr.1: 223619408-223619408 on Build GRCh38
rs772568044 Chr.1: 223619401-223619401 on Build GRCh38
rs1038432368 Chr.1: 223619471-223619471 on Build GRCh38
rs373951019 Chr.1: 223619410-223619410 on Build GRCh38
rs554837985 Chr.1: 223619329-223619329 on Build GRCh38
rs948878463 Chr.1: 223619478-223619478 on Build GRCh38
rs570655166 Chr.1: 223619371-223619371 on Build GRCh38
rs796661732 Chr.1: 223619369-223619369 on Build GRCh38
rs940237629 Chr.1: 223619268-223619268 on Build GRCh38
rs776875184 Chr.1: 223619418-223619418 on Build GRCh38
rs200538064 Chr.1: 223619377-223619377 on Build GRCh38
rs541489689 Chr.1: 223619403-223619403 on Build GRCh38
rs759759114 Chr.1: 223619475-223619475 on Build GRCh38

View Full Product Details