Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77040782-77041280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT6
Entrez Gene ID
192134
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Gene Aliases
B3Gn-T6, BGnT-6, beta-1,3-Gn-T6, beta3Gn-T6
UniGene
Hs.352622
Gene Chromosome Location
Chr.11: 77034341-77041973 on Build GRCh38
Transcripts
NM_138706.4

SNP Detail

SNP ID Chromosome Location
rs140772091 Chr.11: 77040874-77040874 on Build GRCh38
rs563738521 Chr.11: 77040919-77040919 on Build GRCh38
rs1047531418 Chr.11: 77040974-77040974 on Build GRCh38
rs929230030 Chr.11: 77040930-77040930 on Build GRCh38
rs782189095 Chr.11: 77041131-77041131 on Build GRCh38
rs546721713 Chr.11: 77040978-77040978 on Build GRCh38
rs1000996913 Chr.11: 77040992-77040992 on Build GRCh38
rs372555707 Chr.11: 77041247-77041247 on Build GRCh38
rs985864533 Chr.11: 77040858-77040858 on Build GRCh38
rs905247204 Chr.11: 77040988-77040988 on Build GRCh38
rs985769819 Chr.11: 77041122-77041122 on Build GRCh38
rs782122044 Chr.11: 77041257-77041257 on Build GRCh38
rs575924228 Chr.11: 77041088-77041088 on Build GRCh38
rs1009802655 Chr.11: 77041048-77041048 on Build GRCh38
rs1030439455 Chr.11: 77041113-77041113 on Build GRCh38
rs572454531 Chr.11: 77041042-77041042 on Build GRCh38
rs540341951 Chr.11: 77041176-77041176 on Build GRCh38
rs896475526 Chr.11: 77040977-77040977 on Build GRCh38
rs754213742 Chr.11: 77041159-77041159 on Build GRCh38
rs375961635 Chr.11: 77041036-77041036 on Build GRCh38
rs1045146846 Chr.11: 77040982-77040982 on Build GRCh38
rs11237063 Chr.11: 77040869-77040869 on Build GRCh38
rs372242703 Chr.11: 77041160-77041160 on Build GRCh38
rs910274197 Chr.11: 77040865-77040865 on Build GRCh38
rs151165441 Chr.11: 77041121-77041121 on Build GRCh38
rs536362814 Chr.11: 77040848-77040848 on Build GRCh38
rs113733996 Chr.11: 77040842-77040842 on Build GRCh38
rs992773651 Chr.11: 77041205-77041205 on Build GRCh38
rs7950506 Chr.11: 77041201-77041201 on Build GRCh38
rs183065013 Chr.11: 77041237-77041237 on Build GRCh38
rs973192119 Chr.11: 77040883-77040883 on Build GRCh38
rs919092728 Chr.11: 77041163-77041163 on Build GRCh38
rs528922276 Chr.11: 77041258-77041258 on Build GRCh38
rs530804897 Chr.11: 77041105-77041105 on Build GRCh38
rs1045010687 Chr.11: 77041253-77041253 on Build GRCh38
rs538890021 Chr.11: 77041022-77041022 on Build GRCh38

View Full Product Details