Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108505832-108506376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXPH5
Entrez Gene ID
23086
Gene Name
exophilin 5
Gene Aliases
SLAC2-B, SLAC2B
UniGene
Hs.28540
Gene Chromosome Location
Chr.11: 108505431-108607514 on Build GRCh38
Transcripts
NM_001144763.1 NM_001144764.1 NM_001144765.1 NM_001308019.1 NM_015065.2 XM_011542696.2 XM_011542698.2 XM_011542700.2 XM_017017397.1 XM_017017398.1 XM_017017399.1 XM_017017400.1 XM_017017401.1 XM_017017402.1 XM_017017403.1 XM_017017404.1

SNP Detail

SNP ID Chromosome Location
rs994549388 Chr.11: 108506030-108506030 on Build GRCh38
rs868697194 Chr.11: 108506114-108506114 on Build GRCh38
rs988420507 Chr.11: 108506122-108506122 on Build GRCh38
rs946770209 Chr.11: 108505859-108505859 on Build GRCh38
rs754201512 Chr.11: 108506001-108506001 on Build GRCh38
rs930449805 Chr.11: 108506315-108506315 on Build GRCh38
rs940656051 Chr.11: 108505975-108505975 on Build GRCh38
rs944728225 Chr.11: 108506213-108506213 on Build GRCh38
rs969056946 Chr.11: 108506093-108506093 on Build GRCh38
rs866061388 Chr.11: 108506156-108506156 on Build GRCh38
rs573961741 Chr.11: 108506071-108506072 on Build GRCh38
rs940568384 Chr.11: 108506304-108506304 on Build GRCh38
rs920403304 Chr.11: 108506309-108506309 on Build GRCh38
rs555773296 Chr.11: 108506048-108506048 on Build GRCh38
rs770794048 Chr.11: 108506200-108506200 on Build GRCh38
rs915448208 Chr.11: 108505858-108505858 on Build GRCh38
rs994367469 Chr.11: 108505927-108505927 on Build GRCh38
rs975437343 Chr.11: 108506196-108506196 on Build GRCh38
rs987202465 Chr.11: 108506247-108506247 on Build GRCh38
rs957170651 Chr.11: 108506117-108506117 on Build GRCh38
rs986241550 Chr.11: 108506207-108506207 on Build GRCh38
rs910547563 Chr.11: 108506209-108506209 on Build GRCh38
rs759855526 Chr.11: 108506323-108506323 on Build GRCh38
rs1040378487 Chr.11: 108506217-108506217 on Build GRCh38
rs890349301 Chr.11: 108505910-108505910 on Build GRCh38
rs115321903 Chr.11: 108506308-108506308 on Build GRCh38
rs929924027 Chr.11: 108506256-108506256 on Build GRCh38
rs908573213 Chr.11: 108506284-108506284 on Build GRCh38
rs1022382993 Chr.11: 108506191-108506191 on Build GRCh38
rs915428825 Chr.11: 108506142-108506142 on Build GRCh38
rs1051129863 Chr.11: 108505906-108505906 on Build GRCh38
rs192679220 Chr.11: 108506099-108506099 on Build GRCh38
rs574802889 Chr.11: 108506060-108506060 on Build GRCh38
rs934004022 Chr.11: 108505889-108505889 on Build GRCh38
rs114652806 Chr.11: 108505977-108505977 on Build GRCh38
rs531838391 Chr.11: 108506242-108506242 on Build GRCh38
rs747653242 Chr.11: 108505936-108505936 on Build GRCh38
rs968208787 Chr.11: 108506195-108506195 on Build GRCh38
rs866583256 Chr.11: 108506109-108506109 on Build GRCh38
rs921332444 Chr.11: 108506241-108506241 on Build GRCh38
rs1035721825 Chr.11: 108506100-108506100 on Build GRCh38
rs901003321 Chr.11: 108505881-108505881 on Build GRCh38
rs921238817 Chr.11: 108505886-108505886 on Build GRCh38
rs1036595878 Chr.11: 108505976-108505976 on Build GRCh38
rs753193695 Chr.11: 108506281-108506283 on Build GRCh38
rs73548748 Chr.11: 108505864-108505864 on Build GRCh38
rs562864325 Chr.11: 108506106-108506106 on Build GRCh38

View Full Product Details