Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111873915-111874410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDXACB1
Entrez Gene ID
91893
Gene Name
ferredoxin-fold anticodon binding domain containing 1
Gene Aliases
hCG_2033039
UniGene
Hs.697132
Gene Chromosome Location
Chr.11: 111874056-111879457 on Build GRCh38
Transcripts
NM_138378.2 NR_038364.1

SNP Detail

SNP ID Chromosome Location
rs949967727 Chr.11: 111874237-111874237 on Build GRCh38
rs112343480 Chr.11: 111874301-111874301 on Build GRCh38
rs781984438 Chr.11: 111874145-111874145 on Build GRCh38
rs1032742105 Chr.11: 111874088-111874088 on Build GRCh38
rs375372665 Chr.11: 111873952-111873952 on Build GRCh38
rs781922318 Chr.11: 111874296-111874296 on Build GRCh38
rs985794162 Chr.11: 111874110-111874110 on Build GRCh38
rs903037870 Chr.11: 111874285-111874285 on Build GRCh38
rs747133471 Chr.11: 111873953-111873953 on Build GRCh38
rs999921576 Chr.11: 111874087-111874087 on Build GRCh38
rs543286957 Chr.11: 111874369-111874369 on Build GRCh38
rs536237399 Chr.11: 111874332-111874332 on Build GRCh38
rs953050995 Chr.11: 111874109-111874109 on Build GRCh38
rs554503092 Chr.11: 111874334-111874334 on Build GRCh38
rs531995722 Chr.11: 111874083-111874083 on Build GRCh38
rs558735122 Chr.11: 111874374-111874374 on Build GRCh38
rs776865387 Chr.11: 111874175-111874175 on Build GRCh38
rs1019619122 Chr.11: 111874140-111874140 on Build GRCh38
rs759692478 Chr.11: 111874317-111874317 on Build GRCh38
rs183604814 Chr.11: 111874159-111874159 on Build GRCh38
rs1020081534 Chr.11: 111874077-111874077 on Build GRCh38
rs917160600 Chr.11: 111874216-111874216 on Build GRCh38
rs377591505 Chr.11: 111874008-111874008 on Build GRCh38
rs111526632 Chr.11: 111874206-111874206 on Build GRCh38
rs782125732 Chr.11: 111874179-111874179 on Build GRCh38
rs770946976 Chr.11: 111874164-111874164 on Build GRCh38
rs575937193 Chr.11: 111874352-111874352 on Build GRCh38
rs1014506397 Chr.11: 111874046-111874046 on Build GRCh38
rs966287943 Chr.11: 111874144-111874144 on Build GRCh38
rs549735301 Chr.11: 111873981-111873981 on Build GRCh38
rs782362474 Chr.11: 111874248-111874248 on Build GRCh38
rs926702672 Chr.11: 111874177-111874177 on Build GRCh38
rs538343566 Chr.11: 111874370-111874370 on Build GRCh38
rs571713115 Chr.11: 111874165-111874165 on Build GRCh38
rs553954043 Chr.11: 111874282-111874282 on Build GRCh38
rs370741925 Chr.11: 111874082-111874082 on Build GRCh38

View Full Product Details