Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 1028052-1028556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERC1
Entrez Gene ID
23085
Gene Name
ELKS/RAB6-interacting/CAST family member 1
Gene Aliases
Cast2, ELKS, ERC-1, RAB6IP2
UniGene
Hs.601216
Gene Chromosome Location
Chr.12: 990010-1495933 on Build GRCh38
Transcripts
NM_001301248.1 NM_178039.3 NM_178040.3 NR_027946.2 NR_027948.2 NR_027949.2 XM_011520941.2 XM_017019052.1 XM_017019053.1 XM_017019054.1 XM_017019055.1 XM_017019056.1 XM_017019057.1 XM_017019058.1 XM_017019059.1 XM_017019060.1 XM_017019061.1 XM_017019062.1 XM_017019063.1 XM_017019064.1 XM_017019065.1 XM_017019066.1 XM_017019067.1 XM_017019068.1 XM_017019069.1 XM_017019070.1 XM_017019071.1 XM_017019072.1 XM_017019073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6902786 c.182C>T p.S61F Chr.12: 1028085-1028085 on Build GRCh38
5633551 c.622A>G p.I208V Chr.12: 1028525-1028525 on Build GRCh38
6315298 c.401G>T p.S134I Chr.12: 1028304-1028304 on Build GRCh38
691991 c.457G>T p.D153Y Chr.12: 1028360-1028360 on Build GRCh38
935426 c.561G>A p.K187K Chr.12: 1028464-1028464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767513298 Chr.12: 1028192-1028192 on Build GRCh38
rs909367142 Chr.12: 1028286-1028286 on Build GRCh38
rs373614890 Chr.12: 1028149-1028149 on Build GRCh38
rs772898864 Chr.12: 1028152-1028152 on Build GRCh38
rs575749350 Chr.12: 1028277-1028277 on Build GRCh38
rs138529974 Chr.12: 1028524-1028524 on Build GRCh38
rs141624941 Chr.12: 1028483-1028483 on Build GRCh38
rs1057436515 Chr.12: 1028360-1028360 on Build GRCh38
rs924791078 Chr.12: 1028170-1028170 on Build GRCh38
rs141356033 Chr.12: 1028267-1028267 on Build GRCh38
rs1000665011 Chr.12: 1028465-1028465 on Build GRCh38
rs767601504 Chr.12: 1028252-1028252 on Build GRCh38
rs750965800 Chr.12: 1028178-1028178 on Build GRCh38
rs759558423 Chr.12: 1028186-1028186 on Build GRCh38
rs761486520 Chr.12: 1028454-1028454 on Build GRCh38
rs1012978614 Chr.12: 1028462-1028462 on Build GRCh38
rs762129792 Chr.12: 1028145-1028145 on Build GRCh38
rs1021192132 Chr.12: 1028082-1028082 on Build GRCh38
rs756664440 Chr.12: 1028482-1028482 on Build GRCh38
rs751768265 Chr.12: 1028477-1028477 on Build GRCh38
rs146174236 Chr.12: 1028200-1028200 on Build GRCh38
rs762673141 Chr.12: 1028233-1028233 on Build GRCh38
rs750942184 Chr.12: 1028340-1028340 on Build GRCh38
rs906126547 Chr.12: 1028334-1028334 on Build GRCh38
rs762430802 Chr.12: 1028155-1028155 on Build GRCh38
rs1017448580 Chr.12: 1028511-1028511 on Build GRCh38
rs770130777 Chr.12: 1028226-1028226 on Build GRCh38
rs891822337 Chr.12: 1028415-1028415 on Build GRCh38
rs138791726 Chr.12: 1028203-1028203 on Build GRCh38
rs375456123 Chr.12: 1028272-1028272 on Build GRCh38
rs765909910 Chr.12: 1028164-1028164 on Build GRCh38
rs781707711 Chr.12: 1028280-1028280 on Build GRCh38
rs199875182 Chr.12: 1028270-1028270 on Build GRCh38
rs770431604 Chr.12: 1028237-1028237 on Build GRCh38
rs778101125 Chr.12: 1028495-1028495 on Build GRCh38
rs370287516 Chr.12: 1028204-1028204 on Build GRCh38
rs942178384 Chr.12: 1028295-1028295 on Build GRCh38
rs751682690 Chr.12: 1028275-1028275 on Build GRCh38
rs142221098 Chr.12: 1028133-1028133 on Build GRCh38
rs772165451 Chr.12: 1028105-1028105 on Build GRCh38
rs903658680 Chr.12: 1028461-1028461 on Build GRCh38
rs771781513 Chr.12: 1028518-1028518 on Build GRCh38
rs759077188 Chr.12: 1028238-1028238 on Build GRCh38
rs755117710 Chr.12: 1028479-1028479 on Build GRCh38
rs777004711 Chr.12: 1028126-1028126 on Build GRCh38
rs757540017 Chr.12: 1028503-1028503 on Build GRCh38
rs780198520 Chr.12: 1028205-1028205 on Build GRCh38
rs766676015 Chr.12: 1028455-1028455 on Build GRCh38
rs769096127 Chr.12: 1028117-1028117 on Build GRCh38
rs564971510 Chr.12: 1028372-1028372 on Build GRCh38
rs760601427 Chr.12: 1028256-1028256 on Build GRCh38
rs772809702 Chr.12: 1028228-1028228 on Build GRCh38
rs762264236 Chr.12: 1028220-1028220 on Build GRCh38
rs778762418 Chr.12: 1028507-1028507 on Build GRCh38
rs749701492 Chr.12: 1028501-1028501 on Build GRCh38
rs551464705 Chr.12: 1028309-1028309 on Build GRCh38
rs767590463 Chr.12: 1028480-1028480 on Build GRCh38
rs148016557 Chr.12: 1028428-1028428 on Build GRCh38
rs767718877 Chr.12: 1028317-1028317 on Build GRCh38
rs763943046 Chr.12: 1028266-1028266 on Build GRCh38
rs1044633185 Chr.12: 1028320-1028320 on Build GRCh38
rs201939562 Chr.12: 1028292-1028292 on Build GRCh38
rs372323969 Chr.12: 1028450-1028450 on Build GRCh38
rs745507164 Chr.12: 1028508-1028508 on Build GRCh38
rs763892753 Chr.12: 1028448-1028448 on Build GRCh38
rs578145160 Chr.12: 1028386-1028386 on Build GRCh38
rs747544281 Chr.12: 1028110-1028110 on Build GRCh38
rs200160550 Chr.12: 1028409-1028409 on Build GRCh38
rs535851221 Chr.12: 1028211-1028211 on Build GRCh38
rs778212010 Chr.12: 1028358-1028358 on Build GRCh38
rs143485606 Chr.12: 1028417-1028417 on Build GRCh38
rs538802682 Chr.12: 1028107-1028107 on Build GRCh38
rs758672154 Chr.12: 1028271-1028271 on Build GRCh38
rs752570620 Chr.12: 1028193-1028193 on Build GRCh38
rs770220650 Chr.12: 1028347-1028347 on Build GRCh38
rs761918440 Chr.12: 1028210-1028210 on Build GRCh38
rs565050203 Chr.12: 1028297-1028297 on Build GRCh38
rs747014051 Chr.12: 1028206-1028206 on Build GRCh38
rs368784319 Chr.12: 1028341-1028341 on Build GRCh38
rs752656449 Chr.12: 1028254-1028254 on Build GRCh38
rs773843924 Chr.12: 1028408-1028408 on Build GRCh38
rs868467597 Chr.12: 1028104-1028104 on Build GRCh38
rs745971025 Chr.12: 1028098-1028098 on Build GRCh38
rs755938605 Chr.12: 1028199-1028199 on Build GRCh38

View Full Product Details