Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12476441-12476943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP16
Entrez Gene ID
80824
Gene Name
dual specificity phosphatase 16
Gene Aliases
MKP-7, MKP7
UniGene
Hs.536535
Gene Chromosome Location
Chr.12: 12473282-12562839 on Build GRCh38
Transcripts
NM_030640.2 XM_005253488.2 XM_006719155.2 XM_011520856.1 XM_011520857.1 XM_017019988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6650335 c.1927C>T p.R643W Chr.12: 12476904-12476904 on Build GRCh38
5715481 c.1978G>A p.E660K Chr.12: 12476853-12476853 on Build GRCh38
4146811 c.1932A>G p.E644E Chr.12: 12476899-12476899 on Build GRCh38
4040013 c.1981A>C p.I661L Chr.12: 12476850-12476850 on Build GRCh38
4980951 c.1967C>T p.S656L Chr.12: 12476864-12476864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753268899 Chr.12: 12476898-12476898 on Build GRCh38
rs577153387 Chr.12: 12476509-12476509 on Build GRCh38
rs927022982 Chr.12: 12476738-12476738 on Build GRCh38
rs187350433 Chr.12: 12476793-12476793 on Build GRCh38
rs769725354 Chr.12: 12476787-12476789 on Build GRCh38
rs779723853 Chr.12: 12476804-12476804 on Build GRCh38
rs954959692 Chr.12: 12476637-12476637 on Build GRCh38
rs1039915259 Chr.12: 12476756-12476756 on Build GRCh38
rs773507562 Chr.12: 12476793-12476793 on Build GRCh38
rs749892665 Chr.12: 12476911-12476911 on Build GRCh38
rs937000050 Chr.12: 12476739-12476739 on Build GRCh38
rs895741953 Chr.12: 12476773-12476773 on Build GRCh38
rs1013481612 Chr.12: 12476591-12476591 on Build GRCh38
rs765710175 Chr.12: 12476897-12476897 on Build GRCh38
rs981777078 Chr.12: 12476672-12476673 on Build GRCh38
rs749453400 Chr.12: 12476792-12476793 on Build GRCh38
rs773210818 Chr.12: 12476870-12476870 on Build GRCh38
rs1006282749 Chr.12: 12476584-12476584 on Build GRCh38
rs895702094 Chr.12: 12476496-12476496 on Build GRCh38
rs1013104273 Chr.12: 12476875-12476875 on Build GRCh38
rs757870510 Chr.12: 12476774-12476774 on Build GRCh38
rs191898046 Chr.12: 12476822-12476822 on Build GRCh38
rs148434374 Chr.12: 12476864-12476864 on Build GRCh38
rs978547301 Chr.12: 12476737-12476737 on Build GRCh38
rs767054549 Chr.12: 12476711-12476711 on Build GRCh38
rs913431446 Chr.12: 12476692-12476692 on Build GRCh38
rs3088226 Chr.12: 12476658-12476658 on Build GRCh38
rs771368303 Chr.12: 12476873-12476876 on Build GRCh38
rs909607409 Chr.12: 12476750-12476753 on Build GRCh38
rs544718104 Chr.12: 12476594-12476594 on Build GRCh38
rs942579794 Chr.12: 12476751-12476751 on Build GRCh38
rs776927720 Chr.12: 12476886-12476886 on Build GRCh38
rs536500522 Chr.12: 12476595-12476595 on Build GRCh38
rs759912271 Chr.12: 12476892-12476892 on Build GRCh38
rs376039829 Chr.12: 12476805-12476805 on Build GRCh38
rs541821907 Chr.12: 12476685-12476685 on Build GRCh38
rs867907881 Chr.12: 12476516-12476516 on Build GRCh38
rs10845553 Chr.12: 12476608-12476608 on Build GRCh38
rs767135469 Chr.12: 12476909-12476909 on Build GRCh38
rs768518627 Chr.12: 12476808-12476808 on Build GRCh38
rs751069705 Chr.12: 12476789-12476789 on Build GRCh38
rs750957475 Chr.12: 12476715-12476715 on Build GRCh38
rs996598693 Chr.12: 12476904-12476904 on Build GRCh38
rs779729235 Chr.12: 12476918-12476918 on Build GRCh38
rs950818433 Chr.12: 12476535-12476535 on Build GRCh38
rs755885672 Chr.12: 12476917-12476917 on Build GRCh38
rs780800182 Chr.12: 12476797-12476797 on Build GRCh38
rs1056762402 Chr.12: 12476745-12476745 on Build GRCh38
rs145539846 Chr.12: 12476863-12476863 on Build GRCh38
rs1031001557 Chr.12: 12476609-12476609 on Build GRCh38
rs560554348 Chr.12: 12476776-12476776 on Build GRCh38
rs755756417 Chr.12: 12476802-12476802 on Build GRCh38
rs760996793 Chr.12: 12476542-12476542 on Build GRCh38
rs751164918 Chr.12: 12476903-12476903 on Build GRCh38
rs774325525 Chr.12: 12476850-12476850 on Build GRCh38
rs760900919 Chr.12: 12476871-12476871 on Build GRCh38
rs749590610 Chr.12: 12476497-12476497 on Build GRCh38
rs765923726 Chr.12: 12476777-12476777 on Build GRCh38
rs766576635 Chr.12: 12476878-12476878 on Build GRCh38
rs958939359 Chr.12: 12476543-12476543 on Build GRCh38
rs879093967 Chr.12: 12476589-12476589 on Build GRCh38
rs888317376 Chr.12: 12476825-12476825 on Build GRCh38

View Full Product Details