Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 18282627-18283125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3C2G
Entrez Gene ID
5288
Gene Name
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 gamma
Gene Aliases
PI3K-C2-gamma, PI3K-C2GAMMA
UniGene
Hs.22500
Gene Chromosome Location
Chr.12: 18242946-18716861 on Build GRCh38
Transcripts
NM_001288772.1 NM_001288774.1 NM_004570.5 XM_011520696.2 XM_011520697.2 XM_011520700.2 XM_011520701.2 XM_017019470.1 XM_017019471.1 XM_017019472.1 XM_017019473.1 XM_017019474.1 XM_017019475.1 XM_017019476.1 XM_017019477.1 XM_017019478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4605704 c.628G>A p.G210S Chr.12: 18282709-18282709 on Build GRCh38
6946794 c.581G>C p.R194T Chr.12: 18282662-18282662 on Build GRCh38
6242020 c.648G>A p.M216I Chr.12: 18282729-18282729 on Build GRCh38
6979431 c.650G>A p.W217* Chr.12: 18282731-18282731 on Build GRCh38
3458744 c.564G>A p.P188P Chr.12: 18282645-18282645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543632492 Chr.12: 18282964-18282964 on Build GRCh38
rs1022427431 Chr.12: 18282921-18282921 on Build GRCh38
rs949652817 Chr.12: 18282985-18282985 on Build GRCh38
rs765088068 Chr.12: 18282759-18282759 on Build GRCh38
rs1017848749 Chr.12: 18282921-18282922 on Build GRCh38
rs999761264 Chr.12: 18282959-18282959 on Build GRCh38
rs759572822 Chr.12: 18282755-18282755 on Build GRCh38
rs886411633 Chr.12: 18282855-18282855 on Build GRCh38
rs771723064 Chr.12: 18282738-18282738 on Build GRCh38
rs539395234 Chr.12: 18282729-18282729 on Build GRCh38
rs895273078 Chr.12: 18282904-18282904 on Build GRCh38
rs918146642 Chr.12: 18282713-18282713 on Build GRCh38
rs766282161 Chr.12: 18282680-18282680 on Build GRCh38
rs768265732 Chr.12: 18282652-18282652 on Build GRCh38
rs774039288 Chr.12: 18282654-18282654 on Build GRCh38
rs556304309 Chr.12: 18282752-18282752 on Build GRCh38
rs772599393 Chr.12: 18282672-18282672 on Build GRCh38
rs963728269 Chr.12: 18283097-18283097 on Build GRCh38
rs748782538 Chr.12: 18282993-18282993 on Build GRCh38
rs757329312 Chr.12: 18282773-18282773 on Build GRCh38
rs776485084 Chr.12: 18282751-18282751 on Build GRCh38
rs564185158 Chr.12: 18282913-18282913 on Build GRCh38
rs760166580 Chr.12: 18282745-18282745 on Build GRCh38
rs577839519 Chr.12: 18283067-18283067 on Build GRCh38
rs1047440459 Chr.12: 18282842-18282842 on Build GRCh38
rs66539336 Chr.12: 18282945-18282946 on Build GRCh38
rs928917899 Chr.12: 18282681-18282681 on Build GRCh38
rs919760018 Chr.12: 18282798-18282798 on Build GRCh38
rs755642096 Chr.12: 18282663-18282663 on Build GRCh38
rs781615674 Chr.12: 18282723-18282723 on Build GRCh38
rs541096386 Chr.12: 18282875-18282875 on Build GRCh38
rs74942548 Chr.12: 18282750-18282750 on Build GRCh38
rs747075150 Chr.12: 18282718-18282718 on Build GRCh38
rs747730521 Chr.12: 18282733-18282733 on Build GRCh38
rs869306502 Chr.12: 18282946-18282947 on Build GRCh38
rs201664666 Chr.12: 18282686-18282686 on Build GRCh38
rs375191216 Chr.12: 18282767-18282767 on Build GRCh38
rs936812975 Chr.12: 18283085-18283085 on Build GRCh38
rs199657792 Chr.12: 18282728-18282728 on Build GRCh38
rs753527495 Chr.12: 18282874-18282874 on Build GRCh38
rs916947689 Chr.12: 18282982-18282982 on Build GRCh38
rs963282257 Chr.12: 18282936-18282936 on Build GRCh38
rs12372174 Chr.12: 18282956-18282956 on Build GRCh38
rs772586527 Chr.12: 18282810-18282815 on Build GRCh38
rs147251688 Chr.12: 18282864-18282864 on Build GRCh38
rs576024637 Chr.12: 18282811-18282811 on Build GRCh38
rs754864262 Chr.12: 18282688-18282688 on Build GRCh38
rs746276290 Chr.12: 18282649-18282649 on Build GRCh38
rs766973024 Chr.12: 18282671-18282671 on Build GRCh38
rs781353001 Chr.12: 18282779-18282779 on Build GRCh38
rs71440362 Chr.12: 18282956-18282957 on Build GRCh38
rs1031685398 Chr.12: 18282963-18282963 on Build GRCh38
rs1038198987 Chr.12: 18282790-18282790 on Build GRCh38
rs758726931 Chr.12: 18282696-18282696 on Build GRCh38
rs377693233 Chr.12: 18283098-18283098 on Build GRCh38
rs572071437 Chr.12: 18282871-18282871 on Build GRCh38
rs757769822 Chr.12: 18282722-18282722 on Build GRCh38
rs1012444965 Chr.12: 18282667-18282667 on Build GRCh38
rs78254698 Chr.12: 18282948-18282948 on Build GRCh38
rs143896492 Chr.12: 18282724-18282724 on Build GRCh38
rs954763171 Chr.12: 18282895-18282895 on Build GRCh38
rs752548090 Chr.12: 18282761-18282761 on Build GRCh38
rs752922078 Chr.12: 18282694-18282694 on Build GRCh38
rs986467375 Chr.12: 18283058-18283058 on Build GRCh38
rs955029925 Chr.12: 18283008-18283008 on Build GRCh38
rs772761967 Chr.12: 18282742-18282742 on Build GRCh38
rs1026036946 Chr.12: 18282955-18282955 on Build GRCh38
rs112217074 Chr.12: 18282770-18282770 on Build GRCh38
rs892551107 Chr.12: 18282796-18282796 on Build GRCh38
rs777843470 Chr.12: 18282701-18282701 on Build GRCh38
rs869213004 Chr.12: 18282957-18282958 on Build GRCh38
rs761476205 Chr.12: 18282665-18282665 on Build GRCh38
rs758638588 Chr.12: 18282765-18282765 on Build GRCh38
rs942533083 Chr.12: 18282786-18282786 on Build GRCh38
rs973236914 Chr.12: 18282948-18282948 on Build GRCh38
rs981078234 Chr.12: 18282990-18282990 on Build GRCh38
rs765538264 Chr.12: 18282689-18282689 on Build GRCh38
rs535095134 Chr.12: 18282831-18282831 on Build GRCh38
rs760600887 Chr.12: 18282677-18282677 on Build GRCh38

View Full Product Details