Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50986298-50986801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC11A2
Entrez Gene ID
4891
Gene Name
solute carrier family 11 member 2
Gene Aliases
AHMIO1, DCT1, DMT1, NRAMP2
UniGene
Hs.505545
Gene Chromosome Location
Chr.12: 50952263-51028335 on Build GRCh38
Transcripts
NM_000617.2 NM_001174125.1 NM_001174126.1 NM_001174127.1 NM_001174128.1 NM_001174129.1 NM_001174130.1 NR_033421.1 NR_033422.1 XM_005268911.3 XM_005268912.4 XM_011538404.2 XM_011538405.2 XM_017019355.1 XM_017019356.1

SNP Detail

SNP ID Chromosome Location
rs1028196487 Chr.12: 50986611-50986611 on Build GRCh38
rs899143415 Chr.12: 50986580-50986580 on Build GRCh38
rs956658272 Chr.12: 50986434-50986434 on Build GRCh38
rs186583435 Chr.12: 50986619-50986619 on Build GRCh38
rs886049559 Chr.12: 50986726-50986726 on Build GRCh38
rs144968585 Chr.12: 50986742-50986742 on Build GRCh38
rs72561485 Chr.12: 50986376-50986376 on Build GRCh38
rs1023830008 Chr.12: 50986355-50986355 on Build GRCh38
rs967949766 Chr.12: 50986451-50986451 on Build GRCh38
rs1034810700 Chr.12: 50986423-50986423 on Build GRCh38
rs756970075 Chr.12: 50986447-50986447 on Build GRCh38
rs763684336 Chr.12: 50986778-50986778 on Build GRCh38
rs761220255 Chr.12: 50986682-50986682 on Build GRCh38
rs755515112 Chr.12: 50986573-50986573 on Build GRCh38
rs766628921 Chr.12: 50986586-50986586 on Build GRCh38
rs945920509 Chr.12: 50986333-50986333 on Build GRCh38
rs1005518516 Chr.12: 50986632-50986632 on Build GRCh38
rs542347738 Chr.12: 50986751-50986751 on Build GRCh38
rs563773129 Chr.12: 50986354-50986354 on Build GRCh38
rs921160441 Chr.12: 50986526-50986526 on Build GRCh38
rs748156013 Chr.12: 50986597-50986597 on Build GRCh38
rs1045371340 Chr.12: 50986604-50986604 on Build GRCh38
rs751992159 Chr.12: 50986584-50986584 on Build GRCh38
rs899426163 Chr.12: 50986332-50986332 on Build GRCh38
rs149411 Chr.12: 50986449-50986449 on Build GRCh38
rs1036379403 Chr.12: 50986573-50986574 on Build GRCh38
rs537671498 Chr.12: 50986700-50986700 on Build GRCh38
rs750648046 Chr.12: 50986530-50986530 on Build GRCh38
rs763680321 Chr.12: 50986572-50986573 on Build GRCh38
rs762513303 Chr.12: 50986752-50986752 on Build GRCh38
rs59280115 Chr.12: 50986547-50986547 on Build GRCh38
rs755397829 Chr.12: 50986668-50986668 on Build GRCh38
rs755074642 Chr.12: 50986644-50986644 on Build GRCh38
rs377220960 Chr.12: 50986372-50986372 on Build GRCh38
rs557225035 Chr.12: 50986710-50986710 on Build GRCh38
rs760035407 Chr.12: 50986734-50986734 on Build GRCh38
rs773539780 Chr.12: 50986654-50986654 on Build GRCh38
rs975276787 Chr.12: 50986489-50986489 on Build GRCh38
rs144174412 Chr.12: 50986366-50986366 on Build GRCh38
rs777649881 Chr.12: 50986766-50986766 on Build GRCh38
rs578240585 Chr.12: 50986683-50986683 on Build GRCh38
rs772166626 Chr.12: 50986642-50986642 on Build GRCh38
rs528056084 Chr.12: 50986390-50986392 on Build GRCh38
rs796672052 Chr.12: 50986545-50986545 on Build GRCh38
rs886049558 Chr.12: 50986423-50986423 on Build GRCh38
rs777505507 Chr.12: 50986358-50986358 on Build GRCh38
rs375705399 Chr.12: 50986522-50986522 on Build GRCh38
rs971412714 Chr.12: 50986678-50986678 on Build GRCh38
rs914533926 Chr.12: 50986327-50986327 on Build GRCh38
rs191887833 Chr.12: 50986641-50986641 on Build GRCh38

View Full Product Details