Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52014859-52015355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRASP
Entrez Gene ID
160622
Gene Name
general receptor for phosphoinositides 1 associated scaffold protein
Gene Aliases
TAMALIN
UniGene
Hs.407202
Gene Chromosome Location
Chr.12: 52006945-52015889 on Build GRCh38
Transcripts
NM_001271856.1 NM_181711.3 XM_005268691.3 XM_011537996.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6674811 c.1111C>T p.R371C Chr.12: 52015122-52015122 on Build GRCh38
6520272 c.1104C>A p.T368T Chr.12: 52015115-52015115 on Build GRCh38
5681585 c.935C>A p.P312H Chr.12: 52014946-52014946 on Build GRCh38
135589 c.1160C>T p.S387F Chr.12: 52015171-52015171 on Build GRCh38
3462150 c.1161C>T p.S387S Chr.12: 52015172-52015172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937847617 Chr.12: 52014987-52014987 on Build GRCh38
rs751938031 Chr.12: 52015238-52015238 on Build GRCh38
rs767195794 Chr.12: 52015249-52015249 on Build GRCh38
rs765640650 Chr.12: 52015215-52015215 on Build GRCh38
rs781106341 Chr.12: 52015054-52015054 on Build GRCh38
rs749023120 Chr.12: 52015145-52015145 on Build GRCh38
rs771301583 Chr.12: 52015187-52015187 on Build GRCh38
rs757668389 Chr.12: 52015069-52015074 on Build GRCh38
rs746482688 Chr.12: 52015037-52015037 on Build GRCh38
rs905000337 Chr.12: 52014963-52014963 on Build GRCh38
rs747169769 Chr.12: 52015163-52015163 on Build GRCh38
rs139529598 Chr.12: 52015089-52015089 on Build GRCh38
rs775106102 Chr.12: 52015083-52015083 on Build GRCh38
rs745742598 Chr.12: 52015055-52015055 on Build GRCh38
rs954171041 Chr.12: 52015091-52015091 on Build GRCh38
rs1042727758 Chr.12: 52015272-52015272 on Build GRCh38
rs751281622 Chr.12: 52015105-52015105 on Build GRCh38
rs760832588 Chr.12: 52015094-52015094 on Build GRCh38
rs776814897 Chr.12: 52015203-52015203 on Build GRCh38
rs912817276 Chr.12: 52014895-52014895 on Build GRCh38
rs930390417 Chr.12: 52014953-52014953 on Build GRCh38
rs903730105 Chr.12: 52015173-52015173 on Build GRCh38
rs971747132 Chr.12: 52014912-52014912 on Build GRCh38
rs765807414 Chr.12: 52015100-52015100 on Build GRCh38
rs911663683 Chr.12: 52015221-52015221 on Build GRCh38
rs573773880 Chr.12: 52015046-52015046 on Build GRCh38
rs1009933644 Chr.12: 52015016-52015016 on Build GRCh38
rs1000823845 Chr.12: 52015296-52015296 on Build GRCh38
rs756911395 Chr.12: 52015112-52015112 on Build GRCh38
rs944337224 Chr.12: 52014896-52014896 on Build GRCh38
rs749143964 Chr.12: 52015087-52015087 on Build GRCh38
rs541678632 Chr.12: 52015246-52015246 on Build GRCh38
rs750842848 Chr.12: 52015175-52015177 on Build GRCh38
rs921795156 Chr.12: 52014943-52014943 on Build GRCh38
rs745411845 Chr.12: 52015124-52015124 on Build GRCh38
rs764455093 Chr.12: 52015237-52015237 on Build GRCh38
rs997329791 Chr.12: 52015329-52015329 on Build GRCh38
rs961644698 Chr.12: 52015097-52015097 on Build GRCh38
rs973416082 Chr.12: 52015098-52015098 on Build GRCh38
rs945294875 Chr.12: 52015254-52015254 on Build GRCh38
rs200789033 Chr.12: 52015095-52015095 on Build GRCh38
rs780085804 Chr.12: 52015138-52015138 on Build GRCh38
rs1056305803 Chr.12: 52015242-52015242 on Build GRCh38
rs533524051 Chr.12: 52015332-52015332 on Build GRCh38
rs371850904 Chr.12: 52015117-52015117 on Build GRCh38
rs1048868453 Chr.12: 52014957-52014957 on Build GRCh38
rs918954748 Chr.12: 52014920-52014920 on Build GRCh38
rs759729534 Chr.12: 52015213-52015213 on Build GRCh38
rs7974426 Chr.12: 52015104-52015104 on Build GRCh38
rs986936835 Chr.12: 52015093-52015093 on Build GRCh38
rs896413817 Chr.12: 52014992-52014992 on Build GRCh38
rs776126271 Chr.12: 52015216-52015216 on Build GRCh38
rs752973874 Chr.12: 52015101-52015101 on Build GRCh38
rs1016896726 Chr.12: 52015028-52015029 on Build GRCh38
rs564637508 Chr.12: 52015293-52015293 on Build GRCh38
rs1028329972 Chr.12: 52015297-52015297 on Build GRCh38
rs769742423 Chr.12: 52015078-52015078 on Build GRCh38
rs760072380 Chr.12: 52015096-52015096 on Build GRCh38
rs763227859 Chr.12: 52015217-52015217 on Build GRCh38
rs754620705 Chr.12: 52015198-52015199 on Build GRCh38
rs772158859 Chr.12: 52015088-52015088 on Build GRCh38
rs774019783 Chr.12: 52015158-52015158 on Build GRCh38
rs765658157 Chr.12: 52015115-52015116 on Build GRCh38
rs1056359331 Chr.12: 52014988-52014988 on Build GRCh38
rs956537100 Chr.12: 52015311-52015311 on Build GRCh38
rs747606779 Chr.12: 52015223-52015237 on Build GRCh38
rs780903466 Chr.12: 52015221-52015224 on Build GRCh38

View Full Product Details