Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53189395-53189906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF740
Entrez Gene ID
283337
Gene Name
zinc finger protein 740
Gene Aliases
Zfp740
UniGene
Hs.524458
Gene Chromosome Location
Chr.12: 53180715-53190870 on Build GRCh38
Transcripts
NM_001004304.3 XM_006719344.2 XM_011538194.2 XM_017019181.1

SNP Detail

SNP ID Chromosome Location
rs928772521 Chr.12: 53189659-53189659 on Build GRCh38
rs574347454 Chr.12: 53189589-53189589 on Build GRCh38
rs962651179 Chr.12: 53189537-53189537 on Build GRCh38
rs921010641 Chr.12: 53189799-53189799 on Build GRCh38
rs890982175 Chr.12: 53189505-53189506 on Build GRCh38
rs997056554 Chr.12: 53189467-53189467 on Build GRCh38
rs770370212 Chr.12: 53189511-53189514 on Build GRCh38
rs550982609 Chr.12: 53189877-53189877 on Build GRCh38
rs1023608350 Chr.12: 53189588-53189588 on Build GRCh38
rs909731244 Chr.12: 53189842-53189842 on Build GRCh38
rs373662831 Chr.12: 53189765-53189765 on Build GRCh38
rs568386141 Chr.12: 53189822-53189822 on Build GRCh38
rs1036335472 Chr.12: 53189586-53189586 on Build GRCh38
rs932468852 Chr.12: 53189516-53189516 on Build GRCh38
rs769845723 Chr.12: 53189520-53189520 on Build GRCh38
rs1036704968 Chr.12: 53189836-53189836 on Build GRCh38
rs968912274 Chr.12: 53189750-53189750 on Build GRCh38
rs75696435 Chr.12: 53189792-53189792 on Build GRCh38
rs1036953944 Chr.12: 53189871-53189871 on Build GRCh38
rs965424573 Chr.12: 53189781-53189781 on Build GRCh38
rs71443289 Chr.12: 53189482-53189482 on Build GRCh38
rs988893520 Chr.12: 53189713-53189713 on Build GRCh38
rs1029470975 Chr.12: 53189494-53189494 on Build GRCh38
rs34089972 Chr.12: 53189478-53189479 on Build GRCh38
rs780198155 Chr.12: 53189627-53189627 on Build GRCh38
rs898256999 Chr.12: 53189841-53189841 on Build GRCh38
rs939895835 Chr.12: 53189870-53189870 on Build GRCh38
rs919233274 Chr.12: 53189874-53189874 on Build GRCh38
rs73309156 Chr.12: 53189815-53189815 on Build GRCh38
rs953842377 Chr.12: 53189809-53189809 on Build GRCh38
rs563004945 Chr.12: 53189709-53189709 on Build GRCh38
rs1021776023 Chr.12: 53189748-53189748 on Build GRCh38
rs1048297205 Chr.12: 53189519-53189519 on Build GRCh38
rs1015336390 Chr.12: 53189536-53189536 on Build GRCh38
rs367701060 Chr.12: 53189718-53189718 on Build GRCh38
rs1031256258 Chr.12: 53189767-53189767 on Build GRCh38
rs1801641 Chr.12: 53189617-53189617 on Build GRCh38
rs773961117 Chr.12: 53189576-53189576 on Build GRCh38
rs57787354 Chr.12: 53189883-53189883 on Build GRCh38
rs529260407 Chr.12: 53189865-53189865 on Build GRCh38
rs761057319 Chr.12: 53189607-53189607 on Build GRCh38
rs1036193843 Chr.12: 53189652-53189652 on Build GRCh38
rs970352864 Chr.12: 53189616-53189616 on Build GRCh38
rs897838637 Chr.12: 53189622-53189622 on Build GRCh38
rs565276795 Chr.12: 53189784-53189784 on Build GRCh38
rs1004005684 Chr.12: 53189827-53189827 on Build GRCh38
rs369037470 Chr.12: 53189680-53189680 on Build GRCh38
rs947343441 Chr.12: 53189729-53189729 on Build GRCh38
rs539915616 Chr.12: 53189884-53189884 on Build GRCh38
rs981711641 Chr.12: 53189628-53189628 on Build GRCh38
rs545639762 Chr.12: 53189731-53189731 on Build GRCh38
rs3108404 Chr.12: 53189447-53189447 on Build GRCh38
rs571896486 Chr.12: 53189803-53189803 on Build GRCh38
rs939949224 Chr.12: 53189575-53189575 on Build GRCh38
rs921071752 Chr.12: 53189459-53189459 on Build GRCh38

View Full Product Details