Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54373881-54374380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102724050
Entrez Gene ID
102724050
Gene Name
uncharacterized LOC102724050
Gene Aliases
-
UniGene
Hs.670941
Gene Chromosome Location
Chr.12: 54353691-54467030 on Build GRCh38
Transcripts
NR_120486.1 NR_120487.1 NM_001130967.2 NM_001130968.2 NM_001290001.1 NM_001290002.1 NM_001290004.1 NM_015481.2 XM_005268783.4 XM_006719342.2 XM_011538168.2 XM_011538169.2 XM_011538170.1 XM_011538171.2 XM_017019174.1

Gene Symbol
ZNF385A
Entrez Gene ID
25946
Gene Name
zinc finger protein 385A
Gene Aliases
HZF, RZF, ZFP385, ZNF385
UniGene
Hs.505653
Gene Chromosome Location
Chr.12: 54369129-54391609 on Build GRCh38
Transcripts
NR_120486.1 NR_120487.1 NM_001130967.2 NM_001130968.2 NM_001290001.1 NM_001290002.1 NM_001290004.1 NM_015481.2 XM_005268783.4 XM_006719342.2 XM_011538168.2 XM_011538169.2 XM_011538170.1 XM_011538171.2 XM_017019174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1747113 c.208G>A p.E70K Chr.12: 54374126-54374126 on Build GRCh38
5542190 c.307C>T p.P103S Chr.12: 54374027-54374027 on Build GRCh38
5542191 c.307C>T p.P103S Chr.12: 54374027-54374027 on Build GRCh38
1747112 c.208G>A p.E70K Chr.12: 54374126-54374126 on Build GRCh38
5717672 c.270C>T p.T90T Chr.12: 54374064-54374064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200446001 Chr.12: 54373957-54373957 on Build GRCh38
rs748729526 Chr.12: 54373937-54373937 on Build GRCh38
rs577450554 Chr.12: 54374022-54374022 on Build GRCh38
rs765088488 Chr.12: 54373960-54373960 on Build GRCh38
rs758847325 Chr.12: 54374099-54374099 on Build GRCh38
rs755879254 Chr.12: 54374092-54374092 on Build GRCh38
rs373542422 Chr.12: 54374156-54374156 on Build GRCh38
rs764038574 Chr.12: 54373986-54373986 on Build GRCh38
rs1048332652 Chr.12: 54374204-54374204 on Build GRCh38
rs763100927 Chr.12: 54374147-54374147 on Build GRCh38
rs2887875 Chr.12: 54374028-54374028 on Build GRCh38
rs369266291 Chr.12: 54374203-54374203 on Build GRCh38
rs34546338 Chr.12: 54373918-54373918 on Build GRCh38
rs773807053 Chr.12: 54373952-54373952 on Build GRCh38
rs753475854 Chr.12: 54374095-54374095 on Build GRCh38
rs1051561952 Chr.12: 54374310-54374310 on Build GRCh38
rs3832831 Chr.12: 54374197-54374198 on Build GRCh38
rs544481099 Chr.12: 54374058-54374058 on Build GRCh38
rs572437962 Chr.12: 54373995-54373995 on Build GRCh38
rs905856661 Chr.12: 54374348-54374348 on Build GRCh38
rs947501987 Chr.12: 54373914-54373915 on Build GRCh38
rs201286538 Chr.12: 54374045-54374045 on Build GRCh38
rs769128416 Chr.12: 54373944-54373944 on Build GRCh38
rs759793938 Chr.12: 54373958-54373958 on Build GRCh38
rs372451032 Chr.12: 54373956-54373956 on Build GRCh38
rs933874578 Chr.12: 54374042-54374042 on Build GRCh38
rs201064767 Chr.12: 54373942-54373942 on Build GRCh38
rs762764826 Chr.12: 54374071-54374071 on Build GRCh38
rs1004929697 Chr.12: 54373979-54373979 on Build GRCh38
rs769821908 Chr.12: 54374144-54374144 on Build GRCh38
rs76519015 Chr.12: 54374001-54374001 on Build GRCh38
rs929509662 Chr.12: 54374139-54374139 on Build GRCh38
rs774190893 Chr.12: 54374178-54374178 on Build GRCh38
rs770977513 Chr.12: 54374041-54374041 on Build GRCh38
rs773893427 Chr.12: 54374270-54374270 on Build GRCh38
rs199863047 Chr.12: 54374194-54374194 on Build GRCh38
rs761694894 Chr.12: 54374186-54374186 on Build GRCh38
rs376369000 Chr.12: 54374202-54374202 on Build GRCh38
rs925815291 Chr.12: 54373990-54373990 on Build GRCh38
rs747493233 Chr.12: 54374036-54374036 on Build GRCh38
rs141579574 Chr.12: 54373996-54373996 on Build GRCh38
rs751462100 Chr.12: 54374082-54374082 on Build GRCh38
rs144343816 Chr.12: 54374100-54374100 on Build GRCh38
rs375747147 Chr.12: 54374052-54374052 on Build GRCh38
rs773862097 Chr.12: 54374030-54374030 on Build GRCh38
rs369737303 Chr.12: 54374093-54374093 on Build GRCh38
rs759719267 Chr.12: 54374046-54374046 on Build GRCh38
rs138539214 Chr.12: 54374025-54374025 on Build GRCh38
rs141944873 Chr.12: 54373984-54373984 on Build GRCh38
rs747539533 Chr.12: 54374110-54374110 on Build GRCh38
rs386376589 Chr.12: 54374200-54374201 on Build GRCh38
rs745563565 Chr.12: 54374014-54374014 on Build GRCh38
rs752460065 Chr.12: 54373978-54373978 on Build GRCh38
rs1014476636 Chr.12: 54374338-54374338 on Build GRCh38
rs994183122 Chr.12: 54373910-54373910 on Build GRCh38
rs750277898 Chr.12: 54374187-54374187 on Build GRCh38
rs137955935 Chr.12: 54374079-54374079 on Build GRCh38
rs1000719261 Chr.12: 54374352-54374352 on Build GRCh38
rs933124637 Chr.12: 54374255-54374255 on Build GRCh38
rs775690253 Chr.12: 54374146-54374146 on Build GRCh38
rs761796622 Chr.12: 54374083-54374083 on Build GRCh38
rs989025288 Chr.12: 54374010-54374010 on Build GRCh38
rs1023012665 Chr.12: 54374344-54374344 on Build GRCh38
rs1038064840 Chr.12: 54374349-54374349 on Build GRCh38
rs74721071 Chr.12: 54374197-54374197 on Build GRCh38
rs969161326 Chr.12: 54374112-54374112 on Build GRCh38
rs3034352 Chr.12: 54374198-54374199 on Build GRCh38
rs755786085 Chr.12: 54374023-54374023 on Build GRCh38
rs772526402 Chr.12: 54374029-54374029 on Build GRCh38
rs148342828 Chr.12: 54374121-54374121 on Build GRCh38
rs772092279 Chr.12: 54374131-54374131 on Build GRCh38
rs372987947 Chr.12: 54374048-54374048 on Build GRCh38
rs376501983 Chr.12: 54373925-54373925 on Build GRCh38
rs199558604 Chr.12: 54373970-54373970 on Build GRCh38

View Full Product Details