Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 91158114-91158614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCN
Entrez Gene ID
1634
Gene Name
decorin
Gene Aliases
CSCD, DSPG2, PG40, PGII, PGS2, SLRR1B
UniGene
Hs.156316
Gene Chromosome Location
Chr.12: 91143277-91183124 on Build GRCh38
Transcripts
NM_001920.4 NM_133503.3 NM_133504.3 NM_133505.3 NM_133506.3 NM_133507.3 XM_005268693.1 XM_006719270.1 XM_017018917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
42740 c.397C>T p.R133* Chr.12: 91158437-91158437 on Build GRCh38
5964858 c.410C>G p.S137C Chr.12: 91158424-91158424 on Build GRCh38
7204661 c.448A>C p.K150Q Chr.12: 91158386-91158386 on Build GRCh38
4045361 c.430T>A p.L144M Chr.12: 91158404-91158404 on Build GRCh38
4045360 c.497G>A p.R166Q Chr.12: 91158337-91158337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929581369 Chr.12: 91158444-91158444 on Build GRCh38
rs762693864 Chr.12: 91158431-91158431 on Build GRCh38
rs755987970 Chr.12: 91158463-91158463 on Build GRCh38
rs574786139 Chr.12: 91158211-91158211 on Build GRCh38
rs971671690 Chr.12: 91158260-91158260 on Build GRCh38
rs779618769 Chr.12: 91158337-91158337 on Build GRCh38
rs915287751 Chr.12: 91158387-91158387 on Build GRCh38
rs750460433 Chr.12: 91158561-91158562 on Build GRCh38
rs1018127669 Chr.12: 91158161-91158161 on Build GRCh38
rs367777271 Chr.12: 91158261-91158261 on Build GRCh38
rs771153948 Chr.12: 91158368-91158368 on Build GRCh38
rs774468561 Chr.12: 91158282-91158282 on Build GRCh38
rs746492042 Chr.12: 91158341-91158341 on Build GRCh38
rs111244923 Chr.12: 91158510-91158510 on Build GRCh38
rs776646201 Chr.12: 91158408-91158408 on Build GRCh38
rs143385640 Chr.12: 91158438-91158438 on Build GRCh38
rs748665361 Chr.12: 91158350-91158350 on Build GRCh38
rs775738773 Chr.12: 91158524-91158524 on Build GRCh38
rs939390141 Chr.12: 91158485-91158485 on Build GRCh38
rs543407003 Chr.12: 91158223-91158223 on Build GRCh38
rs753885997 Chr.12: 91158316-91158316 on Build GRCh38
rs781652785 Chr.12: 91158346-91158346 on Build GRCh38
rs1029551464 Chr.12: 91158215-91158215 on Build GRCh38
rs199897276 Chr.12: 91158393-91158393 on Build GRCh38
rs201128202 Chr.12: 91158213-91158214 on Build GRCh38
rs372382508 Chr.12: 91158344-91158344 on Build GRCh38
rs772241441 Chr.12: 91158511-91158511 on Build GRCh38
rs1039109019 Chr.12: 91158465-91158465 on Build GRCh38
rs1025929252 Chr.12: 91158303-91158303 on Build GRCh38
rs762793547 Chr.12: 91158558-91158558 on Build GRCh38
rs1047858579 Chr.12: 91158455-91158455 on Build GRCh38
rs758094819 Chr.12: 91158332-91158332 on Build GRCh38
rs867516048 Chr.12: 91158382-91158382 on Build GRCh38
rs895285516 Chr.12: 91158157-91158157 on Build GRCh38
rs201750755 Chr.12: 91158375-91158375 on Build GRCh38
rs770075410 Chr.12: 91158355-91158355 on Build GRCh38
rs779139285 Chr.12: 91158501-91158501 on Build GRCh38
rs989333042 Chr.12: 91158347-91158347 on Build GRCh38
rs771259588 Chr.12: 91158488-91158488 on Build GRCh38
rs201430261 Chr.12: 91158305-91158305 on Build GRCh38
rs773085500 Chr.12: 91158550-91158550 on Build GRCh38
rs1005747170 Chr.12: 91158185-91158185 on Build GRCh38
rs375774029 Chr.12: 91158462-91158462 on Build GRCh38
rs772292920 Chr.12: 91158401-91158401 on Build GRCh38
rs866641462 Chr.12: 91158338-91158338 on Build GRCh38
rs765355380 Chr.12: 91158269-91158269 on Build GRCh38
rs142752401 Chr.12: 91158410-91158410 on Build GRCh38
rs372172875 Chr.12: 91158557-91158557 on Build GRCh38
rs759575808 Chr.12: 91158288-91158288 on Build GRCh38
rs750064319 Chr.12: 91158329-91158329 on Build GRCh38
rs780683499 Chr.12: 91158454-91158454 on Build GRCh38
rs765079976 Chr.12: 91158423-91158423 on Build GRCh38
rs749660390 Chr.12: 91158483-91158483 on Build GRCh38
rs754450177 Chr.12: 91158440-91158440 on Build GRCh38
rs563844840 Chr.12: 91158277-91158277 on Build GRCh38
rs963115740 Chr.12: 91158234-91158234 on Build GRCh38
rs765131074 Chr.12: 91158328-91158328 on Build GRCh38
rs12309644 Chr.12: 91158206-91158206 on Build GRCh38
rs746085883 Chr.12: 91158507-91158507 on Build GRCh38
rs766045934 Chr.12: 91158436-91158436 on Build GRCh38
rs756635987 Chr.12: 91158468-91158468 on Build GRCh38
rs114922731 Chr.12: 91158535-91158535 on Build GRCh38
rs375558672 Chr.12: 91158324-91158324 on Build GRCh38
rs778293007 Chr.12: 91158479-91158479 on Build GRCh38
rs367942727 Chr.12: 91158533-91158533 on Build GRCh38
rs200754930 Chr.12: 91158367-91158367 on Build GRCh38
rs773363001 Chr.12: 91158263-91158263 on Build GRCh38
rs867322719 Chr.12: 91158172-91158172 on Build GRCh38

View Full Product Details