Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109097442-109097950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UNG
Entrez Gene ID
7374
Gene Name
uracil DNA glycosylase
Gene Aliases
DGU, HIGM4, HIGM5, UDG, UNG1, UNG15, UNG2
UniGene
Hs.191334
Gene Chromosome Location
Chr.12: 109097594-109110993 on Build GRCh38
Transcripts
NM_080911.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4404987 c.44C>T p.P15L Chr.12: 109097723-109097723 on Build GRCh38
4941374 c.15G>A p.K5K Chr.12: 109097694-109097694 on Build GRCh38
6553666 c.56G>T p.R19L Chr.12: 109097735-109097735 on Build GRCh38
4998075 c.46G>T p.A16S Chr.12: 109097725-109097725 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774385200 Chr.12: 109097743-109097743 on Build GRCh38
rs73413093 Chr.12: 109097574-109097574 on Build GRCh38
rs775145036 Chr.12: 109097842-109097842 on Build GRCh38
rs979330126 Chr.12: 109097575-109097575 on Build GRCh38
rs770528717 Chr.12: 109097819-109097819 on Build GRCh38
rs755512741 Chr.12: 109097781-109097781 on Build GRCh38
rs775774203 Chr.12: 109097862-109097862 on Build GRCh38
rs982865574 Chr.12: 109097859-109097859 on Build GRCh38
rs769585330 Chr.12: 109097563-109097563 on Build GRCh38
rs982280015 Chr.12: 109097540-109097540 on Build GRCh38
rs556699615 Chr.12: 109097903-109097903 on Build GRCh38
rs967404289 Chr.12: 109097653-109097653 on Build GRCh38
rs768853561 Chr.12: 109097672-109097672 on Build GRCh38
rs538018097 Chr.12: 109097544-109097544 on Build GRCh38
rs750560113 Chr.12: 109097663-109097663 on Build GRCh38
rs377203641 Chr.12: 109097803-109097803 on Build GRCh38
rs539054379 Chr.12: 109097623-109097623 on Build GRCh38
rs572847021 Chr.12: 109097717-109097717 on Build GRCh38
rs998107697 Chr.12: 109097609-109097609 on Build GRCh38
rs1056345717 Chr.12: 109097925-109097925 on Build GRCh38
rs747684225 Chr.12: 109097711-109097711 on Build GRCh38
rs369677873 Chr.12: 109097666-109097666 on Build GRCh38
rs941088015 Chr.12: 109097651-109097651 on Build GRCh38
rs1045024911 Chr.12: 109097491-109097491 on Build GRCh38
rs567830116 Chr.12: 109097541-109097541 on Build GRCh38
rs527980794 Chr.12: 109097493-109097493 on Build GRCh38
rs1050916118 Chr.12: 109097610-109097610 on Build GRCh38
rs765512122 Chr.12: 109097765-109097765 on Build GRCh38
rs775119761 Chr.12: 109097593-109097593 on Build GRCh38
rs775369845 Chr.12: 109097756-109097756 on Build GRCh38
rs886048905 Chr.12: 109097685-109097685 on Build GRCh38
rs886048904 Chr.12: 109097607-109097607 on Build GRCh38
rs942132340 Chr.12: 109097467-109097494 on Build GRCh38
rs941190705 Chr.12: 109097469-109097469 on Build GRCh38
rs897095209 Chr.12: 109097608-109097613 on Build GRCh38
rs1012971028 Chr.12: 109097843-109097843 on Build GRCh38
rs895789870 Chr.12: 109097515-109097515 on Build GRCh38
rs781349708 Chr.12: 109097784-109097784 on Build GRCh38
rs372022075 Chr.12: 109097805-109097805 on Build GRCh38
rs1009949461 Chr.12: 109097916-109097916 on Build GRCh38
rs1020766905 Chr.12: 109097530-109097530 on Build GRCh38
rs1029919102 Chr.12: 109097851-109097851 on Build GRCh38
rs573796572 Chr.12: 109097882-109097882 on Build GRCh38
rs7488798 Chr.12: 109097690-109097690 on Build GRCh38
rs372914375 Chr.12: 109097645-109097645 on Build GRCh38
rs777606593 Chr.12: 109097806-109097806 on Build GRCh38
rs939168204 Chr.12: 109097916-109097916 on Build GRCh38
rs773920326 Chr.12: 109097820-109097820 on Build GRCh38
rs750472259 Chr.12: 109097766-109097766 on Build GRCh38
rs909513284 Chr.12: 109097901-109097901 on Build GRCh38
rs761928219 Chr.12: 109097760-109097760 on Build GRCh38
rs772832695 Chr.12: 109097715-109097715 on Build GRCh38
rs573597388 Chr.12: 109097541-109097542 on Build GRCh38
rs748974541 Chr.12: 109097812-109097812 on Build GRCh38
rs976974524 Chr.12: 109097621-109097621 on Build GRCh38
rs748872362 Chr.12: 109097718-109097718 on Build GRCh38
rs894993490 Chr.12: 109097786-109097786 on Build GRCh38
rs375712849 Chr.12: 109097629-109097629 on Build GRCh38
rs377075959 Chr.12: 109097662-109097662 on Build GRCh38
rs939282895 Chr.12: 109097724-109097724 on Build GRCh38
rs769496984 Chr.12: 109097713-109097713 on Build GRCh38
rs370117219 Chr.12: 109097630-109097630 on Build GRCh38
rs368157435 Chr.12: 109097516-109097516 on Build GRCh38
rs918596003 Chr.12: 109097788-109097788 on Build GRCh38
rs926485159 Chr.12: 109097912-109097912 on Build GRCh38
rs964291842 Chr.12: 109097607-109097607 on Build GRCh38
rs762652189 Chr.12: 109097628-109097628 on Build GRCh38
rs941336481 Chr.12: 109097567-109097567 on Build GRCh38
rs990883898 Chr.12: 109097696-109097696 on Build GRCh38
rs980515453 Chr.12: 109097904-109097904 on Build GRCh38
rs868596879 Chr.12: 109097689-109097689 on Build GRCh38
rs1035670118 Chr.12: 109097503-109097503 on Build GRCh38
rs550280569 Chr.12: 109097562-109097562 on Build GRCh38
rs578115571 Chr.12: 109097918-109097918 on Build GRCh38
rs928133605 Chr.12: 109097542-109097542 on Build GRCh38
rs549361716 Chr.12: 109097504-109097504 on Build GRCh38
rs767154683 Chr.12: 109097749-109097749 on Build GRCh38
rs1010345666 Chr.12: 109097644-109097644 on Build GRCh38
rs745739198 Chr.12: 109097839-109097839 on Build GRCh38
rs759301871 Chr.12: 109097641-109097641 on Build GRCh38
rs373668102 Chr.12: 109097744-109097744 on Build GRCh38
rs544049038 Chr.12: 109097884-109097884 on Build GRCh38
rs1054275430 Chr.12: 109097893-109097893 on Build GRCh38
rs998462991 Chr.12: 109097848-109097848 on Build GRCh38
rs886048906 Chr.12: 109097779-109097779 on Build GRCh38
rs945634412 Chr.12: 109097521-109097521 on Build GRCh38
rs909897963 Chr.12: 109097564-109097564 on Build GRCh38
rs867171362 Chr.12: 109097673-109097673 on Build GRCh38
rs989765313 Chr.12: 109097494-109097494 on Build GRCh38
rs770972381 Chr.12: 109097739-109097739 on Build GRCh38
rs747375477 Chr.12: 109097671-109097671 on Build GRCh38
rs760560169 Chr.12: 109097858-109097858 on Build GRCh38
rs938100668 Chr.12: 109097546-109097546 on Build GRCh38
rs904612774 Chr.12: 109097772-109097772 on Build GRCh38
rs534071307 Chr.12: 109097768-109097768 on Build GRCh38
rs780581104 Chr.12: 109097668-109097668 on Build GRCh38
rs959504476 Chr.12: 109097685-109097685 on Build GRCh38
rs751970160 Chr.12: 109097776-109097776 on Build GRCh38
rs781643146 Chr.12: 109097686-109097686 on Build GRCh38
rs1018876048 Chr.12: 109097810-109097810 on Build GRCh38
rs1003378684 Chr.12: 109097495-109097495 on Build GRCh38
rs767370738 Chr.12: 109097642-109097642 on Build GRCh38
rs766467977 Chr.12: 109097769-109097769 on Build GRCh38

View Full Product Details