Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 48344897-48345404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RB1
Entrez Gene ID
5925
Gene Name
RB transcriptional corepressor 1
Gene Aliases
OSRC, PPP1R130, RB, p105-Rb, pRb, pp110
UniGene
Hs.408528
Gene Chromosome Location
Chr.13: 48303747-48481890 on Build GRCh38
Transcripts
NM_000321.2 XM_011535171.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6438178 c.416A>T p.D139V Chr.13: 48345115-48345115 on Build GRCh38
3936342 c.452T>C p.L151P Chr.13: 48345151-48345151 on Build GRCh38
13413 c.473T>A p.L158* Chr.13: 48345172-48345172 on Build GRCh38
7091266 c.427_428delAA p.K143fs*2 Chr.13: 48345126-48345127 on Build GRCh38
6942233 c.488G>A p.S163N Chr.13: 48345187-48345187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377666789 Chr.13: 48345061-48345061 on Build GRCh38
rs746722035 Chr.13: 48345068-48345068 on Build GRCh38
rs3092899 Chr.13: 48344928-48344928 on Build GRCh38
rs762380973 Chr.13: 48345094-48345094 on Build GRCh38
rs749495284 Chr.13: 48345092-48345092 on Build GRCh38
rs3092901 Chr.13: 48345245-48345245 on Build GRCh38
rs895447213 Chr.13: 48345148-48345148 on Build GRCh38
rs3092902 Chr.13: 48345110-48345110 on Build GRCh38
rs751509066 Chr.13: 48345242-48345242 on Build GRCh38
rs531145220 Chr.13: 48345289-48345289 on Build GRCh38
rs3092900 Chr.13: 48345096-48345096 on Build GRCh38
rs587778870 Chr.13: 48345145-48345145 on Build GRCh38
rs4151455 Chr.13: 48345022-48345022 on Build GRCh38
rs763870325 Chr.13: 48345238-48345238 on Build GRCh38
rs4151456 Chr.13: 48345370-48345370 on Build GRCh38
rs926022870 Chr.13: 48345309-48345309 on Build GRCh38
rs869041987 Chr.13: 48345109-48345110 on Build GRCh38
rs773642334 Chr.13: 48345158-48345158 on Build GRCh38
rs764881599 Chr.13: 48345184-48345184 on Build GRCh38
rs936492093 Chr.13: 48345316-48345316 on Build GRCh38
rs34349663 Chr.13: 48344936-48344936 on Build GRCh38
rs369830657 Chr.13: 48345161-48345161 on Build GRCh38
rs953677695 Chr.13: 48345296-48345296 on Build GRCh38
rs541168997 Chr.13: 48345021-48345021 on Build GRCh38
rs557707018 Chr.13: 48344979-48344979 on Build GRCh38
rs752732091 Chr.13: 48345033-48345033 on Build GRCh38
rs755435269 Chr.13: 48345339-48345339 on Build GRCh38
rs766849295 Chr.13: 48345163-48345163 on Build GRCh38
rs748717860 Chr.13: 48345141-48345141 on Build GRCh38
rs766970080 Chr.13: 48345248-48345248 on Build GRCh38
rs758239290 Chr.13: 48345034-48345034 on Build GRCh38
rs752335969 Chr.13: 48345055-48345055 on Build GRCh38
rs193228367 Chr.13: 48345281-48345281 on Build GRCh38
rs765277265 Chr.13: 48345098-48345098 on Build GRCh38
rs900376429 Chr.13: 48344952-48344952 on Build GRCh38
rs780321508 Chr.13: 48345080-48345080 on Build GRCh38
rs1034616967 Chr.13: 48345165-48345165 on Build GRCh38
rs1030380584 Chr.13: 48345007-48345007 on Build GRCh38
rs915319922 Chr.13: 48345019-48345019 on Build GRCh38
rs570661443 Chr.13: 48344986-48344986 on Build GRCh38
rs535640785 Chr.13: 48345373-48345373 on Build GRCh38
rs777644802 Chr.13: 48345047-48345047 on Build GRCh38
rs198617 Chr.13: 48345222-48345222 on Build GRCh38
rs886043313 Chr.13: 48345095-48345095 on Build GRCh38
rs539131083 Chr.13: 48345011-48345011 on Build GRCh38
rs967359789 Chr.13: 48345251-48345251 on Build GRCh38
rs978874505 Chr.13: 48345268-48345268 on Build GRCh38
rs756363252 Chr.13: 48345069-48345069 on Build GRCh38
rs121913296 Chr.13: 48345108-48345108 on Build GRCh38
rs587778841 Chr.13: 48345137-48345140 on Build GRCh38
rs587778854 Chr.13: 48345162-48345163 on Build GRCh38
rs963149858 Chr.13: 48345230-48345230 on Build GRCh38
rs869188824 Chr.13: 48345192-48345192 on Build GRCh38
rs758368346 Chr.13: 48345225-48345225 on Build GRCh38
rs869136562 Chr.13: 48344997-48344997 on Build GRCh38
rs376166040 Chr.13: 48345030-48345030 on Build GRCh38
rs754234456 Chr.13: 48345335-48345335 on Build GRCh38
rs768768956 Chr.13: 48345095-48345095 on Build GRCh38
rs1051667441 Chr.13: 48345381-48345381 on Build GRCh38
rs779452279 Chr.13: 48345365-48345366 on Build GRCh38
rs1039243676 Chr.13: 48345336-48345336 on Build GRCh38
rs776484051 Chr.13: 48345179-48345179 on Build GRCh38

View Full Product Details