Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 33055826-33056338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPAS3
Entrez Gene ID
64067
Gene Name
neuronal PAS domain protein 3
Gene Aliases
MOP6, PASD6, bHLHe12
UniGene
Hs.657892
Gene Chromosome Location
Chr.14: 32934785-33804176 on Build GRCh38
Transcripts
NM_001164749.1 NM_001165893.1 NM_022123.2 NM_173159.2 XM_005267991.3 XM_005267992.3 XM_011537067.2 XM_011537069.2 XM_011537070.2 XM_011537071.2 XM_011537072.2 XM_017021582.1 XM_017021583.1 XM_017021584.1 XM_017021585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5027848 c.140A>G p.N47S Chr.14: 33055994-33055994 on Build GRCh38
1587389 c.96A>C p.K32N Chr.14: 33055950-33055950 on Build GRCh38
5691228 c.124G>C p.E42Q Chr.14: 33055978-33055978 on Build GRCh38
4560853 c.85G>A p.E29K Chr.14: 33055939-33055939 on Build GRCh38
4245394 c.72G>A p.L24L Chr.14: 33055926-33055926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371179515 Chr.14: 33056171-33056171 on Build GRCh38
rs892065947 Chr.14: 33055872-33055872 on Build GRCh38
rs951381001 Chr.14: 33056120-33056121 on Build GRCh38
rs921953249 Chr.14: 33056290-33056290 on Build GRCh38
rs374977032 Chr.14: 33056304-33056305 on Build GRCh38
rs117917291 Chr.14: 33056227-33056227 on Build GRCh38
rs868716514 Chr.14: 33055966-33055966 on Build GRCh38
rs145614663 Chr.14: 33056186-33056186 on Build GRCh38
rs148815320 Chr.14: 33056312-33056312 on Build GRCh38
rs976088399 Chr.14: 33056260-33056260 on Build GRCh38
rs966371793 Chr.14: 33056254-33056254 on Build GRCh38
rs1043625152 Chr.14: 33055915-33055915 on Build GRCh38
rs951899759 Chr.14: 33056230-33056230 on Build GRCh38
rs12884210 Chr.14: 33056122-33056122 on Build GRCh38
rs201387871 Chr.14: 33056111-33056111 on Build GRCh38
rs866828179 Chr.14: 33055913-33055913 on Build GRCh38
rs1004722841 Chr.14: 33056240-33056240 on Build GRCh38
rs542915721 Chr.14: 33056202-33056202 on Build GRCh38
rs953479180 Chr.14: 33056296-33056296 on Build GRCh38
rs145473872 Chr.14: 33056216-33056216 on Build GRCh38
rs565261611 Chr.14: 33056245-33056245 on Build GRCh38
rs904569830 Chr.14: 33055933-33055933 on Build GRCh38
rs905595380 Chr.14: 33056220-33056220 on Build GRCh38
rs975161776 Chr.14: 33056138-33056138 on Build GRCh38
rs766280068 Chr.14: 33055959-33055959 on Build GRCh38
rs372807918 Chr.14: 33056120-33056121 on Build GRCh38
rs185418496 Chr.14: 33056241-33056241 on Build GRCh38
rs1043884373 Chr.14: 33056125-33056125 on Build GRCh38
rs764367307 Chr.14: 33056015-33056015 on Build GRCh38
rs963821173 Chr.14: 33056137-33056141 on Build GRCh38
rs753362901 Chr.14: 33056238-33056238 on Build GRCh38
rs933635836 Chr.14: 33056173-33056173 on Build GRCh38
rs1014228075 Chr.14: 33056266-33056266 on Build GRCh38
rs765057734 Chr.14: 33055860-33055860 on Build GRCh38
rs878985044 Chr.14: 33055892-33055892 on Build GRCh38
rs773174627 Chr.14: 33055864-33055864 on Build GRCh38
rs752794265 Chr.14: 33055962-33055962 on Build GRCh38
rs1027512853 Chr.14: 33055914-33055914 on Build GRCh38
rs184013307 Chr.14: 33055965-33055965 on Build GRCh38
rs35719018 Chr.14: 33056112-33056113 on Build GRCh38
rs979936010 Chr.14: 33055901-33055901 on Build GRCh38
rs1027483097 Chr.14: 33056229-33056229 on Build GRCh38
rs966034035 Chr.14: 33056053-33056053 on Build GRCh38
rs907873842 Chr.14: 33056048-33056048 on Build GRCh38
rs951930399 Chr.14: 33055943-33055943 on Build GRCh38
rs547314956 Chr.14: 33056313-33056313 on Build GRCh38
rs935582038 Chr.14: 33056180-33056180 on Build GRCh38
rs377486631 Chr.14: 33056119-33056121 on Build GRCh38
rs970224641 Chr.14: 33055886-33055886 on Build GRCh38
rs976499676 Chr.14: 33056067-33056067 on Build GRCh38
rs180858745 Chr.14: 33056187-33056187 on Build GRCh38
rs1025651213 Chr.14: 33056278-33056278 on Build GRCh38
rs188826465 Chr.14: 33056071-33056071 on Build GRCh38
rs71448285 Chr.14: 33056111-33056112 on Build GRCh38
rs762900592 Chr.14: 33055897-33055897 on Build GRCh38
rs983929284 Chr.14: 33056120-33056120 on Build GRCh38
rs11412096 Chr.14: 33056121-33056121 on Build GRCh38
rs753990119 Chr.14: 33056034-33056034 on Build GRCh38

View Full Product Details