Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 36516684-36517190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKX2-1
Entrez Gene ID
7080
Gene Name
NK2 homeobox 1
Gene Aliases
BCH, BHC, NK-2, NKX2.1, NKX2A, NMTC1, T/EBP, TEBP, TITF1, TTF-1, TTF1
UniGene
Hs.94367
Gene Chromosome Location
Chr.14: 36516397-36520225 on Build GRCh38
Transcripts
NM_001079668.2 NM_003317.3

SNP Detail

SNP ID Chromosome Location
rs752548356 Chr.14: 36516910-36516910 on Build GRCh38
rs886070037 Chr.14: 36517052-36517052 on Build GRCh38
rs918090068 Chr.14: 36516970-36516970 on Build GRCh38
rs77021012 Chr.14: 36517101-36517101 on Build GRCh38
rs545347436 Chr.14: 36517067-36517067 on Build GRCh38
rs79849967 Chr.14: 36516842-36516842 on Build GRCh38
rs564521320 Chr.14: 36517135-36517135 on Build GRCh38
rs935411972 Chr.14: 36516965-36516965 on Build GRCh38
rs1030204810 Chr.14: 36517074-36517074 on Build GRCh38
rs1000909186 Chr.14: 36517161-36517161 on Build GRCh38
rs930364005 Chr.14: 36517039-36517039 on Build GRCh38
rs200848675 Chr.14: 36517099-36517102 on Build GRCh38
rs141117763 Chr.14: 36517091-36517092 on Build GRCh38
rs531107509 Chr.14: 36517085-36517085 on Build GRCh38
rs886050480 Chr.14: 36517067-36517068 on Build GRCh38
rs948201959 Chr.14: 36516987-36516987 on Build GRCh38
rs570771635 Chr.14: 36516721-36516721 on Build GRCh38
rs865938094 Chr.14: 36516921-36516921 on Build GRCh38
rs894564701 Chr.14: 36516975-36516975 on Build GRCh38
rs879194477 Chr.14: 36517100-36517100 on Build GRCh38
rs898722223 Chr.14: 36517036-36517036 on Build GRCh38
rs778795444 Chr.14: 36516718-36516718 on Build GRCh38
rs140427692 Chr.14: 36516985-36516986 on Build GRCh38
rs76977831 Chr.14: 36517102-36517102 on Build GRCh38
rs572546897 Chr.14: 36517069-36517069 on Build GRCh38
rs1045335700 Chr.14: 36517030-36517030 on Build GRCh38
rs543274140 Chr.14: 36516900-36516900 on Build GRCh38
rs754698730 Chr.14: 36516724-36516724 on Build GRCh38
rs533460253 Chr.14: 36517136-36517136 on Build GRCh38
rs758339066 Chr.14: 36516945-36516945 on Build GRCh38
rs200610971 Chr.14: 36517093-36517094 on Build GRCh38
rs762731260 Chr.14: 36516816-36516817 on Build GRCh38
rs878959696 Chr.14: 36516818-36516819 on Build GRCh38
rs574552862 Chr.14: 36516886-36516886 on Build GRCh38
rs562274315 Chr.14: 36517080-36517080 on Build GRCh38
rs10139625 Chr.14: 36517068-36517068 on Build GRCh38
rs112799458 Chr.14: 36516817-36516818 on Build GRCh38
rs998761509 Chr.14: 36517055-36517055 on Build GRCh38
rs1055933413 Chr.14: 36516934-36516934 on Build GRCh38
rs11541294 Chr.14: 36516801-36516801 on Build GRCh38
rs200871832 Chr.14: 36517094-36517095 on Build GRCh38
rs1018681554 Chr.14: 36517094-36517094 on Build GRCh38
rs542496269 Chr.14: 36517078-36517078 on Build GRCh38
rs71124756 Chr.14: 36517060-36517061 on Build GRCh38
rs994884781 Chr.14: 36516707-36516707 on Build GRCh38
rs576538982 Chr.14: 36516967-36516967 on Build GRCh38
rs974675029 Chr.14: 36516838-36516838 on Build GRCh38
rs5807883 Chr.14: 36516818-36516818 on Build GRCh38
rs886050479 Chr.14: 36516808-36516808 on Build GRCh38
rs918902794 Chr.14: 36516796-36516796 on Build GRCh38
rs924090033 Chr.14: 36516940-36516940 on Build GRCh38
rs1004750813 Chr.14: 36516756-36516756 on Build GRCh38
rs965829272 Chr.14: 36516836-36516836 on Build GRCh38
rs550912157 Chr.14: 36517099-36517099 on Build GRCh38
rs377039468 Chr.14: 36517128-36517128 on Build GRCh38
rs879228357 Chr.14: 36516818-36516819 on Build GRCh38
rs1049195720 Chr.14: 36517041-36517041 on Build GRCh38
rs1018796402 Chr.14: 36516782-36516782 on Build GRCh38
rs930236894 Chr.14: 36516810-36516810 on Build GRCh38
rs1027426616 Chr.14: 36516719-36516719 on Build GRCh38
rs886050478 Chr.14: 36516734-36516734 on Build GRCh38
rs1028934089 Chr.14: 36517127-36517127 on Build GRCh38
rs886184567 Chr.14: 36516749-36516749 on Build GRCh38
rs144177561 Chr.14: 36516904-36516904 on Build GRCh38

View Full Product Details