Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50591393-50591910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL1
Entrez Gene ID
51062
Gene Name
atlastin GTPase 1
Gene Aliases
AD-FSP, FSP1, GBP3, HSN1D, SPG3, SPG3A, atlastin1
UniGene
Hs.584905
Gene Chromosome Location
Chr.14: 50533082-50633068 on Build GRCh38
Transcripts
NM_001127713.1 NM_015915.4 NM_181598.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6075742 c.445G>A p.G149R Chr.14: 50591562-50591562 on Build GRCh38
4549382 c.471G>A p.L157L Chr.14: 50591588-50591588 on Build GRCh38
3814870 c.453T>C p.F151F Chr.14: 50591570-50591570 on Build GRCh38
4997238 c.424delG p.V142fs*3 Chr.14: 50591541-50591541 on Build GRCh38
279032 c.437A>G p.D146G Chr.14: 50591554-50591554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947333729 Chr.14: 50591782-50591782 on Build GRCh38
rs546750506 Chr.14: 50591710-50591710 on Build GRCh38
rs119476051 Chr.14: 50591587-50591587 on Build GRCh38
rs2934684 Chr.14: 50591516-50591516 on Build GRCh38
rs756989898 Chr.14: 50591659-50591661 on Build GRCh38
rs752643685 Chr.14: 50591514-50591514 on Build GRCh38
rs763396468 Chr.14: 50591666-50591666 on Build GRCh38
rs756910684 Chr.14: 50591463-50591463 on Build GRCh38
rs773825245 Chr.14: 50591665-50591665 on Build GRCh38
rs756976549 Chr.14: 50591561-50591561 on Build GRCh38
rs956545653 Chr.14: 50591797-50591798 on Build GRCh38
rs992098994 Chr.14: 50591817-50591817 on Build GRCh38
rs137852657 Chr.14: 50591584-50591584 on Build GRCh38
rs187545226 Chr.14: 50591734-50591734 on Build GRCh38
rs773772176 Chr.14: 50591502-50591502 on Build GRCh38
rs774908012 Chr.14: 50591781-50591781 on Build GRCh38
rs774056032 Chr.14: 50591685-50591685 on Build GRCh38
rs537044741 Chr.14: 50591795-50591795 on Build GRCh38
rs778224085 Chr.14: 50591677-50591678 on Build GRCh38
rs149031604 Chr.14: 50591557-50591557 on Build GRCh38
rs544702867 Chr.14: 50591444-50591444 on Build GRCh38
rs759052054 Chr.14: 50591691-50591691 on Build GRCh38
rs780349587 Chr.14: 50591579-50591579 on Build GRCh38
rs142257161 Chr.14: 50591450-50591450 on Build GRCh38
rs191641958 Chr.14: 50591774-50591774 on Build GRCh38
rs996306004 Chr.14: 50591720-50591720 on Build GRCh38
rs766612106 Chr.14: 50591673-50591673 on Build GRCh38
rs1024810905 Chr.14: 50591820-50591820 on Build GRCh38
rs575423341 Chr.14: 50591503-50591503 on Build GRCh38
rs908053602 Chr.14: 50591616-50591616 on Build GRCh38
rs146243252 Chr.14: 50591696-50591696 on Build GRCh38
rs952431392 Chr.14: 50591547-50591547 on Build GRCh38
rs373950115 Chr.14: 50591512-50591512 on Build GRCh38
rs1018925869 Chr.14: 50591492-50591492 on Build GRCh38
rs2934683 Chr.14: 50591793-50591793 on Build GRCh38
rs748226901 Chr.14: 50591658-50591658 on Build GRCh38
rs770490028 Chr.14: 50591663-50591663 on Build GRCh38
rs886041897 Chr.14: 50591595-50591595 on Build GRCh38
rs370173293 Chr.14: 50591600-50591600 on Build GRCh38
rs373591459 Chr.14: 50591621-50591621 on Build GRCh38
rs1040462471 Chr.14: 50591846-50591846 on Build GRCh38
rs777680642 Chr.14: 50591533-50591533 on Build GRCh38
rs935378447 Chr.14: 50591823-50591823 on Build GRCh38
rs576856370 Chr.14: 50591729-50591729 on Build GRCh38
rs369764400 Chr.14: 50591509-50591509 on Build GRCh38
rs776796883 Chr.14: 50591643-50591643 on Build GRCh38
rs754547671 Chr.14: 50591511-50591511 on Build GRCh38
rs745384604 Chr.14: 50591678-50591678 on Build GRCh38
rs1040998169 Chr.14: 50591682-50591682 on Build GRCh38
rs984920474 Chr.14: 50591567-50591567 on Build GRCh38
rs766096780 Chr.14: 50591507-50591507 on Build GRCh38
rs532401914 Chr.14: 50591773-50591773 on Build GRCh38
rs753482250 Chr.14: 50591574-50591577 on Build GRCh38

View Full Product Details