Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 61043006-61043512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC38A6
Entrez Gene ID
145389
Gene Name
solute carrier family 38 member 6
Gene Aliases
NAT-1, SNAT6
UniGene
Hs.200738
Gene Chromosome Location
Chr.14: 60981114-61083733 on Build GRCh38
Transcripts
NM_001172702.1 NM_153811.2 NR_033344.1 XM_006720049.3 XM_006720050.1 XM_011536469.2 XM_017021020.1 XM_017021021.1 XM_017021022.1 XM_017021023.1 XM_017021024.1 XM_017021025.1 XM_017021026.1 XM_017021027.1 XM_017021028.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
398708 c.625-1G>T p.? Chr.14: 61043146-61043146 on Build GRCh38
5027871 c.649C>G p.P217A Chr.14: 61043171-61043171 on Build GRCh38
2247065 c.634delA p.K213fs*8 Chr.14: 61043156-61043156 on Build GRCh38
5037394 c.625-7T>A p.? Chr.14: 61043140-61043140 on Build GRCh38
398709 c.625-1G>T p.? Chr.14: 61043146-61043146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051562582 Chr.14: 61043061-61043061 on Build GRCh38
rs781756063 Chr.14: 61043403-61043403 on Build GRCh38
rs2296921 Chr.14: 61043206-61043206 on Build GRCh38
rs1053405457 Chr.14: 61043314-61043314 on Build GRCh38
rs978945417 Chr.14: 61043296-61043296 on Build GRCh38
rs888882596 Chr.14: 61043483-61043483 on Build GRCh38
rs746100108 Chr.14: 61043083-61043083 on Build GRCh38
rs773111678 Chr.14: 61043418-61043418 on Build GRCh38
rs951289478 Chr.14: 61043097-61043097 on Build GRCh38
rs975181007 Chr.14: 61043217-61043217 on Build GRCh38
rs373448076 Chr.14: 61043106-61043106 on Build GRCh38
rs893363846 Chr.14: 61043376-61043376 on Build GRCh38
rs763663785 Chr.14: 61043448-61043448 on Build GRCh38
rs374261108 Chr.14: 61043434-61043434 on Build GRCh38
rs1023080364 Chr.14: 61043216-61043216 on Build GRCh38
rs926084657 Chr.14: 61043328-61043328 on Build GRCh38
rs755613760 Chr.14: 61043238-61043238 on Build GRCh38
rs756792480 Chr.14: 61043252-61043252 on Build GRCh38
rs377689025 Chr.14: 61043225-61043225 on Build GRCh38
rs936309396 Chr.14: 61043287-61043287 on Build GRCh38
rs760469311 Chr.14: 61043438-61043438 on Build GRCh38
rs753540491 Chr.14: 61043248-61043248 on Build GRCh38
rs759292287 Chr.14: 61043429-61043429 on Build GRCh38
rs748306479 Chr.14: 61043414-61043414 on Build GRCh38
rs777446831 Chr.14: 61043137-61043137 on Build GRCh38
rs781779553 Chr.14: 61043472-61043472 on Build GRCh38
rs769906730 Chr.14: 61043415-61043415 on Build GRCh38
rs771022499 Chr.14: 61043421-61043421 on Build GRCh38
rs753245848 Chr.14: 61043456-61043456 on Build GRCh38
rs912380244 Chr.14: 61043147-61043147 on Build GRCh38
rs543874260 Chr.14: 61043194-61043194 on Build GRCh38
rs112008506 Chr.14: 61043275-61043275 on Build GRCh38
rs780222683 Chr.14: 61043161-61043161 on Build GRCh38
rs761963172 Chr.14: 61043195-61043195 on Build GRCh38
rs377644829 Chr.14: 61043469-61043469 on Build GRCh38
rs752241058 Chr.14: 61043230-61043230 on Build GRCh38
rs762716540 Chr.14: 61043210-61043210 on Build GRCh38
rs767416642 Chr.14: 61043229-61043229 on Build GRCh38
rs756955263 Chr.14: 61043457-61043457 on Build GRCh38
rs769244772 Chr.14: 61043190-61043190 on Build GRCh38
rs752011719 Chr.14: 61043466-61043466 on Build GRCh38
rs747521489 Chr.14: 61043172-61043172 on Build GRCh38
rs149677754 Chr.14: 61043036-61043036 on Build GRCh38
rs780498702 Chr.14: 61043259-61043259 on Build GRCh38
rs777942587 Chr.14: 61043485-61043485 on Build GRCh38
rs557433041 Chr.14: 61043198-61043198 on Build GRCh38
rs763710299 Chr.14: 61043239-61043239 on Build GRCh38
rs752167407 Chr.14: 61043108-61043108 on Build GRCh38
rs576243702 Chr.14: 61043150-61043150 on Build GRCh38
rs373814591 Chr.14: 61043480-61043480 on Build GRCh38
rs974489004 Chr.14: 61043048-61043048 on Build GRCh38
rs775390044 Chr.14: 61043436-61043436 on Build GRCh38
rs768794934 Chr.14: 61043480-61043481 on Build GRCh38
rs113118527 Chr.14: 61043458-61043458 on Build GRCh38
rs36052493 Chr.14: 61043300-61043300 on Build GRCh38
rs556321943 Chr.14: 61043090-61043090 on Build GRCh38
rs370897669 Chr.14: 61043304-61043305 on Build GRCh38
rs749532918 Chr.14: 61043420-61043420 on Build GRCh38
rs746282234 Chr.14: 61043146-61043146 on Build GRCh38
rs201429736 Chr.14: 61043423-61043423 on Build GRCh38
rs11845339 Chr.14: 61043290-61043290 on Build GRCh38
rs145440362 Chr.14: 61043478-61043478 on Build GRCh38
rs970114312 Chr.14: 61043285-61043285 on Build GRCh38
rs747235258 Chr.14: 61043463-61043464 on Build GRCh38
rs755592200 Chr.14: 61043264-61043264 on Build GRCh38
rs369427540 Chr.14: 61043470-61043470 on Build GRCh38
rs766211959 Chr.14: 61043214-61043214 on Build GRCh38
rs539837999 Chr.14: 61043348-61043348 on Build GRCh38
rs921018965 Chr.14: 61043277-61043277 on Build GRCh38
rs747329181 Chr.14: 61043263-61043263 on Build GRCh38
rs780414251 Chr.14: 61043211-61043225 on Build GRCh38
rs755808121 Chr.14: 61043125-61043125 on Build GRCh38
rs34502374 Chr.14: 61043406-61043407 on Build GRCh38
rs993339388 Chr.14: 61043166-61043166 on Build GRCh38
rs751291664 Chr.14: 61043218-61043218 on Build GRCh38
rs769950756 Chr.14: 61043203-61043203 on Build GRCh38

View Full Product Details