Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69776137-69776638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC10A1
Entrez Gene ID
6554
Gene Name
solute carrier family 10 member 1
Gene Aliases
NTCP
UniGene
Hs.952
Gene Chromosome Location
Chr.14: 69775835-69797289 on Build GRCh38
Transcripts
NM_003049.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4726344 c.1043C>T p.T348I Chr.14: 69776289-69776289 on Build GRCh38
1607895 c.1003_1006delGGAA p.G335fs*>14 Chr.14: 69776326-69776329 on Build GRCh38
3815191 c.957G>C p.M319I Chr.14: 69776375-69776375 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs945204730 Chr.14: 69776612-69776612 on Build GRCh38
rs542129513 Chr.14: 69776549-69776549 on Build GRCh38
rs780040660 Chr.14: 69776382-69776382 on Build GRCh38
rs985873481 Chr.14: 69776529-69776529 on Build GRCh38
rs768367416 Chr.14: 69776380-69776382 on Build GRCh38
rs935890624 Chr.14: 69776587-69776587 on Build GRCh38
rs567292118 Chr.14: 69776337-69776337 on Build GRCh38
rs199672095 Chr.14: 69776393-69776393 on Build GRCh38
rs764770919 Chr.14: 69776418-69776418 on Build GRCh38
rs1054165400 Chr.14: 69776427-69776427 on Build GRCh38
rs748529124 Chr.14: 69776406-69776406 on Build GRCh38
rs375849915 Chr.14: 69776308-69776308 on Build GRCh38
rs774562376 Chr.14: 69776273-69776273 on Build GRCh38
rs571658472 Chr.14: 69776350-69776350 on Build GRCh38
rs144422250 Chr.14: 69776389-69776389 on Build GRCh38
rs56903885 Chr.14: 69776372-69776372 on Build GRCh38
rs769036919 Chr.14: 69776328-69776328 on Build GRCh38
rs370054875 Chr.14: 69776261-69776261 on Build GRCh38
rs569872092 Chr.14: 69776420-69776420 on Build GRCh38
rs763799355 Chr.14: 69776421-69776421 on Build GRCh38
rs200363313 Chr.14: 69776298-69776298 on Build GRCh38
rs769854391 Chr.14: 69776306-69776306 on Build GRCh38
rs774906600 Chr.14: 69776330-69776330 on Build GRCh38
rs752424517 Chr.14: 69776359-69776359 on Build GRCh38
rs749506324 Chr.14: 69776383-69776383 on Build GRCh38
rs1015178146 Chr.14: 69776499-69776499 on Build GRCh38
rs567746743 Chr.14: 69776278-69776278 on Build GRCh38
rs964404564 Chr.14: 69776163-69776163 on Build GRCh38
rs762452049 Chr.14: 69776264-69776264 on Build GRCh38
rs941571854 Chr.14: 69776532-69776532 on Build GRCh38
rs745789831 Chr.14: 69776276-69776277 on Build GRCh38
rs539927384 Chr.14: 69776440-69776440 on Build GRCh38
rs764747297 Chr.14: 69776353-69776353 on Build GRCh38
rs776215501 Chr.14: 69776342-69776342 on Build GRCh38
rs781231841 Chr.14: 69776289-69776289 on Build GRCh38
rs202036783 Chr.14: 69776271-69776271 on Build GRCh38
rs1045159591 Chr.14: 69776464-69776464 on Build GRCh38
rs745890639 Chr.14: 69776376-69776376 on Build GRCh38
rs765967864 Chr.14: 69776266-69776266 on Build GRCh38
rs761803507 Chr.14: 69776349-69776349 on Build GRCh38
rs200864706 Chr.14: 69776415-69776415 on Build GRCh38
rs955569341 Chr.14: 69776498-69776498 on Build GRCh38
rs944531709 Chr.14: 69776381-69776381 on Build GRCh38
rs200122835 Chr.14: 69776224-69776224 on Build GRCh38
rs770109133 Chr.14: 69776250-69776250 on Build GRCh38
rs73285203 Chr.14: 69776479-69776479 on Build GRCh38
rs769420283 Chr.14: 69776428-69776429 on Build GRCh38
rs1017655827 Chr.14: 69776472-69776472 on Build GRCh38
rs201053766 Chr.14: 69776429-69776429 on Build GRCh38
rs775709718 Chr.14: 69776253-69776253 on Build GRCh38
rs948140381 Chr.14: 69776457-69776457 on Build GRCh38
rs776281571 Chr.14: 69776399-69776401 on Build GRCh38
rs1004115943 Chr.14: 69776493-69776493 on Build GRCh38
rs1027254550 Chr.14: 69776514-69776514 on Build GRCh38
rs762396962 Chr.14: 69776335-69776335 on Build GRCh38
rs994526709 Chr.14: 69776501-69776501 on Build GRCh38
rs898039002 Chr.14: 69776500-69776500 on Build GRCh38
rs746915522 Chr.14: 69776280-69776280 on Build GRCh38
rs774853385 Chr.14: 69776400-69776400 on Build GRCh38
rs924562734 Chr.14: 69776560-69776560 on Build GRCh38
rs573380132 Chr.14: 69776557-69776557 on Build GRCh38
rs770901300 Chr.14: 69776281-69776281 on Build GRCh38
rs753457466 Chr.14: 69776267-69776267 on Build GRCh38
rs756252909 Chr.14: 69776377-69776377 on Build GRCh38
rs768868006 Chr.14: 69776405-69776411 on Build GRCh38
rs746009702 Chr.14: 69776296-69776296 on Build GRCh38
rs1053876192 Chr.14: 69776193-69776193 on Build GRCh38
rs188012028 Chr.14: 69776279-69776279 on Build GRCh38
rs964339783 Chr.14: 69776476-69776476 on Build GRCh38
rs570636573 Chr.14: 69776594-69776594 on Build GRCh38
rs763393936 Chr.14: 69776255-69776255 on Build GRCh38
rs935323163 Chr.14: 69776191-69776191 on Build GRCh38
rs202206428 Chr.14: 69776220-69776220 on Build GRCh38
rs763852512 Chr.14: 69776369-69776369 on Build GRCh38
rs1018643840 Chr.14: 69776544-69776544 on Build GRCh38
rs763189956 Chr.14: 69776312-69776312 on Build GRCh38
rs200696891 Chr.14: 69776165-69776165 on Build GRCh38
rs759178817 Chr.14: 69776553-69776553 on Build GRCh38
rs373480905 Chr.14: 69776412-69776412 on Build GRCh38
rs201537069 Chr.14: 69776338-69776338 on Build GRCh38
rs149272163 Chr.14: 69776375-69776375 on Build GRCh38
rs975562624 Chr.14: 69776179-69776179 on Build GRCh38
rs372221415 Chr.14: 69776568-69776568 on Build GRCh38
rs201371965 Chr.14: 69776171-69776171 on Build GRCh38
rs893859505 Chr.14: 69776198-69776198 on Build GRCh38
rs758024722 Chr.14: 69776367-69776367 on Build GRCh38
rs201184437 Chr.14: 69776218-69776218 on Build GRCh38
rs549615149 Chr.14: 69776269-69776269 on Build GRCh38
rs752376832 Chr.14: 69776275-69776275 on Build GRCh38
rs113335157 Chr.14: 69776522-69776522 on Build GRCh38
rs772062475 Chr.14: 69776284-69776285 on Build GRCh38
rs937989913 Chr.14: 69776570-69776570 on Build GRCh38
rs776730041 Chr.14: 69776234-69776234 on Build GRCh38
rs778631398 Chr.14: 69776273-69776273 on Build GRCh38
rs201989635 Chr.14: 69776371-69776371 on Build GRCh38
rs200015023 Chr.14: 69776186-69776186 on Build GRCh38
rs759729646 Chr.14: 69776239-69776239 on Build GRCh38
rs953103964 Chr.14: 69776366-69776366 on Build GRCh38
rs531107214 Chr.14: 69776214-69776214 on Build GRCh38
rs902542590 Chr.14: 69776177-69776177 on Build GRCh38
rs779693088 Chr.14: 69776375-69776375 on Build GRCh38
rs774909893 Chr.14: 69776419-69776419 on Build GRCh38
rs139756142 Chr.14: 69776512-69776512 on Build GRCh38
rs867085434 Chr.14: 69776481-69776481 on Build GRCh38
rs372916955 Chr.14: 69776584-69776584 on Build GRCh38
rs201079588 Chr.14: 69776252-69776252 on Build GRCh38
rs199688922 Chr.14: 69776432-69776432 on Build GRCh38
rs746771881 Chr.14: 69776378-69776382 on Build GRCh38
rs1000554630 Chr.14: 69776443-69776443 on Build GRCh38
rs1003825308 Chr.14: 69776282-69776282 on Build GRCh38

View Full Product Details