Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69778249-69778759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC10A1
Entrez Gene ID
6554
Gene Name
solute carrier family 10 member 1
Gene Aliases
NTCP
UniGene
Hs.952
Gene Chromosome Location
Chr.14: 69775835-69797289 on Build GRCh38
Transcripts
NM_003049.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6338457 c.902T>G p.I301S Chr.14: 69778374-69778374 on Build GRCh38
433302 c.797G>A p.C266Y Chr.14: 69778479-69778479 on Build GRCh38
72653 c.782A>C p.Q261P Chr.14: 69778494-69778494 on Build GRCh38
6299057 c.819C>G p.A273A Chr.14: 69778457-69778457 on Build GRCh38
7188371 c.785delA p.N262fs*29 Chr.14: 69778491-69778491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028009431 Chr.14: 69778621-69778621 on Build GRCh38
rs199949242 Chr.14: 69778369-69778369 on Build GRCh38
rs201653371 Chr.14: 69778361-69778361 on Build GRCh38
rs1025755319 Chr.14: 69778732-69778732 on Build GRCh38
rs529819230 Chr.14: 69778537-69778537 on Build GRCh38
rs772762685 Chr.14: 69778479-69778479 on Build GRCh38
rs773680632 Chr.14: 69778316-69778316 on Build GRCh38
rs200475806 Chr.14: 69778564-69778564 on Build GRCh38
rs202082807 Chr.14: 69778374-69778374 on Build GRCh38
rs201592464 Chr.14: 69778553-69778553 on Build GRCh38
rs773488386 Chr.14: 69778331-69778331 on Build GRCh38
rs199613401 Chr.14: 69778536-69778536 on Build GRCh38
rs763520236 Chr.14: 69778399-69778399 on Build GRCh38
rs200859649 Chr.14: 69778433-69778433 on Build GRCh38
rs774740424 Chr.14: 69778448-69778449 on Build GRCh38
rs953575031 Chr.14: 69778628-69778628 on Build GRCh38
rs200344196 Chr.14: 69778579-69778579 on Build GRCh38
rs770673347 Chr.14: 69778527-69778527 on Build GRCh38
rs527571638 Chr.14: 69778700-69778700 on Build GRCh38
rs202169699 Chr.14: 69778498-69778498 on Build GRCh38
rs986367945 Chr.14: 69778490-69778490 on Build GRCh38
rs200739093 Chr.14: 69778452-69778452 on Build GRCh38
rs374736609 Chr.14: 69778368-69778368 on Build GRCh38
rs776615277 Chr.14: 69778486-69778486 on Build GRCh38
rs772595706 Chr.14: 69778318-69778318 on Build GRCh38
rs202023002 Chr.14: 69778390-69778390 on Build GRCh38
rs201929677 Chr.14: 69778461-69778461 on Build GRCh38
rs773902401 Chr.14: 69778403-69778403 on Build GRCh38
rs775642688 Chr.14: 69778496-69778496 on Build GRCh38
rs894537374 Chr.14: 69778709-69778709 on Build GRCh38
rs772927310 Chr.14: 69778312-69778312 on Build GRCh38
rs781519262 Chr.14: 69778603-69778603 on Build GRCh38
rs760401548 Chr.14: 69778430-69778430 on Build GRCh38
rs752881267 Chr.14: 69778442-69778442 on Build GRCh38
rs746619552 Chr.14: 69778526-69778526 on Build GRCh38
rs912149818 Chr.14: 69778655-69778655 on Build GRCh38
rs199689913 Chr.14: 69778422-69778422 on Build GRCh38
rs779531420 Chr.14: 69778639-69778640 on Build GRCh38
rs781425586 Chr.14: 69778333-69778333 on Build GRCh38
rs371490029 Chr.14: 69778345-69778345 on Build GRCh38
rs765329350 Chr.14: 69778495-69778495 on Build GRCh38
rs200505958 Chr.14: 69778377-69778377 on Build GRCh38
rs200498007 Chr.14: 69778539-69778539 on Build GRCh38
rs114386061 Chr.14: 69778613-69778613 on Build GRCh38
rs200685480 Chr.14: 69778314-69778314 on Build GRCh38
rs199566799 Chr.14: 69778557-69778557 on Build GRCh38
rs552554983 Chr.14: 69778711-69778714 on Build GRCh38
rs201290531 Chr.14: 69778406-69778406 on Build GRCh38
rs944919886 Chr.14: 69778661-69778661 on Build GRCh38
rs777859204 Chr.14: 69778444-69778444 on Build GRCh38
rs201128499 Chr.14: 69778371-69778371 on Build GRCh38
rs141269120 Chr.14: 69778522-69778522 on Build GRCh38
rs759902090 Chr.14: 69778483-69778484 on Build GRCh38
rs200849985 Chr.14: 69778468-69778468 on Build GRCh38
rs748474136 Chr.14: 69778315-69778315 on Build GRCh38
rs201266539 Chr.14: 69778483-69778483 on Build GRCh38
rs374103998 Chr.14: 69778332-69778332 on Build GRCh38
rs201076064 Chr.14: 69778384-69778384 on Build GRCh38
rs766291686 Chr.14: 69778431-69778431 on Build GRCh38
rs774552057 Chr.14: 69778572-69778572 on Build GRCh38
rs10559679 Chr.14: 69778562-69778563 on Build GRCh38
rs748761023 Chr.14: 69778393-69778393 on Build GRCh38
rs972336153 Chr.14: 69778677-69778677 on Build GRCh38
rs770345573 Chr.14: 69778453-69778453 on Build GRCh38
rs1001662940 Chr.14: 69778695-69778695 on Build GRCh38
rs147226818 Chr.14: 69778521-69778521 on Build GRCh38
rs747396551 Chr.14: 69778387-69778387 on Build GRCh38
rs915493778 Chr.14: 69778282-69778282 on Build GRCh38
rs780171633 Chr.14: 69778289-69778289 on Build GRCh38
rs201672997 Chr.14: 69778470-69778470 on Build GRCh38
rs56063215 Chr.14: 69778690-69778690 on Build GRCh38
rs201677740 Chr.14: 69778348-69778348 on Build GRCh38
rs200116485 Chr.14: 69778299-69778299 on Build GRCh38
rs749396382 Chr.14: 69778293-69778293 on Build GRCh38
rs757552873 Chr.14: 69778502-69778502 on Build GRCh38
rs201142538 Chr.14: 69778340-69778340 on Build GRCh38
rs559096531 Chr.14: 69778287-69778287 on Build GRCh38
rs200824416 Chr.14: 69778519-69778519 on Build GRCh38
rs199910121 Chr.14: 69778375-69778375 on Build GRCh38
rs750245386 Chr.14: 69778360-69778360 on Build GRCh38
rs758741220 Chr.14: 69778383-69778383 on Build GRCh38
rs199623470 Chr.14: 69778450-69778450 on Build GRCh38
rs866366803 Chr.14: 69778406-69778406 on Build GRCh38
rs750857596 Chr.14: 69778509-69778509 on Build GRCh38
rs565189992 Chr.14: 69778606-69778606 on Build GRCh38
rs200641056 Chr.14: 69778283-69778283 on Build GRCh38
rs372034615 Chr.14: 69778305-69778305 on Build GRCh38
rs773375062 Chr.14: 69778616-69778616 on Build GRCh38
rs767038940 Chr.14: 69778412-69778412 on Build GRCh38
rs376884319 Chr.14: 69778391-69778391 on Build GRCh38
rs1036462422 Chr.14: 69778319-69778319 on Build GRCh38
rs749671644 Chr.14: 69778721-69778721 on Build GRCh38
rs113282691 Chr.14: 69778698-69778698 on Build GRCh38
rs201049760 Chr.14: 69778529-69778529 on Build GRCh38
rs750418349 Chr.14: 69778378-69778378 on Build GRCh38
rs946139306 Chr.14: 69778643-69778643 on Build GRCh38
rs201200099 Chr.14: 69778357-69778357 on Build GRCh38
rs375377594 Chr.14: 69778641-69778641 on Build GRCh38
rs200070799 Chr.14: 69778351-69778351 on Build GRCh38
rs201618208 Chr.14: 69778291-69778291 on Build GRCh38
rs780554510 Chr.14: 69778515-69778515 on Build GRCh38
rs199604394 Chr.14: 69778407-69778407 on Build GRCh38
rs894337988 Chr.14: 69778549-69778549 on Build GRCh38
rs200425043 Chr.14: 69778501-69778501 on Build GRCh38
rs200093127 Chr.14: 69778464-69778464 on Build GRCh38
rs757581074 Chr.14: 69778447-69778447 on Build GRCh38
rs1013939071 Chr.14: 69778710-69778710 on Build GRCh38
rs761497776 Chr.14: 69778355-69778355 on Build GRCh38
rs759587022 Chr.14: 69778437-69778437 on Build GRCh38
rs1001818628 Chr.14: 69778463-69778463 on Build GRCh38
rs368086757 Chr.14: 69778512-69778512 on Build GRCh38
rs770599507 Chr.14: 69778484-69778484 on Build GRCh38
rs746779969 Chr.14: 69778482-69778482 on Build GRCh38
rs200184307 Chr.14: 69778408-69778408 on Build GRCh38
rs541801766 Chr.14: 69778503-69778503 on Build GRCh38
rs770267154 Chr.14: 69778633-69778633 on Build GRCh38
rs2296651 Chr.14: 69778476-69778476 on Build GRCh38
rs755882151 Chr.14: 69778436-69778436 on Build GRCh38
rs759531965 Chr.14: 69778489-69778489 on Build GRCh38
rs879828613 Chr.14: 69778723-69778723 on Build GRCh38
rs751889477 Chr.14: 69778445-69778445 on Build GRCh38
rs904729305 Chr.14: 69778691-69778691 on Build GRCh38
rs753750057 Chr.14: 69778435-69778435 on Build GRCh38
rs529840936 Chr.14: 69778731-69778731 on Build GRCh38
rs530495479 Chr.14: 69778583-69778584 on Build GRCh38
rs146982168 Chr.14: 69778701-69778701 on Build GRCh38
rs757387093 Chr.14: 69778575-69778575 on Build GRCh38
rs189313778 Chr.14: 69778399-69778399 on Build GRCh38
rs72547507 Chr.14: 69778440-69778440 on Build GRCh38
rs72547506 Chr.14: 69778336-69778336 on Build GRCh38
rs201089510 Chr.14: 69778414-69778414 on Build GRCh38
rs769794075 Chr.14: 69778546-69778546 on Build GRCh38

View Full Product Details