Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 76788349-76788853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGEL1
Entrez Gene ID
23357
Gene Name
angel homolog 1
Gene Aliases
Ccr4e, KIAA0759
UniGene
Hs.497448
Gene Chromosome Location
Chr.14: 76786473-76812947 on Build GRCh38
Transcripts
NM_015305.3 XM_017021115.1

SNP Detail

SNP ID Chromosome Location
rs936388290 Chr.14: 76788537-76788537 on Build GRCh38
rs549078315 Chr.14: 76788570-76788570 on Build GRCh38
rs975561494 Chr.14: 76788535-76788535 on Build GRCh38
rs968964121 Chr.14: 76788660-76788660 on Build GRCh38
rs545488533 Chr.14: 76788797-76788797 on Build GRCh38
rs887367281 Chr.14: 76788404-76788404 on Build GRCh38
rs534321945 Chr.14: 76788611-76788611 on Build GRCh38
rs1003088624 Chr.14: 76788405-76788405 on Build GRCh38
rs549743846 Chr.14: 76788460-76788460 on Build GRCh38
rs1012697704 Chr.14: 76788469-76788469 on Build GRCh38
rs373416074 Chr.14: 76788778-76788778 on Build GRCh38
rs937646615 Chr.14: 76788689-76788689 on Build GRCh38
rs968452850 Chr.14: 76788483-76788483 on Build GRCh38
rs189093672 Chr.14: 76788475-76788475 on Build GRCh38
rs931382627 Chr.14: 76788568-76788568 on Build GRCh38
rs910413820 Chr.14: 76788509-76788509 on Build GRCh38
rs919999021 Chr.14: 76788567-76788567 on Build GRCh38
rs571651007 Chr.14: 76788750-76788750 on Build GRCh38
rs886316743 Chr.14: 76788431-76788431 on Build GRCh38
rs765287886 Chr.14: 76788663-76788663 on Build GRCh38
rs2242624 Chr.14: 76788493-76788493 on Build GRCh38
rs938928810 Chr.14: 76788625-76788625 on Build GRCh38
rs117186683 Chr.14: 76788417-76788417 on Build GRCh38
rs76710953 Chr.14: 76788375-76788375 on Build GRCh38
rs924950158 Chr.14: 76788681-76788681 on Build GRCh38
rs1021774268 Chr.14: 76788482-76788482 on Build GRCh38
rs35761281 Chr.14: 76788570-76788571 on Build GRCh38
rs952927246 Chr.14: 76788800-76788800 on Build GRCh38
rs141168105 Chr.14: 76788790-76788790 on Build GRCh38
rs776494579 Chr.14: 76788498-76788498 on Build GRCh38
rs1030361961 Chr.14: 76788580-76788580 on Build GRCh38
rs535320224 Chr.14: 76788712-76788712 on Build GRCh38
rs537335713 Chr.14: 76788785-76788785 on Build GRCh38
rs564419846 Chr.14: 76788432-76788432 on Build GRCh38
rs998934736 Chr.14: 76788490-76788490 on Build GRCh38
rs761745080 Chr.14: 76788508-76788508 on Build GRCh38
rs949190977 Chr.14: 76788406-76788406 on Build GRCh38
rs987318707 Chr.14: 76788505-76788505 on Build GRCh38
rs954828418 Chr.14: 76788585-76788585 on Build GRCh38
rs565396947 Chr.14: 76788604-76788604 on Build GRCh38
rs1057383950 Chr.14: 76788655-76788655 on Build GRCh38
rs964880969 Chr.14: 76788512-76788512 on Build GRCh38
rs1031310480 Chr.14: 76788772-76788772 on Build GRCh38
rs973654751 Chr.14: 76788794-76788794 on Build GRCh38
rs765920145 Chr.14: 76788768-76788768 on Build GRCh38
rs1013499552 Chr.14: 76788690-76788690 on Build GRCh38
rs1020874862 Chr.14: 76788646-76788646 on Build GRCh38
rs553918921 Chr.14: 76788734-76788734 on Build GRCh38
rs557300890 Chr.14: 76788786-76788786 on Build GRCh38
rs1047345580 Chr.14: 76788393-76788393 on Build GRCh38
rs1027454064 Chr.14: 76788796-76788796 on Build GRCh38
rs908522729 Chr.14: 76788612-76788612 on Build GRCh38
rs1047559566 Chr.14: 76788774-76788774 on Build GRCh38
rs533255331 Chr.14: 76788454-76788454 on Build GRCh38

View Full Product Details